Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 21808173-21808728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF43
Entrez Gene ID
7594
Gene Name
zinc finger protein 43
Gene Aliases
HTF6, KOX27, ZNF39L1
UniGene
Hs.534365
Gene Chromosome Location
Chr.19: 21804946-21852095 on Build GRCh38
Transcripts
NM_001256648.1 NM_001256649.1 NM_001256650.1 NM_001256651.1 NM_001256653.1 NM_001256654.1 NM_003423.3 XM_011528257.2 XM_011528259.2 XM_017027207.1 XM_017027208.1 XM_017027209.1 XM_017027210.1 XM_017027211.1 XM_017027212.1 XM_017027213.1 XM_017027214.1 XM_017027215.1 XM_017027216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
993722 c.1849A>G p.T617A Chr.19: 21808188-21808188 on Build GRCh38
1271382 c.1640T>A p.F547Y Chr.19: 21808397-21808397 on Build GRCh38
5855241 c.1648T>A p.F550I Chr.19: 21808389-21808389 on Build GRCh38
7423818 c.1327A>T p.K443* Chr.19: 21808710-21808710 on Build GRCh38
1271380 c.1754A>C p.K585T Chr.19: 21808283-21808283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145677006 Chr.19: 21808383-21808383 on Build GRCh38
rs760218843 Chr.19: 21808260-21808260 on Build GRCh38
rs140619848 Chr.19: 21808342-21808342 on Build GRCh38
rs148377521 Chr.19: 21808674-21808674 on Build GRCh38
rs750307096 Chr.19: 21808428-21808428 on Build GRCh38
rs949997030 Chr.19: 21808465-21808465 on Build GRCh38
rs771438539 Chr.19: 21808355-21808355 on Build GRCh38
rs774564038 Chr.19: 21808213-21808213 on Build GRCh38
rs967654523 Chr.19: 21808309-21808309 on Build GRCh38
rs199705979 Chr.19: 21808553-21808553 on Build GRCh38
rs781049145 Chr.19: 21808308-21808308 on Build GRCh38
rs777338829 Chr.19: 21808274-21808274 on Build GRCh38
rs137982117 Chr.19: 21808257-21808257 on Build GRCh38
rs759113415 Chr.19: 21808629-21808634 on Build GRCh38
rs550420786 Chr.19: 21808207-21808207 on Build GRCh38
rs762589553 Chr.19: 21808386-21808386 on Build GRCh38
rs750362062 Chr.19: 21808317-21808317 on Build GRCh38
rs760588871 Chr.19: 21808595-21808595 on Build GRCh38
rs759540747 Chr.19: 21808692-21808692 on Build GRCh38
rs773431108 Chr.19: 21808262-21808262 on Build GRCh38
rs370200354 Chr.19: 21808586-21808586 on Build GRCh38
rs761166182 Chr.19: 21808269-21808269 on Build GRCh38
rs972045512 Chr.19: 21808624-21808624 on Build GRCh38
rs751224291 Chr.19: 21808391-21808391 on Build GRCh38
rs772381041 Chr.19: 21808199-21808203 on Build GRCh38
rs566321351 Chr.19: 21808554-21808554 on Build GRCh38
rs760153901 Chr.19: 21808382-21808382 on Build GRCh38
rs752718593 Chr.19: 21808449-21808449 on Build GRCh38
rs777213962 Chr.19: 21808370-21808370 on Build GRCh38
rs138296910 Chr.19: 21808578-21808578 on Build GRCh38
rs926974599 Chr.19: 21808340-21808340 on Build GRCh38
rs747717128 Chr.19: 21808336-21808336 on Build GRCh38
rs757099787 Chr.19: 21808306-21808306 on Build GRCh38
rs764083760 Chr.19: 21808361-21808361 on Build GRCh38
rs781605873 Chr.19: 21808646-21808646 on Build GRCh38
rs144229123 Chr.19: 21808658-21808658 on Build GRCh38
rs773824688 Chr.19: 21808673-21808673 on Build GRCh38
rs200283818 Chr.19: 21808622-21808622 on Build GRCh38
rs1045764458 Chr.19: 21808512-21808512 on Build GRCh38
rs548001388 Chr.19: 21808244-21808244 on Build GRCh38
rs751784442 Chr.19: 21808631-21808631 on Build GRCh38
rs149571562 Chr.19: 21808629-21808629 on Build GRCh38
rs776393899 Chr.19: 21808688-21808688 on Build GRCh38
rs758565363 Chr.19: 21808471-21808471 on Build GRCh38
rs770196661 Chr.19: 21808509-21808509 on Build GRCh38
rs771710969 Chr.19: 21808678-21808678 on Build GRCh38
rs781707064 Chr.19: 21808506-21808506 on Build GRCh38
rs950350179 Chr.19: 21808279-21808279 on Build GRCh38
rs756019136 Chr.19: 21808440-21808440 on Build GRCh38
rs984464377 Chr.19: 21808664-21808664 on Build GRCh38
rs759060782 Chr.19: 21808278-21808278 on Build GRCh38
rs780083833 Chr.19: 21808445-21808445 on Build GRCh38
rs373631790 Chr.19: 21808413-21808413 on Build GRCh38
rs764964833 Chr.19: 21808592-21808592 on Build GRCh38
rs533846548 Chr.19: 21808594-21808594 on Build GRCh38
rs866204061 Chr.19: 21808566-21808566 on Build GRCh38
rs751307204 Chr.19: 21808296-21808296 on Build GRCh38
rs776122213 Chr.19: 21808524-21808524 on Build GRCh38
rs756514645 Chr.19: 21808657-21808657 on Build GRCh38
rs558384217 Chr.19: 21808670-21808670 on Build GRCh38
rs753829937 Chr.19: 21808619-21808619 on Build GRCh38
rs752354982 Chr.19: 21808282-21808282 on Build GRCh38
rs766205499 Chr.19: 21808687-21808687 on Build GRCh38
rs778877896 Chr.19: 21808330-21808330 on Build GRCh38
rs761647152 Chr.19: 21808498-21808498 on Build GRCh38
rs777803776 Chr.19: 21808472-21808472 on Build GRCh38
rs757531400 Chr.19: 21808645-21808645 on Build GRCh38
rs879057783 Chr.19: 21808375-21808375 on Build GRCh38
rs778652111 Chr.19: 21808219-21808219 on Build GRCh38
rs897859037 Chr.19: 21808538-21808538 on Build GRCh38
rs780200433 Chr.19: 21808702-21808702 on Build GRCh38
rs769385499 Chr.19: 21808634-21808635 on Build GRCh38
rs772429186 Chr.19: 21808222-21808222 on Build GRCh38
rs761528488 Chr.19: 21808406-21808406 on Build GRCh38
rs773949501 Chr.19: 21808559-21808559 on Build GRCh38
rs748305049 Chr.19: 21808339-21808339 on Build GRCh38
rs554390607 Chr.19: 21808390-21808390 on Build GRCh38
rs746221385 Chr.19: 21808649-21808649 on Build GRCh38
rs191638211 Chr.19: 21808385-21808385 on Build GRCh38
rs747239613 Chr.19: 21808493-21808493 on Build GRCh38
rs544627657 Chr.19: 21808680-21808680 on Build GRCh38
rs929230243 Chr.19: 21808539-21808539 on Build GRCh38
rs748344967 Chr.19: 21808217-21808217 on Build GRCh38
rs746284893 Chr.19: 21808508-21808508 on Build GRCh38
rs766217155 Chr.19: 21808596-21808596 on Build GRCh38
rs767202213 Chr.19: 21808582-21808582 on Build GRCh38
rs755009019 Chr.19: 21808322-21808322 on Build GRCh38
rs763846945 Chr.19: 21808295-21808295 on Build GRCh38
rs1045783959 Chr.19: 21808294-21808294 on Build GRCh38
rs1006925244 Chr.19: 21808540-21808540 on Build GRCh38
rs778128366 Chr.19: 21808345-21808345 on Build GRCh38
rs760334254 Chr.19: 21808681-21808681 on Build GRCh38
rs764660079 Chr.19: 21808281-21808281 on Build GRCh38
rs776285885 Chr.19: 21808228-21808228 on Build GRCh38
rs1033089839 Chr.19: 21808560-21808560 on Build GRCh38
rs879152451 Chr.19: 21808289-21808289 on Build GRCh38
rs878968141 Chr.19: 21808259-21808259 on Build GRCh38
rs889805392 Chr.19: 21808502-21808502 on Build GRCh38
rs536331309 Chr.19: 21808496-21808496 on Build GRCh38
rs747368038 Chr.19: 21808354-21808354 on Build GRCh38
rs758010270 Chr.19: 21808286-21808286 on Build GRCh38
rs201252231 Chr.19: 21808660-21808660 on Build GRCh38
rs926130978 Chr.19: 21808371-21808371 on Build GRCh38

View Full Product Details