Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 22313851-22314415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF729
Entrez Gene ID
100287226
Gene Name
zinc finger protein 729
Gene Aliases
-
UniGene
Hs.531629
Gene Chromosome Location
Chr.19: 22286404-22317438 on Build GRCh38
Transcripts
NM_001242680.1 XM_017026122.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7403310 c.604C>A p.H202N Chr.19: 22314021-22314021 on Build GRCh38
4438735 c.544T>G p.C182G Chr.19: 22313961-22313961 on Build GRCh38
2747963 c.772C>T p.H258Y Chr.19: 22314189-22314189 on Build GRCh38
5724502 c.778G>A p.G260R Chr.19: 22314195-22314195 on Build GRCh38
5662510 c.965G>T p.C322F Chr.19: 22314382-22314382 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769027109 Chr.19: 22314320-22314321 on Build GRCh38
rs370660922 Chr.19: 22313969-22313969 on Build GRCh38
rs1004077855 Chr.19: 22314115-22314115 on Build GRCh38
rs370723012 Chr.19: 22314172-22314172 on Build GRCh38
rs766929482 Chr.19: 22314372-22314372 on Build GRCh38
rs772649799 Chr.19: 22313878-22313878 on Build GRCh38
rs762174779 Chr.19: 22314205-22314205 on Build GRCh38
rs764401998 Chr.19: 22314196-22314196 on Build GRCh38
rs750900578 Chr.19: 22314346-22314346 on Build GRCh38
rs879061878 Chr.19: 22314288-22314288 on Build GRCh38
rs538175767 Chr.19: 22314315-22314315 on Build GRCh38
rs747884789 Chr.19: 22314287-22314287 on Build GRCh38
rs570634417 Chr.19: 22314277-22314277 on Build GRCh38
rs201828154 Chr.19: 22314165-22314165 on Build GRCh38
rs763110611 Chr.19: 22314310-22314310 on Build GRCh38
rs766297588 Chr.19: 22314074-22314074 on Build GRCh38
rs763048640 Chr.19: 22314195-22314195 on Build GRCh38
rs762819302 Chr.19: 22313885-22313885 on Build GRCh38
rs754521076 Chr.19: 22314378-22314378 on Build GRCh38
rs556200784 Chr.19: 22314351-22314351 on Build GRCh38
rs768698249 Chr.19: 22314313-22314313 on Build GRCh38
rs754536532 Chr.19: 22314274-22314274 on Build GRCh38
rs184947462 Chr.19: 22314389-22314389 on Build GRCh38
rs767919901 Chr.19: 22314344-22314344 on Build GRCh38
rs773975059 Chr.19: 22314153-22314153 on Build GRCh38
rs532057714 Chr.19: 22314112-22314112 on Build GRCh38
rs778639544 Chr.19: 22314276-22314276 on Build GRCh38
rs537642567 Chr.19: 22313906-22313906 on Build GRCh38
rs749105225 Chr.19: 22313910-22313910 on Build GRCh38
rs866848128 Chr.19: 22314105-22314105 on Build GRCh38
rs542315410 Chr.19: 22313974-22313974 on Build GRCh38
rs546258167 Chr.19: 22314073-22314073 on Build GRCh38
rs45473510 Chr.19: 22314155-22314155 on Build GRCh38
rs776802851 Chr.19: 22313997-22313997 on Build GRCh38
rs780644444 Chr.19: 22313877-22313877 on Build GRCh38
rs752926644 Chr.19: 22314032-22314032 on Build GRCh38
rs914793302 Chr.19: 22314268-22314268 on Build GRCh38
rs777835783 Chr.19: 22314294-22314294 on Build GRCh38
rs200971874 Chr.19: 22314295-22314295 on Build GRCh38
rs748943577 Chr.19: 22314119-22314119 on Build GRCh38
rs200054460 Chr.19: 22314278-22314278 on Build GRCh38
rs550089222 Chr.19: 22314113-22314113 on Build GRCh38
rs946328541 Chr.19: 22314006-22314006 on Build GRCh38
rs753417867 Chr.19: 22314265-22314265 on Build GRCh38
rs577110860 Chr.19: 22313896-22313896 on Build GRCh38
rs768007988 Chr.19: 22314233-22314233 on Build GRCh38
rs867421154 Chr.19: 22313994-22313994 on Build GRCh38
rs762152432 Chr.19: 22314342-22314342 on Build GRCh38
rs572428489 Chr.19: 22314054-22314054 on Build GRCh38
rs768468876 Chr.19: 22314149-22314149 on Build GRCh38
rs772055493 Chr.19: 22314289-22314289 on Build GRCh38
rs954244206 Chr.19: 22314170-22314170 on Build GRCh38
rs774325488 Chr.19: 22313948-22313948 on Build GRCh38
rs538312515 Chr.19: 22314114-22314114 on Build GRCh38
rs556351936 Chr.19: 22313932-22313932 on Build GRCh38
rs772076103 Chr.19: 22314161-22314161 on Build GRCh38
rs779743802 Chr.19: 22314118-22314118 on Build GRCh38
rs545943741 Chr.19: 22314123-22314123 on Build GRCh38
rs765485088 Chr.19: 22314030-22314030 on Build GRCh38
rs376543990 Chr.19: 22314167-22314167 on Build GRCh38
rs552198706 Chr.19: 22314244-22314244 on Build GRCh38
rs868862586 Chr.19: 22314384-22314384 on Build GRCh38
rs759701802 Chr.19: 22314007-22314007 on Build GRCh38
rs867459870 Chr.19: 22314025-22314025 on Build GRCh38
rs113578175 Chr.19: 22314300-22314300 on Build GRCh38
rs564619363 Chr.19: 22314098-22314098 on Build GRCh38
rs879041439 Chr.19: 22314325-22314325 on Build GRCh38
rs751816512 Chr.19: 22314204-22314204 on Build GRCh38
rs912747228 Chr.19: 22313926-22313926 on Build GRCh38
rs558252488 Chr.19: 22314375-22314375 on Build GRCh38
rs913569121 Chr.19: 22313962-22313962 on Build GRCh38
rs767593021 Chr.19: 22313988-22313988 on Build GRCh38
rs745320904 Chr.19: 22314094-22314094 on Build GRCh38
rs149979717 Chr.19: 22314166-22314166 on Build GRCh38
rs779471922 Chr.19: 22314255-22314255 on Build GRCh38
rs191961918 Chr.19: 22313943-22313943 on Build GRCh38
rs1001979167 Chr.19: 22314055-22314055 on Build GRCh38
rs574638391 Chr.19: 22314363-22314363 on Build GRCh38
rs751987651 Chr.19: 22314058-22314058 on Build GRCh38
rs750987929 Chr.19: 22314241-22314241 on Build GRCh38
rs745408163 Chr.19: 22313883-22313883 on Build GRCh38
rs775010341 Chr.19: 22313901-22313901 on Build GRCh38
rs749766206 Chr.19: 22314232-22314232 on Build GRCh38
rs775386659 Chr.19: 22314302-22314302 on Build GRCh38
rs139546086 Chr.19: 22314046-22314046 on Build GRCh38

View Full Product Details