Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51338033-51338620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VSIG10L
Entrez Gene ID
147645
Gene Name
V-set and immunoglobulin domain containing 10 like
Gene Aliases
-
UniGene
Hs.293236
Gene Chromosome Location
Chr.19: 51331541-51342124 on Build GRCh38
Transcripts
NM_001163922.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7220544 c.1857C>T p.P619P Chr.19: 51338081-51338081 on Build GRCh38
1232419 c.1903C>T p.R635W Chr.19: 51338035-51338035 on Build GRCh38
5438343 c.1775G>A p.R592Q Chr.19: 51338163-51338163 on Build GRCh38
5005146 c.1872C>T p.G624G Chr.19: 51338066-51338066 on Build GRCh38
7257089 c.1886G>A p.R629Q Chr.19: 51338052-51338052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139916395 Chr.19: 51338081-51338081 on Build GRCh38
rs753258109 Chr.19: 51338184-51338184 on Build GRCh38
rs945227981 Chr.19: 51338348-51338348 on Build GRCh38
rs768946792 Chr.19: 51338221-51338221 on Build GRCh38
rs920518333 Chr.19: 51338195-51338195 on Build GRCh38
rs935572483 Chr.19: 51338591-51338591 on Build GRCh38
rs542795609 Chr.19: 51338375-51338378 on Build GRCh38
rs921006361 Chr.19: 51338521-51338521 on Build GRCh38
rs964818308 Chr.19: 51338499-51338499 on Build GRCh38
rs751537282 Chr.19: 51338145-51338145 on Build GRCh38
rs201414524 Chr.19: 51338237-51338237 on Build GRCh38
rs1002307809 Chr.19: 51338378-51338378 on Build GRCh38
rs192493912 Chr.19: 51338356-51338356 on Build GRCh38
rs879220442 Chr.19: 51338109-51338109 on Build GRCh38
rs532340677 Chr.19: 51338183-51338183 on Build GRCh38
rs772454929 Chr.19: 51338064-51338064 on Build GRCh38
rs777439173 Chr.19: 51338053-51338053 on Build GRCh38
rs57710066 Chr.19: 51338058-51338058 on Build GRCh38
rs975163813 Chr.19: 51338509-51338509 on Build GRCh38
rs536447218 Chr.19: 51338449-51338450 on Build GRCh38
rs919498085 Chr.19: 51338251-51338251 on Build GRCh38
rs1054139309 Chr.19: 51338327-51338327 on Build GRCh38
rs556379609 Chr.19: 51338092-51338092 on Build GRCh38
rs188080705 Chr.19: 51338282-51338282 on Build GRCh38
rs780654060 Chr.19: 51338080-51338080 on Build GRCh38
rs775895204 Chr.19: 51338275-51338275 on Build GRCh38
rs747587201 Chr.19: 51338210-51338210 on Build GRCh38
rs983393802 Chr.19: 51338373-51338373 on Build GRCh38
rs766202750 Chr.19: 51338119-51338119 on Build GRCh38
rs533862299 Chr.19: 51338358-51338358 on Build GRCh38
rs554860468 Chr.19: 51338311-51338311 on Build GRCh38
rs989154255 Chr.19: 51338543-51338543 on Build GRCh38
rs920940372 Chr.19: 51338318-51338318 on Build GRCh38
rs550886822 Chr.19: 51338213-51338213 on Build GRCh38
rs987590929 Chr.19: 51338266-51338266 on Build GRCh38
rs527748484 Chr.19: 51338139-51338139 on Build GRCh38
rs577495235 Chr.19: 51338059-51338059 on Build GRCh38
rs767423678 Chr.19: 51338149-51338149 on Build GRCh38
rs778055038 Chr.19: 51338066-51338066 on Build GRCh38
rs950974800 Chr.19: 51338467-51338467 on Build GRCh38
rs951954725 Chr.19: 51338364-51338364 on Build GRCh38
rs1053683700 Chr.19: 51338114-51338114 on Build GRCh38
rs551072962 Chr.19: 51338322-51338322 on Build GRCh38
rs1033740879 Chr.19: 51338382-51338382 on Build GRCh38
rs761894506 Chr.19: 51338575-51338575 on Build GRCh38
rs1046324195 Chr.19: 51338576-51338576 on Build GRCh38
rs200590126 Chr.19: 51338094-51338094 on Build GRCh38
rs560220952 Chr.19: 51338120-51338120 on Build GRCh38
rs763131064 Chr.19: 51338115-51338115 on Build GRCh38
rs1002255545 Chr.19: 51338176-51338176 on Build GRCh38
rs34770137 Chr.19: 51338285-51338286 on Build GRCh38
rs200219374 Chr.19: 51338205-51338205 on Build GRCh38
rs34380065 Chr.19: 51338163-51338163 on Build GRCh38
rs974679296 Chr.19: 51338165-51338165 on Build GRCh38
rs1008309416 Chr.19: 51338351-51338351 on Build GRCh38
rs892381804 Chr.19: 51338337-51338337 on Build GRCh38
rs558302131 Chr.19: 51338407-51338407 on Build GRCh38
rs936375624 Chr.19: 51338525-51338525 on Build GRCh38
rs201452734 Chr.19: 51338110-51338110 on Build GRCh38
rs1027015449 Chr.19: 51338447-51338447 on Build GRCh38
rs765286703 Chr.19: 51338601-51338601 on Build GRCh38
rs907593660 Chr.19: 51338250-51338250 on Build GRCh38
rs538744025 Chr.19: 51338274-51338274 on Build GRCh38
rs987681974 Chr.19: 51338136-51338136 on Build GRCh38
rs1019086678 Chr.19: 51338498-51338498 on Build GRCh38
rs569123009 Chr.19: 51338269-51338269 on Build GRCh38
rs936340412 Chr.19: 51338321-51338321 on Build GRCh38
rs920485575 Chr.19: 51338362-51338362 on Build GRCh38
rs1039403151 Chr.19: 51338298-51338298 on Build GRCh38
rs950987000 Chr.19: 51338264-51338264 on Build GRCh38
rs1009386805 Chr.19: 51338140-51338140 on Build GRCh38
rs376841016 Chr.19: 51338083-51338083 on Build GRCh38
rs1056211110 Chr.19: 51338504-51338504 on Build GRCh38
rs892325151 Chr.19: 51338121-51338121 on Build GRCh38
rs10424579 Chr.19: 51338488-51338488 on Build GRCh38
rs376814997 Chr.19: 51338427-51338427 on Build GRCh38
rs904086032 Chr.19: 51338587-51338587 on Build GRCh38
rs756842849 Chr.19: 51338199-51338199 on Build GRCh38
rs1018808934 Chr.19: 51338356-51338357 on Build GRCh38
rs143741110 Chr.19: 51338426-51338426 on Build GRCh38
rs913525708 Chr.19: 51338558-51338558 on Build GRCh38
rs756506584 Chr.19: 51338199-51338200 on Build GRCh38
rs899184926 Chr.19: 51338413-51338413 on Build GRCh38
rs201785306 Chr.19: 51338180-51338180 on Build GRCh38
rs759211948 Chr.19: 51338147-51338147 on Build GRCh38

View Full Product Details