Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19766614-19767169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX1
Entrez Gene ID
6899
Gene Name
T-box 1
Gene Aliases
CAFS, CATCH22, CTHM, DGCR, DGS, DORV, TBX1C, TGA, VCF, VCFS
UniGene
Hs.173984
Gene Chromosome Location
Chr.22: 19756703-19783593 on Build GRCh38
Transcripts
NM_005992.1 NM_080646.1 NM_080647.1 XM_006724312.2 XM_017028925.1 XM_017028926.1 XM_017028927.1 XM_017028928.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6019111 c.1307A>C p.Y436S Chr.22: 19766686-19766686 on Build GRCh38
6038524 c.1259C>T p.A420V Chr.22: 19766638-19766638 on Build GRCh38
7076012 c.1281C>T p.L427L Chr.22: 19766660-19766660 on Build GRCh38
5057677 c.1315C>T p.P439S Chr.22: 19766694-19766694 on Build GRCh38
6797858 c.1317C>T p.P439P Chr.22: 19766696-19766696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967630757 Chr.22: 19767137-19767137 on Build GRCh38
rs574947516 Chr.22: 19766666-19766666 on Build GRCh38
rs750574356 Chr.22: 19766837-19766837 on Build GRCh38
rs770702512 Chr.22: 19766798-19766798 on Build GRCh38
rs537729175 Chr.22: 19767131-19767131 on Build GRCh38
rs1050524818 Chr.22: 19766986-19766986 on Build GRCh38
rs748434309 Chr.22: 19766783-19766784 on Build GRCh38
rs751476947 Chr.22: 19766916-19766916 on Build GRCh38
rs749601692 Chr.22: 19766772-19766786 on Build GRCh38
rs1034090888 Chr.22: 19767105-19767105 on Build GRCh38
rs72646969 Chr.22: 19766985-19766985 on Build GRCh38
rs938102809 Chr.22: 19766872-19766872 on Build GRCh38
rs987951145 Chr.22: 19767055-19767055 on Build GRCh38
rs754416536 Chr.22: 19766693-19766693 on Build GRCh38
rs760098496 Chr.22: 19766695-19766695 on Build GRCh38
rs751740430 Chr.22: 19766700-19766700 on Build GRCh38
rs971086340 Chr.22: 19767078-19767078 on Build GRCh38
rs761590228 Chr.22: 19766679-19766679 on Build GRCh38
rs367711718 Chr.22: 19766732-19766732 on Build GRCh38
rs368172043 Chr.22: 19766876-19766876 on Build GRCh38
rs962345052 Chr.22: 19766664-19766664 on Build GRCh38
rs769522876 Chr.22: 19766658-19766658 on Build GRCh38
rs745479292 Chr.22: 19766865-19766865 on Build GRCh38
rs569976109 Chr.22: 19766653-19766653 on Build GRCh38
rs756769832 Chr.22: 19766860-19766860 on Build GRCh38
rs1029918703 Chr.22: 19766762-19766763 on Build GRCh38
rs968396290 Chr.22: 19767095-19767095 on Build GRCh38
rs528896157 Chr.22: 19766963-19766963 on Build GRCh38
rs958886232 Chr.22: 19767109-19767109 on Build GRCh38
rs373987708 Chr.22: 19766824-19766824 on Build GRCh38
rs770129913 Chr.22: 19766821-19766821 on Build GRCh38
rs754328637 Chr.22: 19766638-19766638 on Build GRCh38
rs962376130 Chr.22: 19766779-19766779 on Build GRCh38
rs779779535 Chr.22: 19766709-19766709 on Build GRCh38
rs775581777 Chr.22: 19766886-19766886 on Build GRCh38
rs551528783 Chr.22: 19767061-19767061 on Build GRCh38
rs1027757586 Chr.22: 19766950-19766950 on Build GRCh38
rs541198585 Chr.22: 19766826-19766826 on Build GRCh38
rs979441252 Chr.22: 19767101-19767101 on Build GRCh38
rs746699675 Chr.22: 19766794-19766794 on Build GRCh38
rs1037619193 Chr.22: 19767146-19767146 on Build GRCh38
rs781108085 Chr.22: 19766651-19766651 on Build GRCh38
rs764911027 Chr.22: 19766761-19766799 on Build GRCh38
rs1042125095 Chr.22: 19766957-19766957 on Build GRCh38
rs765173550 Chr.22: 19766802-19766804 on Build GRCh38
rs756826624 Chr.22: 19766763-19766801 on Build GRCh38
rs760188566 Chr.22: 19766750-19766750 on Build GRCh38
rs764132783 Chr.22: 19766827-19766827 on Build GRCh38
rs573980571 Chr.22: 19766696-19766696 on Build GRCh38
rs1047538218 Chr.22: 19767106-19767106 on Build GRCh38
rs764978950 Chr.22: 19766893-19766893 on Build GRCh38
rs768181576 Chr.22: 19766674-19766674 on Build GRCh38
rs780453322 Chr.22: 19766772-19766774 on Build GRCh38
rs1020839083 Chr.22: 19767086-19767086 on Build GRCh38
rs1044127370 Chr.22: 19766853-19766853 on Build GRCh38
rs780998431 Chr.22: 19766862-19766862 on Build GRCh38
rs375239385 Chr.22: 19766894-19766894 on Build GRCh38
rs774489872 Chr.22: 19766778-19766786 on Build GRCh38
rs777873225 Chr.22: 19766697-19766697 on Build GRCh38
rs774778199 Chr.22: 19766882-19766882 on Build GRCh38
rs1018657043 Chr.22: 19767006-19767006 on Build GRCh38
rs765827356 Chr.22: 19766751-19766751 on Build GRCh38
rs759569338 Chr.22: 19766760-19766768 on Build GRCh38
rs753192876 Chr.22: 19766753-19766753 on Build GRCh38
rs746335599 Chr.22: 19766766-19766795 on Build GRCh38
rs777103863 Chr.22: 19766744-19766744 on Build GRCh38
rs775903697 Chr.22: 19766874-19766874 on Build GRCh38
rs775895785 Chr.22: 19766825-19766825 on Build GRCh38
rs889645777 Chr.22: 19766991-19766991 on Build GRCh38
rs767638460 Chr.22: 19766735-19766736 on Build GRCh38
rs773442513 Chr.22: 19766889-19766889 on Build GRCh38
rs747623105 Chr.22: 19766726-19766726 on Build GRCh38
rs543378212 Chr.22: 19766880-19766880 on Build GRCh38
rs761822829 Chr.22: 19766736-19766738 on Build GRCh38
rs777313046 Chr.22: 19766792-19766792 on Build GRCh38
rs887219265 Chr.22: 19767113-19767113 on Build GRCh38
rs757452865 Chr.22: 19766773-19766773 on Build GRCh38
rs965925805 Chr.22: 19766808-19766808 on Build GRCh38
rs758517884 Chr.22: 19766649-19766649 on Build GRCh38
rs946257292 Chr.22: 19766953-19766953 on Build GRCh38
rs113153036 Chr.22: 19766909-19766909 on Build GRCh38
rs72646968 Chr.22: 19766780-19766780 on Build GRCh38
rs778718533 Chr.22: 19766781-19766781 on Build GRCh38
rs773384513 Chr.22: 19766786-19766787 on Build GRCh38
rs760927067 Chr.22: 19766890-19766890 on Build GRCh38
rs777213879 Chr.22: 19766852-19766852 on Build GRCh38
rs754795532 Chr.22: 19766840-19766840 on Build GRCh38
rs1005628844 Chr.22: 19767119-19767119 on Build GRCh38
rs779087360 Chr.22: 19766640-19766640 on Build GRCh38
rs1037247493 Chr.22: 19766906-19766906 on Build GRCh38
rs754705790 Chr.22: 19766777-19766777 on Build GRCh38
rs910653383 Chr.22: 19767148-19767148 on Build GRCh38
rs751755076 Chr.22: 19766768-19766768 on Build GRCh38
rs772030062 Chr.22: 19766784-19766792 on Build GRCh38
rs753713234 Chr.22: 19766736-19766741 on Build GRCh38
rs913808829 Chr.22: 19766838-19766838 on Build GRCh38
rs764197422 Chr.22: 19766754-19766754 on Build GRCh38
rs753613632 Chr.22: 19766776-19766776 on Build GRCh38
rs746505468 Chr.22: 19766859-19766859 on Build GRCh38
rs774386479 Chr.22: 19766832-19766832 on Build GRCh38
rs558979141 Chr.22: 19767019-19767019 on Build GRCh38
rs990431507 Chr.22: 19766922-19766922 on Build GRCh38
rs772661629 Chr.22: 19767129-19767129 on Build GRCh38
rs771540171 Chr.22: 19766729-19766729 on Build GRCh38
rs780815537 Chr.22: 19766763-19766780 on Build GRCh38
rs768572721 Chr.22: 19766723-19766723 on Build GRCh38
rs573376280 Chr.22: 19766765-19766765 on Build GRCh38
rs761882175 Chr.22: 19766883-19766883 on Build GRCh38
rs571783270 Chr.22: 19767070-19767070 on Build GRCh38
rs371856634 Chr.22: 19766888-19766888 on Build GRCh38
rs982028212 Chr.22: 19766708-19766708 on Build GRCh38
rs760701410 Chr.22: 19766792-19766793 on Build GRCh38
rs766206180 Chr.22: 19766775-19766775 on Build GRCh38
rs938071683 Chr.22: 19767120-19767120 on Build GRCh38
rs762076391 Chr.22: 19766836-19766836 on Build GRCh38
rs923299226 Chr.22: 19767143-19767144 on Build GRCh38
rs551038678 Chr.22: 19767145-19767145 on Build GRCh38
rs771606854 Chr.22: 19766680-19766680 on Build GRCh38
rs977663085 Chr.22: 19767142-19767142 on Build GRCh38
rs898430564 Chr.22: 19767033-19767033 on Build GRCh38
rs770080000 Chr.22: 19766871-19766871 on Build GRCh38
rs772860144 Chr.22: 19766681-19766681 on Build GRCh38
rs778539160 Chr.22: 19766724-19766724 on Build GRCh38
rs751264690 Chr.22: 19766763-19766774 on Build GRCh38
rs758290031 Chr.22: 19766761-19766769 on Build GRCh38
rs971560981 Chr.22: 19766817-19766819 on Build GRCh38
rs757867291 Chr.22: 19766785-19766785 on Build GRCh38
rs553461195 Chr.22: 19766762-19766762 on Build GRCh38
rs1003072874 Chr.22: 19767084-19767084 on Build GRCh38
rs563290139 Chr.22: 19766900-19766900 on Build GRCh38
rs769022249 Chr.22: 19766772-19766780 on Build GRCh38
rs901809779 Chr.22: 19766978-19766978 on Build GRCh38
rs528197589 Chr.22: 19767041-19767041 on Build GRCh38
rs992212631 Chr.22: 19767123-19767123 on Build GRCh38
rs926658302 Chr.22: 19767116-19767116 on Build GRCh38
rs953153020 Chr.22: 19766981-19766981 on Build GRCh38
rs771471094 Chr.22: 19766741-19766741 on Build GRCh38
rs965810583 Chr.22: 19767039-19767039 on Build GRCh38
rs201993443 Chr.22: 19766688-19766688 on Build GRCh38
rs752498057 Chr.22: 19766761-19766790 on Build GRCh38
rs773715388 Chr.22: 19766676-19766676 on Build GRCh38
rs749275474 Chr.22: 19766718-19766718 on Build GRCh38
rs371506174 Chr.22: 19766813-19766813 on Build GRCh38
rs41298842 Chr.22: 19767031-19767032 on Build GRCh38
rs946713267 Chr.22: 19766851-19766851 on Build GRCh38
rs752531044 Chr.22: 19766905-19766905 on Build GRCh38
rs756861421 Chr.22: 19766919-19766919 on Build GRCh38
rs972389680 Chr.22: 19766786-19766786 on Build GRCh38
rs748075946 Chr.22: 19766783-19766783 on Build GRCh38
rs762856074 Chr.22: 19766908-19766908 on Build GRCh38
rs918009540 Chr.22: 19767133-19767133 on Build GRCh38
rs774578030 Chr.22: 19766717-19766718 on Build GRCh38
rs373863680 Chr.22: 19766959-19766959 on Build GRCh38
rs564141057 Chr.22: 19766845-19766845 on Build GRCh38
rs914809647 Chr.22: 19766925-19766925 on Build GRCh38
rs926713371 Chr.22: 19766863-19766863 on Build GRCh38
rs777514486 Chr.22: 19766763-19766771 on Build GRCh38
rs758325399 Chr.22: 19766846-19766846 on Build GRCh38
rs749185545 Chr.22: 19766667-19766667 on Build GRCh38
rs755937050 Chr.22: 19766704-19766704 on Build GRCh38
rs371557427 Chr.22: 19766639-19766639 on Build GRCh38
rs756680878 Chr.22: 19766771-19766772 on Build GRCh38
rs753011413 Chr.22: 19766644-19766644 on Build GRCh38
rs746250147 Chr.22: 19766814-19766814 on Build GRCh38

View Full Product Details