Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 52593110-52593589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP46
Entrez Gene ID
64854
Gene Name
ubiquitin specific peptidase 46
Gene Aliases
-
UniGene
Hs.7966
Gene Chromosome Location
Chr.4: 52590960-52659335 on Build GRCh38
Transcripts
NM_001134223.1 NM_001286767.1 NM_001286768.1 NM_022832.3 XM_017008557.1

SNP Detail

SNP ID Chromosome Location
rs3733506 Chr.4: 52593454-52593454 on Build GRCh38
rs889587726 Chr.4: 52593189-52593189 on Build GRCh38
rs751338804 Chr.4: 52593165-52593165 on Build GRCh38
rs970893737 Chr.4: 52593362-52593362 on Build GRCh38
rs767378022 Chr.4: 52593369-52593369 on Build GRCh38
rs922805159 Chr.4: 52593485-52593485 on Build GRCh38
rs891482652 Chr.4: 52593210-52593210 on Build GRCh38
rs960104265 Chr.4: 52593328-52593328 on Build GRCh38
rs1011221766 Chr.4: 52593230-52593230 on Build GRCh38
rs1053083443 Chr.4: 52593207-52593207 on Build GRCh38
rs935444861 Chr.4: 52593390-52593390 on Build GRCh38
rs915283239 Chr.4: 52593404-52593404 on Build GRCh38
rs771339170 Chr.4: 52593153-52593153 on Build GRCh38
rs10034164 Chr.4: 52593565-52593565 on Build GRCh38
rs901337507 Chr.4: 52593177-52593177 on Build GRCh38
rs953074949 Chr.4: 52593296-52593296 on Build GRCh38
rs879182777 Chr.4: 52593332-52593332 on Build GRCh38
rs1019351433 Chr.4: 52593230-52593230 on Build GRCh38
rs115667430 Chr.4: 52593448-52593448 on Build GRCh38
rs935472006 Chr.4: 52593499-52593499 on Build GRCh38
rs150183803 Chr.4: 52593138-52593138 on Build GRCh38
rs186428903 Chr.4: 52593422-52593422 on Build GRCh38
rs567936762 Chr.4: 52593426-52593426 on Build GRCh38
rs138732602 Chr.4: 52593469-52593469 on Build GRCh38
rs762663732 Chr.4: 52593553-52593553 on Build GRCh38
rs999786722 Chr.4: 52593195-52593195 on Build GRCh38
rs1021279895 Chr.4: 52593239-52593239 on Build GRCh38
rs972683688 Chr.4: 52593329-52593329 on Build GRCh38
rs1035566888 Chr.4: 52593327-52593327 on Build GRCh38
rs77059667 Chr.4: 52593278-52593278 on Build GRCh38
rs180954976 Chr.4: 52593386-52593386 on Build GRCh38
rs374004581 Chr.4: 52593192-52593192 on Build GRCh38
rs1041250971 Chr.4: 52593171-52593171 on Build GRCh38
rs3733505 Chr.4: 52593525-52593525 on Build GRCh38
rs1040928478 Chr.4: 52593566-52593566 on Build GRCh38
rs17051581 Chr.4: 52593397-52593397 on Build GRCh38
rs534371132 Chr.4: 52593498-52593498 on Build GRCh38
rs961140948 Chr.4: 52593234-52593234 on Build GRCh38
rs952612717 Chr.4: 52593396-52593396 on Build GRCh38
rs545532200 Chr.4: 52593316-52593316 on Build GRCh38

View Full Product Details