Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 149724500-149725052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP43
Entrez Gene ID
348995
Gene Name
nucleoporin 43
Gene Aliases
bA350J20.1, p42
UniGene
Hs.510375
Gene Chromosome Location
Chr.6: 149724315-149746572 on Build GRCh38
Transcripts
NM_198887.2 NR_104456.1 XM_005266960.4 XM_005266961.3 XM_005266962.3 XM_011535799.2

SNP Detail

SNP ID Chromosome Location
rs1048113 Chr.6: 149724613-149724613 on Build GRCh38
rs767511255 Chr.6: 149724651-149724651 on Build GRCh38
rs981221785 Chr.6: 149724741-149724741 on Build GRCh38
rs369822792 Chr.6: 149724770-149724770 on Build GRCh38
rs187804112 Chr.6: 149724564-149724564 on Build GRCh38
rs1044985976 Chr.6: 149724675-149724675 on Build GRCh38
rs567686061 Chr.6: 149724936-149724936 on Build GRCh38
rs867663690 Chr.6: 149724736-149724736 on Build GRCh38
rs1055408405 Chr.6: 149724587-149724587 on Build GRCh38
rs1048137 Chr.6: 149724611-149724611 on Build GRCh38
rs11961809 Chr.6: 149724838-149724838 on Build GRCh38
rs1129806 Chr.6: 149724619-149724619 on Build GRCh38
rs1047975216 Chr.6: 149724841-149724841 on Build GRCh38
rs1028599110 Chr.6: 149724861-149724862 on Build GRCh38
rs1129805 Chr.6: 149724620-149724620 on Build GRCh38
rs533744793 Chr.6: 149724640-149724640 on Build GRCh38
rs759545326 Chr.6: 149724609-149724609 on Build GRCh38
rs527718149 Chr.6: 149725009-149725009 on Build GRCh38
rs930885495 Chr.6: 149724604-149724604 on Build GRCh38
rs183753778 Chr.6: 149724786-149724786 on Build GRCh38
rs1054873029 Chr.6: 149725002-149725002 on Build GRCh38
rs926984368 Chr.6: 149724540-149724540 on Build GRCh38
rs571982238 Chr.6: 149724934-149724934 on Build GRCh38
rs766465675 Chr.6: 149724610-149724610 on Build GRCh38
rs899392724 Chr.6: 149724593-149724593 on Build GRCh38
rs375620957 Chr.6: 149724605-149724605 on Build GRCh38
rs560246942 Chr.6: 149724965-149724965 on Build GRCh38
rs765730251 Chr.6: 149724709-149724709 on Build GRCh38
rs753232088 Chr.6: 149724726-149724726 on Build GRCh38
rs150832752 Chr.6: 149724798-149724798 on Build GRCh38
rs751288977 Chr.6: 149724943-149724943 on Build GRCh38
rs1048117 Chr.6: 149724592-149724592 on Build GRCh38
rs141467915 Chr.6: 149724524-149724524 on Build GRCh38
rs531074867 Chr.6: 149724809-149724809 on Build GRCh38
rs73779549 Chr.6: 149724586-149724586 on Build GRCh38
rs564817062 Chr.6: 149724885-149724885 on Build GRCh38
rs553459376 Chr.6: 149724672-149724672 on Build GRCh38
rs1010609047 Chr.6: 149725026-149725026 on Build GRCh38
rs780863863 Chr.6: 149725022-149725022 on Build GRCh38
rs188131224 Chr.6: 149724950-149724950 on Build GRCh38
rs908086401 Chr.6: 149724880-149724880 on Build GRCh38
rs1129798 Chr.6: 149724705-149724705 on Build GRCh38
rs961376015 Chr.6: 149724909-149724909 on Build GRCh38
rs192682722 Chr.6: 149724719-149724719 on Build GRCh38
rs565332615 Chr.6: 149724567-149724567 on Build GRCh38
rs776632093 Chr.6: 149724668-149724668 on Build GRCh38
rs1032991581 Chr.6: 149724703-149724703 on Build GRCh38
rs760090022 Chr.6: 149724677-149724677 on Build GRCh38
rs3177640 Chr.6: 149724682-149724682 on Build GRCh38
rs377396941 Chr.6: 149724777-149724777 on Build GRCh38
rs886572923 Chr.6: 149724634-149724634 on Build GRCh38
rs952819017 Chr.6: 149724879-149724879 on Build GRCh38
rs573690702 Chr.6: 149724744-149724744 on Build GRCh38
rs900541094 Chr.6: 149724676-149724676 on Build GRCh38
rs565374713 Chr.6: 149724539-149724539 on Build GRCh38
rs926371689 Chr.6: 149724707-149724707 on Build GRCh38
rs1129820 Chr.6: 149724660-149724660 on Build GRCh38
rs540637736 Chr.6: 149724959-149724959 on Build GRCh38
rs892146520 Chr.6: 149724750-149724751 on Build GRCh38
rs1020539241 Chr.6: 149724727-149724727 on Build GRCh38
rs1129792 Chr.6: 149724792-149724792 on Build GRCh38
rs1048114 Chr.6: 149724612-149724612 on Build GRCh38

View Full Product Details