Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 90160788-90161339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STEAP1
Entrez Gene ID
26872
Gene Name
six transmembrane epithelial antigen of the prostate 1
Gene Aliases
PRSS24, STEAP
UniGene
Hs.61635
Gene Chromosome Location
Chr.7: 90154375-90164827 on Build GRCh38
Transcripts
NM_012449.2 NR_110029.2

Gene Symbol
STEAP2-AS1
Entrez Gene ID
100874100
Gene Name
STEAP2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.573339
Gene Chromosome Location
Chr.7: 89882353-90211635 on Build GRCh38
Transcripts
NM_012449.2 NR_110029.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1092797 c.205C>A p.P69T Chr.7: 90160925-90160925 on Build GRCh38
5632447 c.250C>G p.L84V Chr.7: 90160970-90160970 on Build GRCh38
1092798 c.515C>T p.A172V Chr.7: 90161235-90161235 on Build GRCh38
5710733 c.316T>G p.F106V Chr.7: 90161036-90161036 on Build GRCh38
6007371 c.536G>A p.S179N Chr.7: 90161256-90161256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982170332 Chr.7: 90160905-90160905 on Build GRCh38
rs758441219 Chr.7: 90161098-90161098 on Build GRCh38
rs777722158 Chr.7: 90160826-90160826 on Build GRCh38
rs754108733 Chr.7: 90160993-90160993 on Build GRCh38
rs61737479 Chr.7: 90161001-90161001 on Build GRCh38
rs749388502 Chr.7: 90161164-90161164 on Build GRCh38
rs143969433 Chr.7: 90161277-90161277 on Build GRCh38
rs777630691 Chr.7: 90161132-90161132 on Build GRCh38
rs771964794 Chr.7: 90160831-90160831 on Build GRCh38
rs374381276 Chr.7: 90161140-90161140 on Build GRCh38
rs773941822 Chr.7: 90160970-90160970 on Build GRCh38
rs771726443 Chr.7: 90161145-90161145 on Build GRCh38
rs747961520 Chr.7: 90160943-90160943 on Build GRCh38
rs762142125 Chr.7: 90161209-90161209 on Build GRCh38
rs374442133 Chr.7: 90160907-90160907 on Build GRCh38
rs200418160 Chr.7: 90160863-90160863 on Build GRCh38
rs776038298 Chr.7: 90161045-90161045 on Build GRCh38
rs71557077 Chr.7: 90160910-90160910 on Build GRCh38
rs751164868 Chr.7: 90160940-90160942 on Build GRCh38
rs776059791 Chr.7: 90160986-90160986 on Build GRCh38
rs772759291 Chr.7: 90161040-90161040 on Build GRCh38
rs762159939 Chr.7: 90161081-90161081 on Build GRCh38
rs779678323 Chr.7: 90161276-90161276 on Build GRCh38
rs532811070 Chr.7: 90161022-90161022 on Build GRCh38
rs773454536 Chr.7: 90161237-90161237 on Build GRCh38
rs149449071 Chr.7: 90161246-90161246 on Build GRCh38
rs761081630 Chr.7: 90160988-90160988 on Build GRCh38
rs137931598 Chr.7: 90161133-90161133 on Build GRCh38
rs375618251 Chr.7: 90160862-90160862 on Build GRCh38
rs748985562 Chr.7: 90161024-90161024 on Build GRCh38
rs768459695 Chr.7: 90160844-90160844 on Build GRCh38
rs371063299 Chr.7: 90160830-90160830 on Build GRCh38
rs368900941 Chr.7: 90160816-90160816 on Build GRCh38
rs760074090 Chr.7: 90160872-90160872 on Build GRCh38
rs770589860 Chr.7: 90160967-90160967 on Build GRCh38
rs201507772 Chr.7: 90161306-90161306 on Build GRCh38
rs201266997 Chr.7: 90161025-90161025 on Build GRCh38
rs781085814 Chr.7: 90161017-90161017 on Build GRCh38
rs565591003 Chr.7: 90160931-90160931 on Build GRCh38
rs750961483 Chr.7: 90161057-90161058 on Build GRCh38
rs4015376 Chr.7: 90160958-90160958 on Build GRCh38
rs967102323 Chr.7: 90160894-90160894 on Build GRCh38
rs376248650 Chr.7: 90161135-90161135 on Build GRCh38
rs761034705 Chr.7: 90161050-90161050 on Build GRCh38
rs11538915 Chr.7: 90160837-90160837 on Build GRCh38
rs765431839 Chr.7: 90161088-90161088 on Build GRCh38
rs71557078 Chr.7: 90161113-90161113 on Build GRCh38
rs772656393 Chr.7: 90160860-90160860 on Build GRCh38
rs757361648 Chr.7: 90160997-90160997 on Build GRCh38
rs755363804 Chr.7: 90160815-90160815 on Build GRCh38
rs769037451 Chr.7: 90161180-90161180 on Build GRCh38
rs766494139 Chr.7: 90161247-90161247 on Build GRCh38
rs780653386 Chr.7: 90161119-90161133 on Build GRCh38
rs762473739 Chr.7: 90160965-90160965 on Build GRCh38
rs757090228 Chr.7: 90161267-90161267 on Build GRCh38
rs778046788 Chr.7: 90160882-90160882 on Build GRCh38
rs778690758 Chr.7: 90161139-90161139 on Build GRCh38
rs778815521 Chr.7: 90161269-90161269 on Build GRCh38
rs553569756 Chr.7: 90160841-90160841 on Build GRCh38
rs928554723 Chr.7: 90160915-90160915 on Build GRCh38
rs1003916381 Chr.7: 90161289-90161289 on Build GRCh38
rs746959889 Chr.7: 90160919-90160919 on Build GRCh38
rs765303046 Chr.7: 90160999-90160999 on Build GRCh38
rs61737480 Chr.7: 90161274-90161274 on Build GRCh38
rs868419381 Chr.7: 90161290-90161290 on Build GRCh38
rs375178272 Chr.7: 90161293-90161293 on Build GRCh38
rs776350036 Chr.7: 90160846-90160846 on Build GRCh38
rs750496522 Chr.7: 90161006-90161006 on Build GRCh38
rs148631795 Chr.7: 90161283-90161283 on Build GRCh38
rs756634962 Chr.7: 90161059-90161060 on Build GRCh38
rs755948533 Chr.7: 90161126-90161126 on Build GRCh38
rs2888782 Chr.7: 90161227-90161227 on Build GRCh38
rs747552710 Chr.7: 90161179-90161179 on Build GRCh38
rs750109299 Chr.7: 90161270-90161270 on Build GRCh38
rs138781089 Chr.7: 90160877-90160877 on Build GRCh38
rs769480944 Chr.7: 90160853-90160853 on Build GRCh38
rs371335343 Chr.7: 90160984-90160984 on Build GRCh38
rs761458488 Chr.7: 90160847-90160847 on Build GRCh38
rs4015375 Chr.7: 90160861-90160861 on Build GRCh38
rs368652491 Chr.7: 90160873-90160873 on Build GRCh38
rs192792079 Chr.7: 90160878-90160878 on Build GRCh38
rs745360602 Chr.7: 90161034-90161034 on Build GRCh38
rs146259721 Chr.7: 90161059-90161059 on Build GRCh38
rs770042356 Chr.7: 90161214-90161214 on Build GRCh38
rs61737476 Chr.7: 90161078-90161078 on Build GRCh38
rs199539511 Chr.7: 90161285-90161285 on Build GRCh38
rs781591726 Chr.7: 90161032-90161033 on Build GRCh38
rs766406635 Chr.7: 90161111-90161111 on Build GRCh38
rs753718544 Chr.7: 90161262-90161262 on Build GRCh38
rs4015377 Chr.7: 90161141-90161141 on Build GRCh38
rs760734771 Chr.7: 90161256-90161256 on Build GRCh38
rs200282338 Chr.7: 90161261-90161261 on Build GRCh38
rs3953584 Chr.7: 90160963-90160963 on Build GRCh38
rs778910446 Chr.7: 90160916-90160916 on Build GRCh38
rs774074314 Chr.7: 90161027-90161027 on Build GRCh38
rs369205794 Chr.7: 90161250-90161250 on Build GRCh38
rs753918145 Chr.7: 90160886-90160886 on Build GRCh38
rs756305874 Chr.7: 90160824-90160824 on Build GRCh38
rs774801733 Chr.7: 90161146-90161146 on Build GRCh38
rs777110316 Chr.7: 90161200-90161200 on Build GRCh38
rs781030625 Chr.7: 90160936-90160936 on Build GRCh38
rs779932615 Chr.7: 90160838-90160838 on Build GRCh38
rs750549921 Chr.7: 90161092-90161092 on Build GRCh38
rs757611958 Chr.7: 90160975-90160975 on Build GRCh38
rs760127479 Chr.7: 90160978-90160978 on Build GRCh38
rs754964368 Chr.7: 90161009-90161009 on Build GRCh38
rs367749466 Chr.7: 90160953-90160953 on Build GRCh38
rs751527919 Chr.7: 90161255-90161255 on Build GRCh38

View Full Product Details