Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133308480-133309024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBGCP2
Entrez Gene ID
10844
Gene Name
tubulin gamma complex associated protein 2
Gene Aliases
GCP-2, GCP2, Grip103, SPBC97, Spc97p, h103p, hGCP2, hSpc97
UniGene
Hs.523370
Gene Chromosome Location
Chr.10: 133278630-133312337 on Build GRCh38
Transcripts
NM_001256617.1 NM_001256618.1 NM_006659.3 NR_046330.1 NM_145806.3 NR_130127.1

Gene Symbol
ZNF511
Entrez Gene ID
118472
Gene Name
zinc finger protein 511
Gene Aliases
-
UniGene
Hs.422113
Gene Chromosome Location
Chr.10: 133308889-133313162 on Build GRCh38
Transcripts
NM_001256617.1 NM_001256618.1 NM_006659.3 NR_046330.1 NM_145806.3 NR_130127.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3751664 c.60G>A p.P20P Chr.10: 133309003-133309003 on Build GRCh38
3751665 c.60G>A p.P20P Chr.10: 133309003-133309003 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs918632811 Chr.10: 133308560-133308560 on Build GRCh38
rs1003999859 Chr.10: 133308678-133308679 on Build GRCh38
rs370707138 Chr.10: 133308867-133308867 on Build GRCh38
rs560087256 Chr.10: 133308715-133308715 on Build GRCh38
rs1014665469 Chr.10: 133308909-133308909 on Build GRCh38
rs1018848078 Chr.10: 133308525-133308525 on Build GRCh38
rs562838613 Chr.10: 133308739-133308739 on Build GRCh38
rs76864287 Chr.10: 133308583-133308583 on Build GRCh38
rs1032899660 Chr.10: 133308505-133308505 on Build GRCh38
rs1038456044 Chr.10: 133308954-133308954 on Build GRCh38
rs906993975 Chr.10: 133308846-133308846 on Build GRCh38
rs973372967 Chr.10: 133308614-133308614 on Build GRCh38
rs977112240 Chr.10: 133308815-133308815 on Build GRCh38
rs780810429 Chr.10: 133308997-133308997 on Build GRCh38
rs3008356 Chr.10: 133308733-133308733 on Build GRCh38
rs942857664 Chr.10: 133308596-133308596 on Build GRCh38
rs1047623525 Chr.10: 133308953-133308953 on Build GRCh38
rs975599096 Chr.10: 133308769-133308769 on Build GRCh38
rs946802030 Chr.10: 133308968-133308968 on Build GRCh38
rs935788328 Chr.10: 133308913-133308913 on Build GRCh38
rs574994706 Chr.10: 133308536-133308536 on Build GRCh38
rs540540558 Chr.10: 133308647-133308647 on Build GRCh38
rs981944305 Chr.10: 133308824-133308824 on Build GRCh38
rs900053060 Chr.10: 133308862-133308862 on Build GRCh38
rs998328755 Chr.10: 133308724-133308724 on Build GRCh38
rs865926353 Chr.10: 133308910-133308910 on Build GRCh38
rs956567043 Chr.10: 133308953-133308953 on Build GRCh38
rs955705091 Chr.10: 133308902-133308902 on Build GRCh38
rs983048186 Chr.10: 133308593-133308593 on Build GRCh38
rs947950940 Chr.10: 133308634-133308634 on Build GRCh38
rs935391835 Chr.10: 133308827-133308827 on Build GRCh38
rs971250349 Chr.10: 133308537-133308537 on Build GRCh38
rs1016979551 Chr.10: 133308865-133308865 on Build GRCh38
rs555150577 Chr.10: 133308507-133308507 on Build GRCh38
rs902822939 Chr.10: 133308633-133308633 on Build GRCh38
rs915243650 Chr.10: 133308853-133308853 on Build GRCh38
rs984336874 Chr.10: 133308581-133308581 on Build GRCh38
rs547691200 Chr.10: 133308863-133308863 on Build GRCh38
rs1004580983 Chr.10: 133308879-133308879 on Build GRCh38
rs1013184718 Chr.10: 133308521-133308521 on Build GRCh38
rs905067711 Chr.10: 133308896-133308907 on Build GRCh38
rs1029996462 Chr.10: 133308571-133308571 on Build GRCh38
rs955678638 Chr.10: 133308957-133308957 on Build GRCh38
rs948686910 Chr.10: 133308841-133308841 on Build GRCh38
rs561933203 Chr.10: 133308790-133308790 on Build GRCh38
rs767967051 Chr.10: 133308796-133308796 on Build GRCh38
rs916467777 Chr.10: 133308806-133308806 on Build GRCh38
rs1018569933 Chr.10: 133308682-133308682 on Build GRCh38
rs532494020 Chr.10: 133308630-133308630 on Build GRCh38
rs1030310229 Chr.10: 133308956-133308956 on Build GRCh38
rs964280043 Chr.10: 133308546-133308546 on Build GRCh38
rs982056716 Chr.10: 133308936-133308936 on Build GRCh38
rs1040116342 Chr.10: 133308858-133308858 on Build GRCh38
rs778325014 Chr.10: 133308988-133308988 on Build GRCh38
rs987976908 Chr.10: 133308955-133308955 on Build GRCh38
rs570792225 Chr.10: 133308875-133308875 on Build GRCh38
rs969724432 Chr.10: 133308814-133308814 on Build GRCh38
rs1017177874 Chr.10: 133308755-133308756 on Build GRCh38
rs927776215 Chr.10: 133308952-133308952 on Build GRCh38
rs894386217 Chr.10: 133308838-133308838 on Build GRCh38
rs752943294 Chr.10: 133308962-133308962 on Build GRCh38
rs527554063 Chr.10: 133308847-133308847 on Build GRCh38
rs927805702 Chr.10: 133308826-133308826 on Build GRCh38
rs1041229069 Chr.10: 133308540-133308540 on Build GRCh38
rs974897989 Chr.10: 133308548-133308548 on Build GRCh38
rs948423707 Chr.10: 133308934-133308934 on Build GRCh38
rs964673209 Chr.10: 133308757-133308757 on Build GRCh38
rs569605747 Chr.10: 133308984-133308984 on Build GRCh38
rs1052419574 Chr.10: 133308666-133308666 on Build GRCh38
rs973725421 Chr.10: 133308797-133308797 on Build GRCh38
rs969314211 Chr.10: 133308926-133308926 on Build GRCh38
rs772761864 Chr.10: 133308993-133308993 on Build GRCh38
rs941934232 Chr.10: 133308606-133308606 on Build GRCh38
rs539433068 Chr.10: 133308924-133308924 on Build GRCh38
rs946701735 Chr.10: 133308856-133308856 on Build GRCh38
rs750962236 Chr.10: 133308861-133308861 on Build GRCh38
rs1023741116 Chr.10: 133308925-133308925 on Build GRCh38
rs989826410 Chr.10: 133308983-133308983 on Build GRCh38
rs1014529913 Chr.10: 133308763-133308763 on Build GRCh38
rs549797543 Chr.10: 133308971-133308971 on Build GRCh38
rs916390995 Chr.10: 133308616-133308616 on Build GRCh38
rs958504272 Chr.10: 133308517-133308517 on Build GRCh38
rs746642397 Chr.10: 133308994-133308994 on Build GRCh38
rs548037467 Chr.10: 133308785-133308785 on Build GRCh38
rs531835310 Chr.10: 133308784-133308784 on Build GRCh38
rs1041361672 Chr.10: 133308620-133308620 on Build GRCh38
rs1046668606 Chr.10: 133308641-133308641 on Build GRCh38
rs935421173 Chr.10: 133308650-133308650 on Build GRCh38
rs3008357 Chr.10: 133309003-133309003 on Build GRCh38
rs893843076 Chr.10: 133308673-133308673 on Build GRCh38
rs907133614 Chr.10: 133308940-133308940 on Build GRCh38
rs2995336 Chr.10: 133308727-133308727 on Build GRCh38
rs935652407 Chr.10: 133308825-133308825 on Build GRCh38
rs1045654966 Chr.10: 133308845-133308845 on Build GRCh38
rs758788592 Chr.10: 133308982-133308982 on Build GRCh38
rs922800627 Chr.10: 133308790-133308791 on Build GRCh38
rs1011568032 Chr.10: 133308674-133308674 on Build GRCh38
rs1004612104 Chr.10: 133308755-133308755 on Build GRCh38
rs770733163 Chr.10: 133308996-133308996 on Build GRCh38
rs954467511 Chr.10: 133308592-133308592 on Build GRCh38
rs910147970 Chr.10: 133308595-133308595 on Build GRCh38
rs1054277413 Chr.10: 133308646-133308646 on Build GRCh38
rs907563261 Chr.10: 133308602-133308602 on Build GRCh38
rs900084672 Chr.10: 133308708-133308708 on Build GRCh38
rs894421688 Chr.10: 133308648-133308648 on Build GRCh38

View Full Product Details