Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6002759-6003160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR56A4
Entrez Gene ID
120793
Gene Name
olfactory receptor family 56 subfamily A member 4
Gene Aliases
OR11-49
UniGene
Hs.553571
Gene Chromosome Location
Chr.11: 6002051-6003148 on Build GRCh38
Transcripts
NM_001005179.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
929250 c.278C>A p.A93D Chr.11: 6002871-6002871 on Build GRCh38
3450494 c.378C>T p.I126I Chr.11: 6002771-6002771 on Build GRCh38
6518612 c.175G>T p.D59Y Chr.11: 6002974-6002974 on Build GRCh38
4433780 c.6G>A p.M2I Chr.11: 6003143-6003143 on Build GRCh38
1703487 c.284G>A p.G95E Chr.11: 6002865-6002865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980038400 Chr.11: 6002876-6002876 on Build GRCh38
rs376077628 Chr.11: 6002850-6002850 on Build GRCh38
rs779572872 Chr.11: 6002975-6002975 on Build GRCh38
rs201265542 Chr.11: 6003045-6003045 on Build GRCh38
rs764324245 Chr.11: 6003124-6003124 on Build GRCh38
rs200362281 Chr.11: 6002896-6002896 on Build GRCh38
rs759582750 Chr.11: 6003020-6003020 on Build GRCh38
rs181754672 Chr.11: 6003077-6003077 on Build GRCh38
rs748520158 Chr.11: 6002796-6002796 on Build GRCh38
rs755325772 Chr.11: 6002786-6002786 on Build GRCh38
rs762673544 Chr.11: 6003010-6003010 on Build GRCh38
rs771597917 Chr.11: 6003072-6003073 on Build GRCh38
rs775774883 Chr.11: 6003075-6003076 on Build GRCh38
rs761472498 Chr.11: 6002921-6002921 on Build GRCh38
rs777318512 Chr.11: 6003074-6003075 on Build GRCh38
rs770754028 Chr.11: 6003082-6003082 on Build GRCh38
rs774478006 Chr.11: 6002792-6002792 on Build GRCh38
rs765033799 Chr.11: 6002848-6002848 on Build GRCh38
rs761678088 Chr.11: 6002830-6002830 on Build GRCh38
rs114240585 Chr.11: 6003083-6003083 on Build GRCh38
rs778256994 Chr.11: 6002968-6002968 on Build GRCh38
rs751658027 Chr.11: 6003127-6003127 on Build GRCh38
rs776746926 Chr.11: 6003125-6003125 on Build GRCh38
rs1020102370 Chr.11: 6003086-6003086 on Build GRCh38
rs368657390 Chr.11: 6003058-6003058 on Build GRCh38
rs767686748 Chr.11: 6002962-6002962 on Build GRCh38
rs370692842 Chr.11: 6002892-6002892 on Build GRCh38
rs201699749 Chr.11: 6002905-6002905 on Build GRCh38
rs200091716 Chr.11: 6003119-6003119 on Build GRCh38
rs751780544 Chr.11: 6002859-6002859 on Build GRCh38
rs773013591 Chr.11: 6002928-6002928 on Build GRCh38
rs768148185 Chr.11: 6002953-6002953 on Build GRCh38
rs891646227 Chr.11: 6002969-6002969 on Build GRCh38
rs767635248 Chr.11: 6003024-6003024 on Build GRCh38
rs773791496 Chr.11: 6002807-6002807 on Build GRCh38
rs754295492 Chr.11: 6002973-6002973 on Build GRCh38
rs768362201 Chr.11: 6002912-6002912 on Build GRCh38
rs368204471 Chr.11: 6002779-6002779 on Build GRCh38
rs973503489 Chr.11: 6002808-6002808 on Build GRCh38
rs756413126 Chr.11: 6002964-6002964 on Build GRCh38
rs374820484 Chr.11: 6003037-6003037 on Build GRCh38
rs765795064 Chr.11: 6003059-6003059 on Build GRCh38
rs369835055 Chr.11: 6003121-6003121 on Build GRCh38
rs202064495 Chr.11: 6002780-6002780 on Build GRCh38
rs772132002 Chr.11: 6003111-6003111 on Build GRCh38
rs544610117 Chr.11: 6002918-6002918 on Build GRCh38
rs115418679 Chr.11: 6002875-6002875 on Build GRCh38
rs143488149 Chr.11: 6002801-6002801 on Build GRCh38
rs745865391 Chr.11: 6003091-6003091 on Build GRCh38
rs750402228 Chr.11: 6002854-6002854 on Build GRCh38
rs768310858 Chr.11: 6002982-6002982 on Build GRCh38
rs745332709 Chr.11: 6002812-6002812 on Build GRCh38
rs189213766 Chr.11: 6002932-6002932 on Build GRCh38
rs116472611 Chr.11: 6003078-6003078 on Build GRCh38
rs372111623 Chr.11: 6002809-6002809 on Build GRCh38
rs769668588 Chr.11: 6002927-6002927 on Build GRCh38
rs751584545 Chr.11: 6002945-6002945 on Build GRCh38
rs760462406 Chr.11: 6002816-6002816 on Build GRCh38
rs1020367117 Chr.11: 6002841-6002841 on Build GRCh38
rs746927168 Chr.11: 6002977-6002977 on Build GRCh38
rs371788687 Chr.11: 6003032-6003032 on Build GRCh38
rs200789526 Chr.11: 6003021-6003021 on Build GRCh38
rs779777718 Chr.11: 6002904-6002904 on Build GRCh38
rs772997785 Chr.11: 6003002-6003002 on Build GRCh38
rs573348744 Chr.11: 6002814-6002814 on Build GRCh38
rs780957796 Chr.11: 6002984-6002984 on Build GRCh38
rs771468001 Chr.11: 6002898-6002898 on Build GRCh38
rs780918915 Chr.11: 6003073-6003073 on Build GRCh38
rs367924197 Chr.11: 6003012-6003012 on Build GRCh38
rs751003660 Chr.11: 6003064-6003064 on Build GRCh38
rs749071277 Chr.11: 6003079-6003079 on Build GRCh38
rs763089460 Chr.11: 6003077-6003078 on Build GRCh38
rs940851434 Chr.11: 6002924-6002924 on Build GRCh38
rs757714591 Chr.11: 6002974-6002974 on Build GRCh38
rs199693548 Chr.11: 6002888-6002888 on Build GRCh38
rs374846573 Chr.11: 6002895-6002895 on Build GRCh38
rs533058344 Chr.11: 6002992-6002992 on Build GRCh38
rs747895571 Chr.11: 6002987-6002987 on Build GRCh38
rs371483800 Chr.11: 6002825-6002825 on Build GRCh38
rs372809864 Chr.11: 6003093-6003093 on Build GRCh38
rs374429426 Chr.11: 6002857-6002857 on Build GRCh38
rs889408530 Chr.11: 6003028-6003028 on Build GRCh38
rs754512143 Chr.11: 6003068-6003068 on Build GRCh38
rs776498358 Chr.11: 6002829-6002829 on Build GRCh38
rs373194993 Chr.11: 6003019-6003019 on Build GRCh38
rs779747700 Chr.11: 6002845-6002845 on Build GRCh38
rs773324649 Chr.11: 6002937-6002937 on Build GRCh38
rs150304188 Chr.11: 6003051-6003051 on Build GRCh38
rs752958653 Chr.11: 6002963-6002963 on Build GRCh38
rs748964070 Chr.11: 6002939-6002941 on Build GRCh38
rs759635784 Chr.11: 6002949-6002949 on Build GRCh38
rs146654386 Chr.11: 6002883-6002883 on Build GRCh38
rs1007184485 Chr.11: 6002782-6002782 on Build GRCh38
rs540849050 Chr.11: 6002856-6002856 on Build GRCh38
rs747472279 Chr.11: 6002995-6002996 on Build GRCh38
rs983519493 Chr.11: 6002835-6002835 on Build GRCh38
rs773749998 Chr.11: 6002961-6002961 on Build GRCh38

View Full Product Details