Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41754528-41755030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JUP
Entrez Gene ID
3728
Gene Name
junction plakoglobin
Gene Aliases
ARVD12, CTNNG, DP3, DPIII, PDGB, PKGB
UniGene
Hs.514174
Gene Chromosome Location
Chr.17: 41754607-41786853 on Build GRCh38
Transcripts
NM_002230.2 NM_021991.2 XM_006721873.2 XM_006721874.2 XM_006721875.1 XM_006721878.1 XM_011524753.2 XM_011524755.1 XM_011524756.1 XM_011524757.2 XM_011524758.1 XM_017024588.1 XM_017024589.1 XM_017024590.1

SNP Detail

SNP ID Chromosome Location
rs73983655 Chr.17: 41754593-41754593 on Build GRCh38
rs539534709 Chr.17: 41754608-41754608 on Build GRCh38
rs545304563 Chr.17: 41754737-41754738 on Build GRCh38
rs144670936 Chr.17: 41754715-41754715 on Build GRCh38
rs185419450 Chr.17: 41754639-41754639 on Build GRCh38
rs951066482 Chr.17: 41754758-41754758 on Build GRCh38
rs892429843 Chr.17: 41754547-41754547 on Build GRCh38
rs575636161 Chr.17: 41754620-41754620 on Build GRCh38
rs543772543 Chr.17: 41754737-41754737 on Build GRCh38
rs1126985 Chr.17: 41754787-41754787 on Build GRCh38
rs372393793 Chr.17: 41754638-41754638 on Build GRCh38
rs548302699 Chr.17: 41754836-41754836 on Build GRCh38
rs551870163 Chr.17: 41754957-41754957 on Build GRCh38
rs925107477 Chr.17: 41754944-41754944 on Build GRCh38
rs1052343211 Chr.17: 41754951-41754951 on Build GRCh38
rs569650118 Chr.17: 41754973-41754973 on Build GRCh38
rs55919561 Chr.17: 41754891-41754891 on Build GRCh38
rs559563707 Chr.17: 41754743-41754743 on Build GRCh38
rs936596217 Chr.17: 41754945-41754945 on Build GRCh38
rs1025317179 Chr.17: 41754742-41754742 on Build GRCh38
rs993715499 Chr.17: 41754738-41754738 on Build GRCh38
rs781969877 Chr.17: 41754963-41754963 on Build GRCh38
rs529819746 Chr.17: 41754759-41754759 on Build GRCh38
rs187950810 Chr.17: 41754718-41754718 on Build GRCh38
rs782385635 Chr.17: 41754981-41754981 on Build GRCh38
rs1008162338 Chr.17: 41754677-41754677 on Build GRCh38
rs569644513 Chr.17: 41754838-41754838 on Build GRCh38
rs997472174 Chr.17: 41754600-41754600 on Build GRCh38
rs917683435 Chr.17: 41754852-41754852 on Build GRCh38
rs578142472 Chr.17: 41754888-41754888 on Build GRCh38
rs563541627 Chr.17: 41754555-41754555 on Build GRCh38

View Full Product Details