Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41974844-41975352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNP
Entrez Gene ID
1267
Gene Name
2',3'-cyclic nucleotide 3' phosphodiesterase
Gene Aliases
CNP1
UniGene
Hs.273621
Gene Chromosome Location
Chr.17: 41966741-41977736 on Build GRCh38
Transcripts
NM_033133.4 XM_006721701.3 XM_011524340.2

SNP Detail

SNP ID Chromosome Location
rs528942789 Chr.17: 41974950-41974950 on Build GRCh38
rs936489934 Chr.17: 41975294-41975294 on Build GRCh38
rs550164128 Chr.17: 41974972-41974972 on Build GRCh38
rs951177954 Chr.17: 41975079-41975079 on Build GRCh38
rs567080846 Chr.17: 41975118-41975118 on Build GRCh38
rs534125948 Chr.17: 41975170-41975170 on Build GRCh38
rs782316874 Chr.17: 41975271-41975271 on Build GRCh38
rs963529911 Chr.17: 41975056-41975056 on Build GRCh38
rs782602070 Chr.17: 41974995-41974996 on Build GRCh38
rs568054718 Chr.17: 41974946-41974946 on Build GRCh38
rs997912218 Chr.17: 41974989-41974989 on Build GRCh38
rs555868383 Chr.17: 41975171-41975171 on Build GRCh38
rs553386621 Chr.17: 41974908-41974908 on Build GRCh38
rs11296 Chr.17: 41975042-41975042 on Build GRCh38
rs111525390 Chr.17: 41975175-41975175 on Build GRCh38
rs1052470553 Chr.17: 41974971-41974971 on Build GRCh38
rs767497305 Chr.17: 41974959-41974959 on Build GRCh38
rs913958844 Chr.17: 41975322-41975322 on Build GRCh38
rs782581288 Chr.17: 41975242-41975242 on Build GRCh38
rs568172748 Chr.17: 41975016-41975016 on Build GRCh38
rs1052248993 Chr.17: 41975308-41975308 on Build GRCh38
rs936543755 Chr.17: 41974953-41974953 on Build GRCh38
rs781856979 Chr.17: 41975165-41975165 on Build GRCh38
rs1045263348 Chr.17: 41974943-41974943 on Build GRCh38
rs782182093 Chr.17: 41975270-41975270 on Build GRCh38
rs759557900 Chr.17: 41974952-41974952 on Build GRCh38
rs969678365 Chr.17: 41975260-41975260 on Build GRCh38
rs1007811506 Chr.17: 41975032-41975032 on Build GRCh38
rs948017677 Chr.17: 41974923-41974923 on Build GRCh38
rs756066409 Chr.17: 41975033-41975033 on Build GRCh38
rs993703871 Chr.17: 41975069-41975069 on Build GRCh38
rs558013427 Chr.17: 41975070-41975070 on Build GRCh38
rs113598886 Chr.17: 41975176-41975176 on Build GRCh38
rs889342111 Chr.17: 41975019-41975019 on Build GRCh38

View Full Product Details