Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42027497-42027987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF385C
Entrez Gene ID
201181
Gene Name
zinc finger protein 385C
Gene Aliases
-
UniGene
Hs.443140
Gene Chromosome Location
Chr.17: 42025576-42065668 on Build GRCh38
Transcripts
NM_001242704.1 XM_006721751.3 XM_011524479.2 XM_011524482.2 XM_011524483.2 XM_017024314.1 XM_017024315.1 XM_017024316.1 XM_017024317.1 XM_017024318.1 XM_017024319.1 XM_017024320.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6829807 c.264A>G p.A88A Chr.17: 42027642-42027642 on Build GRCh38
4563906 c.245G>A p.S82N Chr.17: 42027661-42027661 on Build GRCh38
5084849 c.206G>A p.R69H Chr.17: 42027700-42027700 on Build GRCh38
6189940 c.178-1G>T p.? Chr.17: 42027729-42027729 on Build GRCh38
7399034 c.229C>G p.P77A Chr.17: 42027677-42027677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782342323 Chr.17: 42027700-42027700 on Build GRCh38
rs925377743 Chr.17: 42027538-42027538 on Build GRCh38
rs377419568 Chr.17: 42027649-42027649 on Build GRCh38
rs370180261 Chr.17: 42027721-42027721 on Build GRCh38
rs1052405111 Chr.17: 42027545-42027545 on Build GRCh38
rs781984849 Chr.17: 42027631-42027631 on Build GRCh38
rs782795172 Chr.17: 42027748-42027748 on Build GRCh38
rs573411707 Chr.17: 42027678-42027678 on Build GRCh38
rs530693699 Chr.17: 42027886-42027886 on Build GRCh38
rs995234827 Chr.17: 42027953-42027953 on Build GRCh38
rs781970480 Chr.17: 42027701-42027701 on Build GRCh38
rs145255298 Chr.17: 42027645-42027645 on Build GRCh38
rs563277863 Chr.17: 42027861-42027861 on Build GRCh38
rs782024393 Chr.17: 42027712-42027712 on Build GRCh38
rs538917044 Chr.17: 42027797-42027797 on Build GRCh38
rs782747386 Chr.17: 42027644-42027644 on Build GRCh38
rs781999775 Chr.17: 42027822-42027822 on Build GRCh38
rs575977771 Chr.17: 42027575-42027575 on Build GRCh38
rs782077036 Chr.17: 42027725-42027725 on Build GRCh38
rs782783397 Chr.17: 42027717-42027717 on Build GRCh38
rs782731350 Chr.17: 42027731-42027731 on Build GRCh38
rs947950773 Chr.17: 42027533-42027533 on Build GRCh38
rs116660545 Chr.17: 42027858-42027858 on Build GRCh38
rs150004992 Chr.17: 42027640-42027640 on Build GRCh38
rs782225562 Chr.17: 42027577-42027577 on Build GRCh38
rs781816864 Chr.17: 42027681-42027681 on Build GRCh38
rs151041175 Chr.17: 42027715-42027715 on Build GRCh38
rs782494610 Chr.17: 42027764-42027764 on Build GRCh38
rs186155621 Chr.17: 42027904-42027904 on Build GRCh38
rs563930327 Chr.17: 42027927-42027927 on Build GRCh38
rs782794673 Chr.17: 42027673-42027673 on Build GRCh38
rs374964032 Chr.17: 42027613-42027613 on Build GRCh38
rs988214284 Chr.17: 42027743-42027743 on Build GRCh38
rs782333255 Chr.17: 42027705-42027705 on Build GRCh38
rs782075181 Chr.17: 42027656-42027656 on Build GRCh38
rs375508779 Chr.17: 42027824-42027824 on Build GRCh38
rs73983697 Chr.17: 42027965-42027965 on Build GRCh38
rs1024924082 Chr.17: 42027689-42027689 on Build GRCh38
rs781900519 Chr.17: 42027751-42027751 on Build GRCh38
rs782542339 Chr.17: 42027692-42027692 on Build GRCh38
rs782002065 Chr.17: 42027634-42027634 on Build GRCh38
rs782387060 Chr.17: 42027569-42027569 on Build GRCh38
rs933376243 Chr.17: 42027777-42027777 on Build GRCh38
rs781909186 Chr.17: 42027674-42027674 on Build GRCh38
rs782482146 Chr.17: 42027683-42027683 on Build GRCh38
rs145565025 Chr.17: 42027966-42027966 on Build GRCh38
rs781784798 Chr.17: 42027734-42027734 on Build GRCh38
rs782787838 Chr.17: 42027680-42027680 on Build GRCh38
rs145333824 Chr.17: 42027630-42027630 on Build GRCh38
rs546678227 Chr.17: 42027931-42027931 on Build GRCh38
rs150227256 Chr.17: 42027930-42027930 on Build GRCh38
rs148762556 Chr.17: 42027693-42027693 on Build GRCh38
rs782613235 Chr.17: 42027767-42027767 on Build GRCh38
rs782199531 Chr.17: 42027691-42027691 on Build GRCh38
rs373988096 Chr.17: 42027737-42027737 on Build GRCh38
rs370131301 Chr.17: 42027548-42027548 on Build GRCh38
rs540075849 Chr.17: 42027626-42027626 on Build GRCh38
rs140982734 Chr.17: 42027768-42027768 on Build GRCh38
rs782184069 Chr.17: 42027704-42027704 on Build GRCh38
rs995149430 Chr.17: 42027650-42027650 on Build GRCh38
rs781940212 Chr.17: 42027618-42027618 on Build GRCh38
rs782260940 Chr.17: 42027699-42027699 on Build GRCh38
rs782310937 Chr.17: 42027580-42027580 on Build GRCh38

View Full Product Details