Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31781585-31782086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VARS
Entrez Gene ID
7407
Gene Name
valyl-tRNA synthetase
Gene Aliases
G7A, VARS1, VARS2
UniGene
Hs.520026
Gene Chromosome Location
Chr.6: 31777518-31795935 on Build GRCh38
Transcripts
NM_006295.2 XM_005249362.2 XM_017011246.1 XM_017011247.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6106563 c.2348-1G>T p.? Chr.6: 31781762-31781762 on Build GRCh38
356191 c.2248G>A p.D750N Chr.6: 31781946-31781946 on Build GRCh38
4740148 c.2295G>A p.E765E Chr.6: 31781899-31781899 on Build GRCh38
741859 c.2283G>T p.A761A Chr.6: 31781911-31781911 on Build GRCh38
6173936 c.2293G>T p.E765* Chr.6: 31781901-31781901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760270776 Chr.6: 31781823-31781823 on Build GRCh38
rs759184244 Chr.6: 31781941-31781941 on Build GRCh38
rs55786236 Chr.6: 31781904-31781904 on Build GRCh38
rs368747450 Chr.6: 31781665-31781665 on Build GRCh38
rs369603016 Chr.6: 31781880-31781880 on Build GRCh38
rs776260072 Chr.6: 31782058-31782058 on Build GRCh38
rs930599654 Chr.6: 31781781-31781781 on Build GRCh38
rs772110719 Chr.6: 31781918-31781918 on Build GRCh38
rs148246629 Chr.6: 31781845-31781845 on Build GRCh38
rs375854405 Chr.6: 31781664-31781664 on Build GRCh38
rs758822153 Chr.6: 31781755-31781755 on Build GRCh38
rs780328097 Chr.6: 31781998-31781998 on Build GRCh38
rs555708628 Chr.6: 31781715-31781715 on Build GRCh38
rs1038728929 Chr.6: 31781672-31781672 on Build GRCh38
rs755472759 Chr.6: 31781721-31781721 on Build GRCh38
rs535208710 Chr.6: 31782018-31782018 on Build GRCh38
rs758873313 Chr.6: 31781877-31781877 on Build GRCh38
rs368988920 Chr.6: 31781940-31781940 on Build GRCh38
rs367909977 Chr.6: 31781671-31781671 on Build GRCh38
rs761826212 Chr.6: 31781809-31781809 on Build GRCh38
rs748476862 Chr.6: 31781645-31781645 on Build GRCh38
rs371885173 Chr.6: 31781674-31781674 on Build GRCh38
rs778162056 Chr.6: 31781879-31781879 on Build GRCh38
rs568116738 Chr.6: 31781818-31781818 on Build GRCh38
rs753296702 Chr.6: 31781832-31781832 on Build GRCh38
rs1049425064 Chr.6: 31781812-31781812 on Build GRCh38
rs772289276 Chr.6: 31781656-31781656 on Build GRCh38
rs758417251 Chr.6: 31782045-31782045 on Build GRCh38
rs543561819 Chr.6: 31781805-31781806 on Build GRCh38
rs753077261 Chr.6: 31781750-31781750 on Build GRCh38
rs375095621 Chr.6: 31781839-31781839 on Build GRCh38
rs945520107 Chr.6: 31781659-31781659 on Build GRCh38
rs112667566 Chr.6: 31781846-31781846 on Build GRCh38
rs764768184 Chr.6: 31781951-31781951 on Build GRCh38
rs140366323 Chr.6: 31781871-31781871 on Build GRCh38
rs781152286 Chr.6: 31781792-31781792 on Build GRCh38
rs371948566 Chr.6: 31781784-31781784 on Build GRCh38
rs370411565 Chr.6: 31782035-31782035 on Build GRCh38
rs746911944 Chr.6: 31782053-31782053 on Build GRCh38
rs766017359 Chr.6: 31781828-31781828 on Build GRCh38
rs774369035 Chr.6: 31781801-31781801 on Build GRCh38
rs755491126 Chr.6: 31781636-31781636 on Build GRCh38
rs756370926 Chr.6: 31781985-31781985 on Build GRCh38
rs145442830 Chr.6: 31781881-31781881 on Build GRCh38
rs746755292 Chr.6: 31781921-31781921 on Build GRCh38
rs745919752 Chr.6: 31781797-31781797 on Build GRCh38
rs751874117 Chr.6: 31781971-31781971 on Build GRCh38
rs932437475 Chr.6: 31781811-31781811 on Build GRCh38
rs743399 Chr.6: 31782027-31782027 on Build GRCh38
rs779329829 Chr.6: 31781742-31781742 on Build GRCh38
rs752569643 Chr.6: 31782052-31782052 on Build GRCh38
rs944459535 Chr.6: 31781666-31781666 on Build GRCh38
rs143162168 Chr.6: 31781759-31781759 on Build GRCh38
rs745571001 Chr.6: 31782064-31782064 on Build GRCh38
rs990121445 Chr.6: 31781609-31781609 on Build GRCh38
rs762587470 Chr.6: 31781962-31781962 on Build GRCh38
rs749824510 Chr.6: 31781627-31781627 on Build GRCh38
rs1022471020 Chr.6: 31782034-31782034 on Build GRCh38
rs924551525 Chr.6: 31781743-31781743 on Build GRCh38
rs751148803 Chr.6: 31781969-31781969 on Build GRCh38
rs754676615 Chr.6: 31781983-31781983 on Build GRCh38
rs112783854 Chr.6: 31781764-31781764 on Build GRCh38
rs373146932 Chr.6: 31782039-31782039 on Build GRCh38
rs765738671 Chr.6: 31781991-31781991 on Build GRCh38
rs765943997 Chr.6: 31781937-31781937 on Build GRCh38
rs776894128 Chr.6: 31781764-31781765 on Build GRCh38
rs767288425 Chr.6: 31781680-31781680 on Build GRCh38
rs759605189 Chr.6: 31781805-31781805 on Build GRCh38
rs777712330 Chr.6: 31782049-31782049 on Build GRCh38
rs867991903 Chr.6: 31781926-31781926 on Build GRCh38
rs940705577 Chr.6: 31781826-31781826 on Build GRCh38
rs776988621 Chr.6: 31781653-31781653 on Build GRCh38
rs771161682 Chr.6: 31781767-31781767 on Build GRCh38
rs762845967 Chr.6: 31781668-31781668 on Build GRCh38
rs138353806 Chr.6: 31781912-31781912 on Build GRCh38
rs770826721 Chr.6: 31782055-31782055 on Build GRCh38
rs759033237 Chr.6: 31781840-31781840 on Build GRCh38
rs1051628034 Chr.6: 31781819-31781819 on Build GRCh38
rs781407972 Chr.6: 31781887-31781887 on Build GRCh38
rs915432003 Chr.6: 31781631-31781631 on Build GRCh38
rs773366658 Chr.6: 31781663-31781663 on Build GRCh38
rs764330687 Chr.6: 31782051-31782051 on Build GRCh38
rs534149314 Chr.6: 31781667-31781667 on Build GRCh38
rs763541565 Chr.6: 31781979-31781979 on Build GRCh38
rs747236858 Chr.6: 31781763-31781763 on Build GRCh38
rs757583659 Chr.6: 31781972-31781972 on Build GRCh38
rs779274444 Chr.6: 31781641-31781641 on Build GRCh38
rs376163798 Chr.6: 31781939-31781939 on Build GRCh38
rs760368252 Chr.6: 31781704-31781704 on Build GRCh38
rs149706716 Chr.6: 31782033-31782033 on Build GRCh38
rs761632486 Chr.6: 31781675-31781675 on Build GRCh38
rs748132817 Chr.6: 31781913-31781913 on Build GRCh38
rs750154252 Chr.6: 31781694-31781694 on Build GRCh38
rs372693339 Chr.6: 31781919-31781919 on Build GRCh38
rs765844833 Chr.6: 31781711-31781711 on Build GRCh38
rs771152490 Chr.6: 31781670-31781671 on Build GRCh38
rs768717531 Chr.6: 31781798-31781798 on Build GRCh38
rs377367472 Chr.6: 31781978-31781979 on Build GRCh38

View Full Product Details