Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36306161-36306696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNPLA1
Entrez Gene ID
285848
Gene Name
patatin like phospholipase domain containing 1
Gene Aliases
ARCI10, dJ50J22.1
UniGene
Hs.407002
Gene Chromosome Location
Chr.6: 36242523-36313955 on Build GRCh38
Transcripts
NM_001145716.2 NM_001145717.1 NM_173676.2 XM_011514519.2 XM_011514520.2 XM_017010776.1 XM_017010777.1 XM_017010778.1 XM_017010779.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4631112 c.1422C>T p.A474A Chr.6: 36306329-36306329 on Build GRCh38
6248711 c.1133G>T p.S378I Chr.6: 36306325-36306325 on Build GRCh38
4716222 c.1422C>T p.S474S Chr.6: 36306326-36306326 on Build GRCh38
3873514 c.1472+1G>A p.? Chr.6: 36306377-36306377 on Build GRCh38
4631113 c.1137C>T p.A379A Chr.6: 36306329-36306329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs946046733 Chr.6: 36306439-36306439 on Build GRCh38
rs917289436 Chr.6: 36306438-36306438 on Build GRCh38
rs768399485 Chr.6: 36306255-36306255 on Build GRCh38
rs780350401 Chr.6: 36306251-36306251 on Build GRCh38
rs771277982 Chr.6: 36306671-36306671 on Build GRCh38
rs780841373 Chr.6: 36306410-36306410 on Build GRCh38
rs759230393 Chr.6: 36306301-36306301 on Build GRCh38
rs139909055 Chr.6: 36306327-36306327 on Build GRCh38
rs764436384 Chr.6: 36306308-36306308 on Build GRCh38
rs527867287 Chr.6: 36306581-36306581 on Build GRCh38
rs757845483 Chr.6: 36306610-36306612 on Build GRCh38
rs549597236 Chr.6: 36306663-36306663 on Build GRCh38
rs373131816 Chr.6: 36306386-36306386 on Build GRCh38
rs371589583 Chr.6: 36306421-36306421 on Build GRCh38
rs751556946 Chr.6: 36306332-36306332 on Build GRCh38
rs766879431 Chr.6: 36306425-36306425 on Build GRCh38
rs1051298374 Chr.6: 36306588-36306588 on Build GRCh38
rs140803733 Chr.6: 36306429-36306429 on Build GRCh38
rs544187428 Chr.6: 36306580-36306580 on Build GRCh38
rs1023035692 Chr.6: 36306522-36306522 on Build GRCh38
rs745678573 Chr.6: 36306400-36306400 on Build GRCh38
rs768460025 Chr.6: 36306404-36306404 on Build GRCh38
rs774690711 Chr.6: 36306407-36306407 on Build GRCh38
rs778413773 Chr.6: 36306256-36306256 on Build GRCh38
rs746892136 Chr.6: 36306273-36306273 on Build GRCh38
rs766844556 Chr.6: 36306334-36306334 on Build GRCh38
rs577964639 Chr.6: 36306250-36306250 on Build GRCh38
rs1004305131 Chr.6: 36306656-36306656 on Build GRCh38
rs747678371 Chr.6: 36306556-36306556 on Build GRCh38
rs922489274 Chr.6: 36306382-36306388 on Build GRCh38
rs575769690 Chr.6: 36306419-36306419 on Build GRCh38
rs747745268 Chr.6: 36306357-36306357 on Build GRCh38
rs952442073 Chr.6: 36306528-36306528 on Build GRCh38
rs368844811 Chr.6: 36306238-36306238 on Build GRCh38
rs866355921 Chr.6: 36306328-36306328 on Build GRCh38
rs776607279 Chr.6: 36306295-36306295 on Build GRCh38
rs560871305 Chr.6: 36306497-36306497 on Build GRCh38
rs755243278 Chr.6: 36306247-36306247 on Build GRCh38
rs765435490 Chr.6: 36306262-36306263 on Build GRCh38
rs12197079 Chr.6: 36306376-36306376 on Build GRCh38
rs371029559 Chr.6: 36306571-36306571 on Build GRCh38
rs925875610 Chr.6: 36306496-36306496 on Build GRCh38
rs543009277 Chr.6: 36306428-36306428 on Build GRCh38
rs45621032 Chr.6: 36306371-36306371 on Build GRCh38
rs1034468211 Chr.6: 36306417-36306417 on Build GRCh38
rs998986566 Chr.6: 36306454-36306454 on Build GRCh38
rs886825301 Chr.6: 36306614-36306614 on Build GRCh38
rs922456784 Chr.6: 36306625-36306625 on Build GRCh38
rs758243716 Chr.6: 36306351-36306351 on Build GRCh38
rs761915938 Chr.6: 36306409-36306409 on Build GRCh38
rs750865772 Chr.6: 36306245-36306245 on Build GRCh38
rs538566938 Chr.6: 36306257-36306257 on Build GRCh38
rs750116578 Chr.6: 36306330-36306330 on Build GRCh38
rs533404664 Chr.6: 36306440-36306440 on Build GRCh38
rs1039584574 Chr.6: 36306518-36306518 on Build GRCh38
rs754299523 Chr.6: 36306426-36306426 on Build GRCh38
rs574942304 Chr.6: 36306406-36306406 on Build GRCh38
rs767621138 Chr.6: 36306416-36306416 on Build GRCh38
rs771414777 Chr.6: 36306267-36306267 on Build GRCh38
rs899666997 Chr.6: 36306519-36306519 on Build GRCh38
rs758032489 Chr.6: 36306365-36306365 on Build GRCh38
rs935871936 Chr.6: 36306516-36306516 on Build GRCh38
rs545401767 Chr.6: 36306486-36306486 on Build GRCh38
rs748929081 Chr.6: 36306252-36306252 on Build GRCh38
rs572168497 Chr.6: 36306286-36306286 on Build GRCh38
rs143723135 Chr.6: 36306338-36306338 on Build GRCh38
rs145988979 Chr.6: 36306326-36306326 on Build GRCh38
rs903350354 Chr.6: 36306447-36306448 on Build GRCh38
rs375616407 Chr.6: 36306268-36306268 on Build GRCh38
rs770880134 Chr.6: 36306277-36306277 on Build GRCh38
rs755289820 Chr.6: 36306344-36306344 on Build GRCh38

View Full Product Details