Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 149213566-149213965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF282
Entrez Gene ID
8427
Gene Name
zinc finger protein 282
Gene Aliases
HUB1
UniGene
Hs.729056
Gene Chromosome Location
Chr.7: 149195462-149226248 on Build GRCh38
Transcripts
NM_001303481.1 NM_003575.3 XM_006716151.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5452167 c.1159C>T p.L387F Chr.7: 149213793-149213793 on Build GRCh38
6829125 c.1180+3G>A p.? Chr.7: 149213817-149213817 on Build GRCh38
3259924 c.1139G>A p.R380H Chr.7: 149213773-149213773 on Build GRCh38
744955 c.1070C>G p.S357C Chr.7: 149213704-149213704 on Build GRCh38
5552120 c.1088A>G p.D363G Chr.7: 149213722-149213722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs947086576 Chr.7: 149213910-149213910 on Build GRCh38
rs770429822 Chr.7: 149213708-149213708 on Build GRCh38
rs779303278 Chr.7: 149213776-149213776 on Build GRCh38
rs748065196 Chr.7: 149213937-149213937 on Build GRCh38
rs920794842 Chr.7: 149213873-149213873 on Build GRCh38
rs762848752 Chr.7: 149213837-149213837 on Build GRCh38
rs750662958 Chr.7: 149213653-149213653 on Build GRCh38
rs915451712 Chr.7: 149213880-149213880 on Build GRCh38
rs1019001203 Chr.7: 149213619-149213619 on Build GRCh38
rs148349879 Chr.7: 149213775-149213775 on Build GRCh38
rs376015918 Chr.7: 149213797-149213797 on Build GRCh38
rs1009316758 Chr.7: 149213662-149213662 on Build GRCh38
rs747358431 Chr.7: 149213821-149213821 on Build GRCh38
rs1002520534 Chr.7: 149213926-149213926 on Build GRCh38
rs147914591 Chr.7: 149213678-149213678 on Build GRCh38
rs114974629 Chr.7: 149213756-149213756 on Build GRCh38
rs993987977 Chr.7: 149213769-149213769 on Build GRCh38
rs746453982 Chr.7: 149213793-149213793 on Build GRCh38
rs748699652 Chr.7: 149213688-149213688 on Build GRCh38
rs759210336 Chr.7: 149213750-149213750 on Build GRCh38
rs538624117 Chr.7: 149213883-149213883 on Build GRCh38
rs1009642172 Chr.7: 149213938-149213938 on Build GRCh38
rs753193861 Chr.7: 149213781-149213781 on Build GRCh38
rs562645389 Chr.7: 149213813-149213813 on Build GRCh38
rs1040781413 Chr.7: 149213930-149213930 on Build GRCh38
rs780678027 Chr.7: 149213814-149213814 on Build GRCh38
rs780331091 Chr.7: 149213661-149213661 on Build GRCh38
rs935616623 Chr.7: 149213857-149213857 on Build GRCh38
rs1056846397 Chr.7: 149213933-149213933 on Build GRCh38
rs930875536 Chr.7: 149213902-149213902 on Build GRCh38
rs775546716 Chr.7: 149213753-149213753 on Build GRCh38
rs552085439 Chr.7: 149213787-149213787 on Build GRCh38
rs142167998 Chr.7: 149213861-149213861 on Build GRCh38
rs961307226 Chr.7: 149213768-149213768 on Build GRCh38
rs776367020 Chr.7: 149213710-149213710 on Build GRCh38
rs567739559 Chr.7: 149213874-149213874 on Build GRCh38
rs889149551 Chr.7: 149213599-149213599 on Build GRCh38
rs1053906477 Chr.7: 149213590-149213590 on Build GRCh38
rs202186203 Chr.7: 149213786-149213786 on Build GRCh38
rs142354798 Chr.7: 149213618-149213618 on Build GRCh38
rs199929554 Chr.7: 149213706-149213706 on Build GRCh38
rs766466237 Chr.7: 149213773-149213773 on Build GRCh38
rs144443267 Chr.7: 149213911-149213911 on Build GRCh38
rs756359858 Chr.7: 149213660-149213660 on Build GRCh38
rs892864382 Chr.7: 149213606-149213606 on Build GRCh38
rs570765039 Chr.7: 149213924-149213924 on Build GRCh38
rs775493804 Chr.7: 149213827-149213827 on Build GRCh38
rs769670725 Chr.7: 149213752-149213752 on Build GRCh38
rs768659891 Chr.7: 149213846-149213846 on Build GRCh38
rs375051278 Chr.7: 149213667-149213667 on Build GRCh38
rs897066739 Chr.7: 149213936-149213936 on Build GRCh38
rs755396813 Chr.7: 149213674-149213674 on Build GRCh38
rs750463028 Chr.7: 149213762-149213762 on Build GRCh38
rs116432078 Chr.7: 149213772-149213772 on Build GRCh38
rs763042254 Chr.7: 149213754-149213754 on Build GRCh38
rs771553474 Chr.7: 149213692-149213692 on Build GRCh38
rs369380272 Chr.7: 149213823-149213823 on Build GRCh38
rs531293375 Chr.7: 149213824-149213824 on Build GRCh38
rs1050731477 Chr.7: 149213908-149213908 on Build GRCh38
rs772772738 Chr.7: 149213698-149213698 on Build GRCh38
rs147846260 Chr.7: 149213919-149213919 on Build GRCh38
rs578062173 Chr.7: 149213774-149213774 on Build GRCh38
rs980752496 Chr.7: 149213751-149213751 on Build GRCh38
rs774455023 Chr.7: 149213856-149213856 on Build GRCh38
rs200623057 Chr.7: 149213652-149213652 on Build GRCh38

View Full Product Details