Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 149223700-149224192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF282
Entrez Gene ID
8427
Gene Name
zinc finger protein 282
Gene Aliases
HUB1
UniGene
Hs.729056
Gene Chromosome Location
Chr.7: 149195462-149226248 on Build GRCh38
Transcripts
NM_001303481.1 NM_003575.3 XM_006716151.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6370732 c.1284G>T p.Q428H Chr.7: 149223915-149223915 on Build GRCh38
7087932 c.1534G>T p.G512C Chr.7: 149224165-149224165 on Build GRCh38
4650741 c.1537G>A p.A513T Chr.7: 149224168-149224168 on Build GRCh38
5431156 c.1212C>A p.P404P Chr.7: 149223843-149223843 on Build GRCh38
6431978 c.1216G>C p.A406P Chr.7: 149223847-149223847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949026035 Chr.7: 149223785-149223785 on Build GRCh38
rs372401444 Chr.7: 149223999-149223999 on Build GRCh38
rs1053148348 Chr.7: 149223851-149223851 on Build GRCh38
rs763271661 Chr.7: 149224105-149224105 on Build GRCh38
rs574494573 Chr.7: 149223750-149223750 on Build GRCh38
rs768606730 Chr.7: 149223899-149223899 on Build GRCh38
rs780178844 Chr.7: 149223755-149223755 on Build GRCh38
rs758783337 Chr.7: 149223804-149223804 on Build GRCh38
rs1053094474 Chr.7: 149224037-149224037 on Build GRCh38
rs1004726495 Chr.7: 149224087-149224087 on Build GRCh38
rs774128320 Chr.7: 149223921-149223921 on Build GRCh38
rs756700401 Chr.7: 149223845-149223845 on Build GRCh38
rs764490886 Chr.7: 149224114-149224114 on Build GRCh38
rs973104910 Chr.7: 149223948-149223948 on Build GRCh38
rs1035955037 Chr.7: 149224072-149224072 on Build GRCh38
rs567367506 Chr.7: 149224071-149224071 on Build GRCh38
rs559639142 Chr.7: 149223761-149223761 on Build GRCh38
rs907577810 Chr.7: 149223932-149223932 on Build GRCh38
rs990565954 Chr.7: 149223758-149223758 on Build GRCh38
rs769712882 Chr.7: 149223855-149223856 on Build GRCh38
rs540265325 Chr.7: 149223752-149223752 on Build GRCh38
rs545260496 Chr.7: 149223888-149223888 on Build GRCh38
rs376301393 Chr.7: 149224155-149224155 on Build GRCh38
rs1021730754 Chr.7: 149223887-149223888 on Build GRCh38
rs781236720 Chr.7: 149223852-149223863 on Build GRCh38
rs907628398 Chr.7: 149223726-149223726 on Build GRCh38
rs775719118 Chr.7: 149224101-149224101 on Build GRCh38
rs1036044800 Chr.7: 149223915-149223915 on Build GRCh38
rs753989714 Chr.7: 149223771-149223771 on Build GRCh38
rs941809908 Chr.7: 149223733-149223733 on Build GRCh38
rs769506197 Chr.7: 149223860-149223860 on Build GRCh38
rs953291766 Chr.7: 149224154-149224154 on Build GRCh38
rs774358396 Chr.7: 149223976-149223978 on Build GRCh38
rs932799721 Chr.7: 149223955-149223955 on Build GRCh38
rs960270116 Chr.7: 149223920-149223920 on Build GRCh38
rs765446421 Chr.7: 149223792-149223792 on Build GRCh38
rs780813532 Chr.7: 149223847-149223847 on Build GRCh38
rs1001372084 Chr.7: 149224064-149224064 on Build GRCh38
rs778211287 Chr.7: 149223836-149223836 on Build GRCh38
rs963030838 Chr.7: 149223940-149223940 on Build GRCh38
rs983121436 Chr.7: 149223922-149223922 on Build GRCh38
rs749044528 Chr.7: 149223871-149223871 on Build GRCh38
rs920066969 Chr.7: 149223746-149223746 on Build GRCh38
rs944310154 Chr.7: 149224005-149224005 on Build GRCh38
rs752146819 Chr.7: 149223840-149223840 on Build GRCh38
rs748170038 Chr.7: 149223923-149223923 on Build GRCh38
rs779867905 Chr.7: 149223866-149223866 on Build GRCh38
rs530683910 Chr.7: 149223979-149223979 on Build GRCh38
rs561005426 Chr.7: 149223728-149223728 on Build GRCh38
rs994550957 Chr.7: 149224056-149224057 on Build GRCh38
rs891866299 Chr.7: 149223882-149223882 on Build GRCh38
rs770907770 Chr.7: 149223954-149223954 on Build GRCh38
rs767915527 Chr.7: 149224151-149224151 on Build GRCh38
rs1014811124 Chr.7: 149224094-149224094 on Build GRCh38
rs3735312 Chr.7: 149223964-149223964 on Build GRCh38
rs759545294 Chr.7: 149224026-149224026 on Build GRCh38
rs1043123191 Chr.7: 149224053-149224053 on Build GRCh38
rs912809543 Chr.7: 149223985-149223985 on Build GRCh38
rs759865322 Chr.7: 149223787-149223787 on Build GRCh38
rs745988429 Chr.7: 149223856-149223861 on Build GRCh38
rs757849526 Chr.7: 149224145-149224145 on Build GRCh38
rs1001782237 Chr.7: 149223907-149223907 on Build GRCh38
rs115346898 Chr.7: 149223802-149223802 on Build GRCh38
rs779816578 Chr.7: 149224168-149224168 on Build GRCh38
rs10255054 Chr.7: 149224048-149224048 on Build GRCh38
rs977528519 Chr.7: 149223743-149223743 on Build GRCh38
rs1011602560 Chr.7: 149224049-149224049 on Build GRCh38
rs771189248 Chr.7: 149223801-149223802 on Build GRCh38
rs745504033 Chr.7: 149223853-149223853 on Build GRCh38
rs113503498 Chr.7: 149223894-149223894 on Build GRCh38
rs111843065 Chr.7: 149223738-149223738 on Build GRCh38
rs928835547 Chr.7: 149223854-149223854 on Build GRCh38
rs529738250 Chr.7: 149224143-149224143 on Build GRCh38
rs1035605862 Chr.7: 149223910-149223910 on Build GRCh38

View Full Product Details