Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150331690-150332198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC61
Entrez Gene ID
65999
Gene Name
leucine rich repeat containing 61
Gene Aliases
HSPC295
UniGene
Hs.647119
Gene Chromosome Location
Chr.7: 150322639-150338156 on Build GRCh38
Transcripts
NM_001142928.1 NM_023942.2 XM_006716095.2 NM_138434.2

Gene Symbol
ZBED6CL
Entrez Gene ID
113763
Gene Name
ZBED6 C-terminal like
Gene Aliases
C7orf29
UniGene
Hs.655915
Gene Chromosome Location
Chr.7: 150329849-150332721 on Build GRCh38
Transcripts
NM_001142928.1 NM_023942.2 XM_006716095.2 NM_138434.2

SNP Detail

SNP ID Chromosome Location
rs960821217 Chr.7: 150331737-150331737 on Build GRCh38
rs149036457 Chr.7: 150331867-150331867 on Build GRCh38
rs531150061 Chr.7: 150331733-150331733 on Build GRCh38
rs554512620 Chr.7: 150332011-150332011 on Build GRCh38
rs879304917 Chr.7: 150331784-150331784 on Build GRCh38
rs928232014 Chr.7: 150332121-150332121 on Build GRCh38
rs1045310336 Chr.7: 150331915-150331915 on Build GRCh38
rs12671192 Chr.7: 150332083-150332083 on Build GRCh38
rs1044233383 Chr.7: 150331953-150331954 on Build GRCh38
rs939354750 Chr.7: 150331926-150331926 on Build GRCh38
rs1015694493 Chr.7: 150332045-150332045 on Build GRCh38
rs759216758 Chr.7: 150332124-150332124 on Build GRCh38
rs988096151 Chr.7: 150332105-150332105 on Build GRCh38
rs374188039 Chr.7: 150331882-150331882 on Build GRCh38
rs1030632042 Chr.7: 150332075-150332075 on Build GRCh38
rs550928657 Chr.7: 150331771-150331771 on Build GRCh38
rs956407953 Chr.7: 150331878-150331878 on Build GRCh38
rs774789321 Chr.7: 150331964-150331964 on Build GRCh38
rs764678100 Chr.7: 150332173-150332173 on Build GRCh38
rs776198313 Chr.7: 150332104-150332104 on Build GRCh38
rs3735172 Chr.7: 150331869-150331869 on Build GRCh38
rs946549879 Chr.7: 150332140-150332140 on Build GRCh38
rs981470901 Chr.7: 150332110-150332110 on Build GRCh38
rs538021587 Chr.7: 150331985-150331985 on Build GRCh38
rs1052254957 Chr.7: 150331924-150331924 on Build GRCh38
rs915386293 Chr.7: 150331897-150331897 on Build GRCh38
rs961754508 Chr.7: 150331820-150331820 on Build GRCh38
rs868408426 Chr.7: 150331854-150331854 on Build GRCh38
rs953052174 Chr.7: 150332080-150332080 on Build GRCh38
rs143206388 Chr.7: 150332096-150332096 on Build GRCh38
rs973582669 Chr.7: 150331844-150331844 on Build GRCh38
rs1007242030 Chr.7: 150332018-150332018 on Build GRCh38
rs897923386 Chr.7: 150331949-150331949 on Build GRCh38
rs532938172 Chr.7: 150331918-150331918 on Build GRCh38
rs1004448176 Chr.7: 150332037-150332037 on Build GRCh38
rs982929079 Chr.7: 150331744-150331744 on Build GRCh38
rs535021890 Chr.7: 150331925-150331925 on Build GRCh38
rs138751323 Chr.7: 150331982-150331982 on Build GRCh38
rs762154374 Chr.7: 150332019-150332019 on Build GRCh38
rs574068408 Chr.7: 150332020-150332020 on Build GRCh38
rs897035194 Chr.7: 150331731-150331731 on Build GRCh38
rs952742158 Chr.7: 150331764-150331764 on Build GRCh38
rs116864401 Chr.7: 150332062-150332062 on Build GRCh38
rs969567629 Chr.7: 150332106-150332106 on Build GRCh38
rs962886450 Chr.7: 150332053-150332053 on Build GRCh38
rs1036439927 Chr.7: 150331929-150331929 on Build GRCh38
rs997857089 Chr.7: 150332074-150332074 on Build GRCh38
rs1002451949 Chr.7: 150331730-150331730 on Build GRCh38
rs993954738 Chr.7: 150331741-150331741 on Build GRCh38
rs1026787178 Chr.7: 150331752-150331752 on Build GRCh38
rs994564516 Chr.7: 150332057-150332057 on Build GRCh38
rs911832419 Chr.7: 150331880-150331880 on Build GRCh38
rs976536901 Chr.7: 150331868-150331868 on Build GRCh38
rs538891376 Chr.7: 150331917-150331917 on Build GRCh38
rs986027935 Chr.7: 150331879-150331879 on Build GRCh38
rs913729316 Chr.7: 150332132-150332132 on Build GRCh38
rs1002618067 Chr.7: 150331989-150331989 on Build GRCh38
rs1043777211 Chr.7: 150332143-150332143 on Build GRCh38
rs867762079 Chr.7: 150332099-150332099 on Build GRCh38
rs28549705 Chr.7: 150331898-150331898 on Build GRCh38

View Full Product Details