Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28107522-28108030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELP3
Entrez Gene ID
55140
Gene Name
elongator acetyltransferase complex subunit 3
Gene Aliases
KAT9
UniGene
Hs.491336
Gene Chromosome Location
Chr.8: 28090231-28191153 on Build GRCh38
Transcripts
NM_001284220.1 NM_001284222.1 NM_001284224.1 NM_001284225.1 NM_001284226.1 NM_018091.5 XM_006716354.2 XM_017013604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6234679 c.344G>T p.G115V Chr.8: 28107927-28107927 on Build GRCh38
3899362 c.370A>G p.T124A Chr.8: 28107953-28107953 on Build GRCh38
3647908 c.361G>A p.E121K Chr.8: 28107944-28107944 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs149788540 Chr.8: 28107586-28107586 on Build GRCh38
rs74652424 Chr.8: 28107607-28107607 on Build GRCh38
rs200017763 Chr.8: 28108000-28108000 on Build GRCh38
rs77315736 Chr.8: 28107822-28107822 on Build GRCh38
rs971126457 Chr.8: 28107853-28107853 on Build GRCh38
rs903085243 Chr.8: 28107649-28107649 on Build GRCh38
rs775459581 Chr.8: 28107909-28107909 on Build GRCh38
rs73573286 Chr.8: 28107583-28107583 on Build GRCh38
rs367881465 Chr.8: 28107887-28107887 on Build GRCh38
rs1034158891 Chr.8: 28107705-28107705 on Build GRCh38
rs567662706 Chr.8: 28107812-28107812 on Build GRCh38
rs961628085 Chr.8: 28107913-28107913 on Build GRCh38
rs1012637590 Chr.8: 28107785-28107785 on Build GRCh38
rs201540756 Chr.8: 28107972-28107972 on Build GRCh38
rs780558095 Chr.8: 28107579-28107579 on Build GRCh38
rs776036635 Chr.8: 28107640-28107640 on Build GRCh38
rs762361402 Chr.8: 28107865-28107865 on Build GRCh38
rs769360945 Chr.8: 28107740-28107740 on Build GRCh38
rs202229366 Chr.8: 28108006-28108006 on Build GRCh38
rs757389948 Chr.8: 28107964-28107964 on Build GRCh38
rs754409273 Chr.8: 28107946-28107946 on Build GRCh38
rs1032653686 Chr.8: 28107723-28107723 on Build GRCh38
rs958560000 Chr.8: 28107732-28107732 on Build GRCh38
rs759137175 Chr.8: 28107893-28107895 on Build GRCh38
rs548224424 Chr.8: 28107987-28107987 on Build GRCh38
rs775660848 Chr.8: 28107903-28107903 on Build GRCh38
rs865818748 Chr.8: 28107961-28107962 on Build GRCh38
rs774655514 Chr.8: 28107629-28107629 on Build GRCh38
rs764772545 Chr.8: 28107896-28107896 on Build GRCh38
rs73226778 Chr.8: 28107702-28107702 on Build GRCh38
rs190274080 Chr.8: 28107996-28107996 on Build GRCh38
rs758624669 Chr.8: 28107993-28107993 on Build GRCh38
rs780434553 Chr.8: 28107994-28107994 on Build GRCh38
rs201016610 Chr.8: 28107879-28107879 on Build GRCh38
rs764504706 Chr.8: 28107931-28107931 on Build GRCh38
rs761161057 Chr.8: 28107929-28107929 on Build GRCh38
rs774948039 Chr.8: 28107962-28107963 on Build GRCh38
rs770482049 Chr.8: 28107868-28107868 on Build GRCh38
rs531150568 Chr.8: 28107643-28107643 on Build GRCh38
rs1009944507 Chr.8: 28107588-28107588 on Build GRCh38
rs116319109 Chr.8: 28107601-28107601 on Build GRCh38

View Full Product Details