Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28342988-28343483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNOC
Entrez Gene ID
5368
Gene Name
prepronociceptin
Gene Aliases
N/OFQ, NOP, OFQ, PPNOC, ppN/OFQ
UniGene
Hs.88218
Gene Chromosome Location
Chr.8: 28316979-28343355 on Build GRCh38
Transcripts
NM_001284244.1 NM_006228.4 XM_005273532.2 XM_011544559.2 XM_017013578.1

SNP Detail

SNP ID Chromosome Location
rs954066019 Chr.8: 28343357-28343357 on Build GRCh38
rs187519507 Chr.8: 28343366-28343366 on Build GRCh38
rs912840041 Chr.8: 28343405-28343405 on Build GRCh38
rs1051617 Chr.8: 28343207-28343207 on Build GRCh38
rs144527098 Chr.8: 28343029-28343029 on Build GRCh38
rs541982547 Chr.8: 28343460-28343460 on Build GRCh38
rs542513553 Chr.8: 28343281-28343281 on Build GRCh38
rs185798961 Chr.8: 28343425-28343425 on Build GRCh38
rs895709224 Chr.8: 28343176-28343176 on Build GRCh38
rs8192510 Chr.8: 28343101-28343101 on Build GRCh38
rs902247456 Chr.8: 28343396-28343396 on Build GRCh38
rs545367108 Chr.8: 28343072-28343072 on Build GRCh38
rs558807680 Chr.8: 28343030-28343030 on Build GRCh38
rs944266883 Chr.8: 28343410-28343410 on Build GRCh38
rs1012259843 Chr.8: 28343157-28343157 on Build GRCh38
rs1042582260 Chr.8: 28343039-28343039 on Build GRCh38
rs76400483 Chr.8: 28343427-28343427 on Build GRCh38
rs544315480 Chr.8: 28343162-28343162 on Build GRCh38
rs1014995061 Chr.8: 28343209-28343209 on Build GRCh38
rs1004866019 Chr.8: 28343449-28343449 on Build GRCh38
rs763983806 Chr.8: 28343293-28343293 on Build GRCh38
rs1025354489 Chr.8: 28343159-28343159 on Build GRCh38
rs766170773 Chr.8: 28343398-28343399 on Build GRCh38
rs192435507 Chr.8: 28343414-28343414 on Build GRCh38
rs906555609 Chr.8: 28343446-28343446 on Build GRCh38
rs762604223 Chr.8: 28343227-28343227 on Build GRCh38
rs895391089 Chr.8: 28343149-28343149 on Build GRCh38
rs751767965 Chr.8: 28343383-28343383 on Build GRCh38
rs17058993 Chr.8: 28343168-28343168 on Build GRCh38
rs985146425 Chr.8: 28343284-28343284 on Build GRCh38
rs147418530 Chr.8: 28343424-28343424 on Build GRCh38
rs1001024104 Chr.8: 28343044-28343044 on Build GRCh38
rs1027031977 Chr.8: 28343305-28343305 on Build GRCh38
rs1032632620 Chr.8: 28343052-28343052 on Build GRCh38
rs182886402 Chr.8: 28343173-28343173 on Build GRCh38
rs905541406 Chr.8: 28343098-28343098 on Build GRCh38
rs775330918 Chr.8: 28343104-28343104 on Build GRCh38
rs529736293 Chr.8: 28343452-28343452 on Build GRCh38
rs961217818 Chr.8: 28343210-28343210 on Build GRCh38
rs113667304 Chr.8: 28343214-28343214 on Build GRCh38
rs558021172 Chr.8: 28343042-28343042 on Build GRCh38
rs376435171 Chr.8: 28343264-28343264 on Build GRCh38
rs559638783 Chr.8: 28343301-28343301 on Build GRCh38
rs376455837 Chr.8: 28343445-28343445 on Build GRCh38
rs1001607247 Chr.8: 28343161-28343161 on Build GRCh38
rs919878405 Chr.8: 28343251-28343251 on Build GRCh38
rs558604116 Chr.8: 28343097-28343097 on Build GRCh38
rs575367926 Chr.8: 28343151-28343151 on Build GRCh38
rs372094677 Chr.8: 28343183-28343183 on Build GRCh38
rs528254343 Chr.8: 28343330-28343330 on Build GRCh38
rs755380341 Chr.8: 28343426-28343426 on Build GRCh38
rs963180567 Chr.8: 28343244-28343244 on Build GRCh38

View Full Product Details