Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28447099-28447633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO16
Entrez Gene ID
157574
Gene Name
F-box protein 16
Gene Aliases
FBX16
UniGene
Hs.741522
Gene Chromosome Location
Chr.8: 28428408-28490318 on Build GRCh38
Transcripts
NM_001258211.1 NM_172366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6113172 c.821G>T p.R274M Chr.8: 28447193-28447193 on Build GRCh38
1472127 c.833C>T p.A278V Chr.8: 28447181-28447181 on Build GRCh38
2788586 c.838T>G p.S280A Chr.8: 28447176-28447176 on Build GRCh38
3763308 c.762G>A p.M254I Chr.8: 28447252-28447252 on Build GRCh38
6662082 c.840A>G p.S280S Chr.8: 28447174-28447174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569980765 Chr.8: 28447490-28447490 on Build GRCh38
rs1009317551 Chr.8: 28447286-28447286 on Build GRCh38
rs767425691 Chr.8: 28447157-28447157 on Build GRCh38
rs778491164 Chr.8: 28447224-28447224 on Build GRCh38
rs373655206 Chr.8: 28447233-28447233 on Build GRCh38
rs572587304 Chr.8: 28447561-28447561 on Build GRCh38
rs960362437 Chr.8: 28447428-28447428 on Build GRCh38
rs915436780 Chr.8: 28447370-28447370 on Build GRCh38
rs750695634 Chr.8: 28447150-28447150 on Build GRCh38
rs184935026 Chr.8: 28447460-28447460 on Build GRCh38
rs138441476 Chr.8: 28447168-28447168 on Build GRCh38
rs555698559 Chr.8: 28447186-28447186 on Build GRCh38
rs765603972 Chr.8: 28447149-28447149 on Build GRCh38
rs753744119 Chr.8: 28447301-28447301 on Build GRCh38
rs778405361 Chr.8: 28447321-28447321 on Build GRCh38
rs1050661108 Chr.8: 28447594-28447594 on Build GRCh38
rs564457153 Chr.8: 28447235-28447235 on Build GRCh38
rs760502143 Chr.8: 28447275-28447275 on Build GRCh38
rs192598873 Chr.8: 28447336-28447336 on Build GRCh38
rs144107749 Chr.8: 28447449-28447449 on Build GRCh38
rs756884862 Chr.8: 28447207-28447207 on Build GRCh38
rs769536194 Chr.8: 28447270-28447270 on Build GRCh38
rs562724571 Chr.8: 28447257-28447257 on Build GRCh38
rs201413261 Chr.8: 28447223-28447223 on Build GRCh38
rs576123750 Chr.8: 28447362-28447362 on Build GRCh38
rs149234031 Chr.8: 28447262-28447262 on Build GRCh38
rs1390963 Chr.8: 28447252-28447252 on Build GRCh38
rs535796041 Chr.8: 28447547-28447547 on Build GRCh38
rs759362503 Chr.8: 28447155-28447155 on Build GRCh38
rs140792800 Chr.8: 28447191-28447191 on Build GRCh38
rs993968833 Chr.8: 28447240-28447240 on Build GRCh38
rs188903503 Chr.8: 28447555-28447555 on Build GRCh38
rs539003224 Chr.8: 28447453-28447453 on Build GRCh38
rs779893708 Chr.8: 28447316-28447317 on Build GRCh38
rs376539326 Chr.8: 28447141-28447141 on Build GRCh38
rs187988580 Chr.8: 28447328-28447328 on Build GRCh38
rs541195992 Chr.8: 28447226-28447226 on Build GRCh38
rs756016710 Chr.8: 28447261-28447262 on Build GRCh38
rs555908982 Chr.8: 28447552-28447552 on Build GRCh38
rs981024906 Chr.8: 28447383-28447383 on Build GRCh38
rs956495228 Chr.8: 28447356-28447356 on Build GRCh38
rs79573885 Chr.8: 28447326-28447326 on Build GRCh38
rs1039517303 Chr.8: 28447576-28447576 on Build GRCh38
rs756134627 Chr.8: 28447173-28447173 on Build GRCh38
rs541505058 Chr.8: 28447575-28447575 on Build GRCh38
rs761909670 Chr.8: 28447146-28447146 on Build GRCh38
rs1022117465 Chr.8: 28447347-28447347 on Build GRCh38
rs939348424 Chr.8: 28447161-28447161 on Build GRCh38
rs767203849 Chr.8: 28447265-28447265 on Build GRCh38
rs1049607040 Chr.8: 28447530-28447530 on Build GRCh38
rs939755224 Chr.8: 28447465-28447465 on Build GRCh38
rs386723954 Chr.8: 28447250-28447252 on Build GRCh38
rs1056488742 Chr.8: 28447459-28447459 on Build GRCh38
rs768602046 Chr.8: 28447137-28447137 on Build GRCh38
rs750411020 Chr.8: 28447225-28447226 on Build GRCh38
rs929428005 Chr.8: 28447493-28447493 on Build GRCh38
rs920735760 Chr.8: 28447556-28447556 on Build GRCh38
rs568586909 Chr.8: 28447196-28447200 on Build GRCh38
rs756718099 Chr.8: 28447312-28447312 on Build GRCh38
rs764144544 Chr.8: 28447290-28447290 on Build GRCh38
rs776550852 Chr.8: 28447236-28447236 on Build GRCh38
rs547132875 Chr.8: 28447375-28447375 on Build GRCh38
rs745522032 Chr.8: 28447225-28447225 on Build GRCh38
rs888313525 Chr.8: 28447559-28447559 on Build GRCh38
rs988282471 Chr.8: 28447364-28447364 on Build GRCh38
rs926539006 Chr.8: 28447384-28447384 on Build GRCh38

View Full Product Details