Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 30105604-30106123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LEPROTL1
Entrez Gene ID
23484
Gene Name
leptin receptor overlapping transcript-like 1
Gene Aliases
HSPC112, Vps55, my047
UniGene
Hs.146585
Gene Chromosome Location
Chr.8: 30095406-30137706 on Build GRCh38
Transcripts
NM_001128208.1 NM_015344.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3899520 c.334A>T p.T112S Chr.8: 30105800-30105800 on Build GRCh38
1099069 c.316A>G p.N106D Chr.8: 30105782-30105782 on Build GRCh38
5952378 c.373G>T p.D125Y Chr.8: 30105839-30105839 on Build GRCh38
273782 c.286_287insG p.A98fs*27 Chr.8: 30105752-30105753 on Build GRCh38
1699930 c.338T>G p.I113R Chr.8: 30105804-30105804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749057013 Chr.8: 30105764-30105764 on Build GRCh38
rs908217155 Chr.8: 30106023-30106023 on Build GRCh38
rs568100342 Chr.8: 30106069-30106069 on Build GRCh38
rs768363991 Chr.8: 30105881-30105881 on Build GRCh38
rs367920230 Chr.8: 30105891-30105891 on Build GRCh38
rs891355680 Chr.8: 30105921-30105921 on Build GRCh38
rs374240229 Chr.8: 30105838-30105838 on Build GRCh38
rs974543255 Chr.8: 30105839-30105839 on Build GRCh38
rs201947181 Chr.8: 30105704-30105704 on Build GRCh38
rs896516529 Chr.8: 30105643-30105643 on Build GRCh38
rs113344745 Chr.8: 30106093-30106093 on Build GRCh38
rs1013593506 Chr.8: 30106009-30106009 on Build GRCh38
rs200526340 Chr.8: 30105906-30105906 on Build GRCh38
rs370989119 Chr.8: 30105723-30105723 on Build GRCh38
rs746681440 Chr.8: 30105866-30105866 on Build GRCh38
rs903922036 Chr.8: 30105993-30105993 on Build GRCh38
rs981081867 Chr.8: 30105674-30105674 on Build GRCh38
rs751823585 Chr.8: 30105735-30105735 on Build GRCh38
rs576013236 Chr.8: 30105631-30105631 on Build GRCh38
rs143499704 Chr.8: 30105834-30105834 on Build GRCh38
rs866642118 Chr.8: 30106063-30106063 on Build GRCh38
rs111836861 Chr.8: 30106014-30106014 on Build GRCh38
rs1011658436 Chr.8: 30105954-30105954 on Build GRCh38
rs368909987 Chr.8: 30105793-30105793 on Build GRCh38
rs371805932 Chr.8: 30105753-30105753 on Build GRCh38
rs144287209 Chr.8: 30105654-30105654 on Build GRCh38
rs886369392 Chr.8: 30106086-30106086 on Build GRCh38
rs377616301 Chr.8: 30105718-30105718 on Build GRCh38
rs13693 Chr.8: 30106007-30106007 on Build GRCh38
rs201372373 Chr.8: 30105705-30105705 on Build GRCh38
rs531685732 Chr.8: 30106002-30106002 on Build GRCh38
rs572098554 Chr.8: 30105657-30105657 on Build GRCh38
rs774841146 Chr.8: 30105723-30105724 on Build GRCh38
rs535456882 Chr.8: 30105682-30105683 on Build GRCh38
rs533268797 Chr.8: 30105884-30105884 on Build GRCh38
rs775327859 Chr.8: 30105695-30105695 on Build GRCh38
rs113932601 Chr.8: 30105728-30105728 on Build GRCh38
rs745765899 Chr.8: 30105837-30105837 on Build GRCh38
rs773439942 Chr.8: 30105906-30105907 on Build GRCh38
rs945511906 Chr.8: 30105944-30105944 on Build GRCh38
rs73669623 Chr.8: 30105919-30105919 on Build GRCh38
rs767855380 Chr.8: 30105906-30105906 on Build GRCh38
rs766425015 Chr.8: 30105729-30105729 on Build GRCh38
rs756994956 Chr.8: 30105808-30105808 on Build GRCh38
rs774421697 Chr.8: 30106003-30106003 on Build GRCh38
rs1043884885 Chr.8: 30105969-30105969 on Build GRCh38
rs762210637 Chr.8: 30105903-30105906 on Build GRCh38
rs765509050 Chr.8: 30105900-30105900 on Build GRCh38
rs921426286 Chr.8: 30105894-30105894 on Build GRCh38
rs927924910 Chr.8: 30105683-30105683 on Build GRCh38
rs1011669796 Chr.8: 30105644-30105644 on Build GRCh38
rs183942469 Chr.8: 30105628-30105628 on Build GRCh38
rs750891687 Chr.8: 30105727-30105727 on Build GRCh38
rs73669622 Chr.8: 30105694-30105694 on Build GRCh38
rs960924239 Chr.8: 30105693-30105693 on Build GRCh38
rs1053068499 Chr.8: 30105935-30105935 on Build GRCh38
rs760368684 Chr.8: 30105699-30105699 on Build GRCh38
rs1006036194 Chr.8: 30106091-30106091 on Build GRCh38
rs397960486 Chr.8: 30105693-30105694 on Build GRCh38
rs969281563 Chr.8: 30105660-30105660 on Build GRCh38
rs914159191 Chr.8: 30105942-30105942 on Build GRCh38
rs1026272883 Chr.8: 30105662-30105662 on Build GRCh38
rs962389245 Chr.8: 30106087-30106087 on Build GRCh38
rs999526806 Chr.8: 30105923-30105923 on Build GRCh38
rs775098803 Chr.8: 30105855-30105855 on Build GRCh38
rs1015992599 Chr.8: 30105791-30105791 on Build GRCh38
rs541922991 Chr.8: 30105639-30105639 on Build GRCh38
rs143574759 Chr.8: 30106089-30106089 on Build GRCh38
rs933034204 Chr.8: 30105917-30105917 on Build GRCh38
rs769208044 Chr.8: 30105699-30105700 on Build GRCh38
rs199590565 Chr.8: 30105776-30105776 on Build GRCh38

View Full Product Details