Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 30156214-30156716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN6
Entrez Gene ID
10671
Gene Name
dynactin subunit 6
Gene Aliases
WS-3, WS3, p27
UniGene
Hs.158427
Gene Chromosome Location
Chr.8: 30156297-30183639 on Build GRCh38
Transcripts
NM_006571.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3669893 c.6G>T p.A2A Chr.8: 30156389-30156389 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376288588 Chr.8: 30156324-30156324 on Build GRCh38
rs765991899 Chr.8: 30156357-30156357 on Build GRCh38
rs938598388 Chr.8: 30156671-30156671 on Build GRCh38
rs529484142 Chr.8: 30156476-30156476 on Build GRCh38
rs773874585 Chr.8: 30156359-30156359 on Build GRCh38
rs11305860 Chr.8: 30156688-30156688 on Build GRCh38
rs914557018 Chr.8: 30156641-30156641 on Build GRCh38
rs201876552 Chr.8: 30156432-30156432 on Build GRCh38
rs748897577 Chr.8: 30156443-30156443 on Build GRCh38
rs896044030 Chr.8: 30156689-30156689 on Build GRCh38
rs1018181646 Chr.8: 30156252-30156252 on Build GRCh38
rs962084002 Chr.8: 30156609-30156609 on Build GRCh38
rs754806061 Chr.8: 30156392-30156392 on Build GRCh38
rs560126915 Chr.8: 30156624-30156624 on Build GRCh38
rs774105467 Chr.8: 30156450-30156450 on Build GRCh38
rs925734589 Chr.8: 30156687-30156687 on Build GRCh38
rs896813960 Chr.8: 30156235-30156235 on Build GRCh38
rs770478787 Chr.8: 30156652-30156652 on Build GRCh38
rs567391022 Chr.8: 30156694-30156694 on Build GRCh38
rs914579720 Chr.8: 30156351-30156351 on Build GRCh38
rs1056417096 Chr.8: 30156523-30156523 on Build GRCh38
rs770335786 Chr.8: 30156448-30156448 on Build GRCh38
rs1048180066 Chr.8: 30156695-30156695 on Build GRCh38
rs7012749 Chr.8: 30156243-30156243 on Build GRCh38
rs918432736 Chr.8: 30156693-30156693 on Build GRCh38
rs772729244 Chr.8: 30156348-30156348 on Build GRCh38
rs904762424 Chr.8: 30156234-30156234 on Build GRCh38
rs779251461 Chr.8: 30156389-30156389 on Build GRCh38
rs757277107 Chr.8: 30156378-30156378 on Build GRCh38
rs769662886 Chr.8: 30156418-30156418 on Build GRCh38
rs980415885 Chr.8: 30156653-30156653 on Build GRCh38
rs966169357 Chr.8: 30156542-30156542 on Build GRCh38
rs948608741 Chr.8: 30156398-30156398 on Build GRCh38
rs367828651 Chr.8: 30156410-30156410 on Build GRCh38
rs199955013 Chr.8: 30156426-30156426 on Build GRCh38
rs976404793 Chr.8: 30156675-30156675 on Build GRCh38
rs1008222586 Chr.8: 30156503-30156503 on Build GRCh38
rs995142856 Chr.8: 30156236-30156236 on Build GRCh38
rs562181077 Chr.8: 30156276-30156276 on Build GRCh38
rs150067726 Chr.8: 30156616-30156616 on Build GRCh38
rs570951322 Chr.8: 30156690-30156690 on Build GRCh38
rs889494963 Chr.8: 30156497-30156497 on Build GRCh38
rs868166857 Chr.8: 30156269-30156269 on Build GRCh38
rs992797141 Chr.8: 30156257-30156257 on Build GRCh38
rs907017860 Chr.8: 30156400-30156400 on Build GRCh38
rs565444718 Chr.8: 30156612-30156612 on Build GRCh38
rs945810343 Chr.8: 30156420-30156420 on Build GRCh38
rs896072403 Chr.8: 30156527-30156527 on Build GRCh38
rs199722675 Chr.8: 30156413-30156413 on Build GRCh38
rs756636605 Chr.8: 30156371-30156371 on Build GRCh38
rs995257944 Chr.8: 30156464-30156464 on Build GRCh38
rs934756937 Chr.8: 30156280-30156280 on Build GRCh38
rs912927979 Chr.8: 30156677-30156677 on Build GRCh38
rs748106898 Chr.8: 30156415-30156415 on Build GRCh38
rs549519501 Chr.8: 30156534-30156534 on Build GRCh38
rs960350559 Chr.8: 30156253-30156253 on Build GRCh38
rs759945668 Chr.8: 30156692-30156692 on Build GRCh38
rs759237655 Chr.8: 30156458-30156458 on Build GRCh38
rs758915963 Chr.8: 30156272-30156272 on Build GRCh38
rs200411897 Chr.8: 30156664-30156665 on Build GRCh38
rs764278794 Chr.8: 30156373-30156373 on Build GRCh38
rs1036827682 Chr.8: 30156687-30156687 on Build GRCh38
rs542780903 Chr.8: 30156267-30156267 on Build GRCh38
rs572532819 Chr.8: 30156277-30156277 on Build GRCh38
rs541440684 Chr.8: 30156278-30156278 on Build GRCh38
rs1044361405 Chr.8: 30156399-30156399 on Build GRCh38
rs976190891 Chr.8: 30156547-30156547 on Build GRCh38
rs746053546 Chr.8: 30156390-30156390 on Build GRCh38
rs533355540 Chr.8: 30156296-30156296 on Build GRCh38
rs945777811 Chr.8: 30156679-30156679 on Build GRCh38
rs972526123 Chr.8: 30156305-30156305 on Build GRCh38
rs564521121 Chr.8: 30156291-30156291 on Build GRCh38
rs1017879164 Chr.8: 30156540-30156540 on Build GRCh38
rs896884007 Chr.8: 30156460-30156460 on Build GRCh38
rs551057827 Chr.8: 30156665-30156665 on Build GRCh38
rs753886811 Chr.8: 30156374-30156374 on Build GRCh38
rs1047295 Chr.8: 30156319-30156319 on Build GRCh38
rs1014906328 Chr.8: 30156600-30156600 on Build GRCh38
rs1043232764 Chr.8: 30156683-30156683 on Build GRCh38
rs1014180918 Chr.8: 30156575-30156575 on Build GRCh38
rs952358485 Chr.8: 30156246-30156246 on Build GRCh38
rs1047296 Chr.8: 30156360-30156360 on Build GRCh38
rs753300610 Chr.8: 30156364-30156364 on Build GRCh38
rs762528872 Chr.8: 30156356-30156356 on Build GRCh38
rs1031817377 Chr.8: 30156572-30156572 on Build GRCh38
rs370007794 Chr.8: 30156339-30156339 on Build GRCh38
rs986407579 Chr.8: 30156247-30156247 on Build GRCh38

View Full Product Details