Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38143254-38143777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAR
Entrez Gene ID
6770
Gene Name
steroidogenic acute regulatory protein
Gene Aliases
STARD1
UniGene
Hs.521535
Gene Chromosome Location
Chr.8: 38141747-38151265 on Build GRCh38
Transcripts
NM_000349.2 XM_006716392.2

SNP Detail

SNP ID Chromosome Location
rs886062903 Chr.8: 38143717-38143717 on Build GRCh38
rs545393727 Chr.8: 38143351-38143351 on Build GRCh38
rs573359224 Chr.8: 38143629-38143629 on Build GRCh38
rs769133584 Chr.8: 38143305-38143306 on Build GRCh38
rs1024148648 Chr.8: 38143711-38143711 on Build GRCh38
rs35514257 Chr.8: 38143547-38143547 on Build GRCh38
rs765198172 Chr.8: 38143607-38143607 on Build GRCh38
rs566567853 Chr.8: 38143389-38143389 on Build GRCh38
rs540152389 Chr.8: 38143574-38143574 on Build GRCh38
rs971312799 Chr.8: 38143372-38143372 on Build GRCh38
rs985513350 Chr.8: 38143549-38143549 on Build GRCh38
rs1047513157 Chr.8: 38143730-38143730 on Build GRCh38
rs922649364 Chr.8: 38143545-38143545 on Build GRCh38
rs886062900 Chr.8: 38143325-38143325 on Build GRCh38
rs1037579047 Chr.8: 38143426-38143426 on Build GRCh38
rs1033910984 Chr.8: 38143282-38143282 on Build GRCh38
rs955564824 Chr.8: 38143548-38143548 on Build GRCh38
rs11326306 Chr.8: 38143324-38143324 on Build GRCh38
rs182316515 Chr.8: 38143399-38143399 on Build GRCh38
rs959681660 Chr.8: 38143477-38143477 on Build GRCh38
rs1004716306 Chr.8: 38143352-38143352 on Build GRCh38
rs944976686 Chr.8: 38143553-38143553 on Build GRCh38
rs898981799 Chr.8: 38143435-38143435 on Build GRCh38
rs886062899 Chr.8: 38143306-38143306 on Build GRCh38
rs548810270 Chr.8: 38143393-38143393 on Build GRCh38
rs886062902 Chr.8: 38143477-38143477 on Build GRCh38
rs558498679 Chr.8: 38143455-38143455 on Build GRCh38
rs973304084 Chr.8: 38143576-38143576 on Build GRCh38
rs907012858 Chr.8: 38143714-38143716 on Build GRCh38
rs906211970 Chr.8: 38143457-38143457 on Build GRCh38
rs188232971 Chr.8: 38143292-38143292 on Build GRCh38
rs978160297 Chr.8: 38143527-38143527 on Build GRCh38
rs1049185010 Chr.8: 38143621-38143621 on Build GRCh38
rs957097988 Chr.8: 38143751-38143751 on Build GRCh38
rs774771797 Chr.8: 38143306-38143308 on Build GRCh38
rs978127643 Chr.8: 38143359-38143359 on Build GRCh38
rs886062901 Chr.8: 38143456-38143456 on Build GRCh38
rs758226281 Chr.8: 38143753-38143753 on Build GRCh38
rs966836158 Chr.8: 38143522-38143522 on Build GRCh38
rs569540827 Chr.8: 38143444-38143444 on Build GRCh38
rs1022441621 Chr.8: 38143496-38143496 on Build GRCh38
rs190975625 Chr.8: 38143398-38143398 on Build GRCh38
rs747926723 Chr.8: 38143308-38143308 on Build GRCh38
rs752456740 Chr.8: 38143687-38143687 on Build GRCh38
rs560285321 Chr.8: 38143506-38143506 on Build GRCh38
rs906245398 Chr.8: 38143738-38143738 on Build GRCh38
rs1001468266 Chr.8: 38143719-38143719 on Build GRCh38
rs747092957 Chr.8: 38143581-38143581 on Build GRCh38
rs527342015 Chr.8: 38143392-38143392 on Build GRCh38
rs1034353358 Chr.8: 38143474-38143474 on Build GRCh38
rs945580576 Chr.8: 38143475-38143475 on Build GRCh38
rs915400542 Chr.8: 38143343-38143343 on Build GRCh38
rs533085477 Chr.8: 38143331-38143331 on Build GRCh38
rs551135472 Chr.8: 38143360-38143360 on Build GRCh38
rs943273677 Chr.8: 38143425-38143425 on Build GRCh38
rs979978784 Chr.8: 38143373-38143373 on Build GRCh38
rs989946911 Chr.8: 38143440-38143440 on Build GRCh38
rs556220711 Chr.8: 38143442-38143442 on Build GRCh38
rs929102876 Chr.8: 38143699-38143699 on Build GRCh38
rs957234746 Chr.8: 38143395-38143395 on Build GRCh38
rs558290486 Chr.8: 38143575-38143575 on Build GRCh38
rs966804852 Chr.8: 38143345-38143345 on Build GRCh38
rs200095542 Chr.8: 38143316-38143317 on Build GRCh38
rs777417661 Chr.8: 38143307-38143308 on Build GRCh38
rs28361934 Chr.8: 38143286-38143286 on Build GRCh38
rs1003169711 Chr.8: 38143465-38143465 on Build GRCh38
rs930620623 Chr.8: 38143592-38143592 on Build GRCh38
rs1011571206 Chr.8: 38143495-38143495 on Build GRCh38

View Full Product Details