Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70926387-70926919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC7A3
Entrez Gene ID
84889
Gene Name
solute carrier family 7 member 3
Gene Aliases
ATRC3, CAT-3, CAT3
UniGene
Hs.175220
Gene Chromosome Location
Chr.X: 70925579-70931125 on Build GRCh38
Transcripts
NM_001048164.2 NM_032803.5 XM_017029912.1

SNP Detail

SNP ID Chromosome Location
rs990360390 Chr.X: 70926581-70926581 on Build GRCh38
rs753230802 Chr.X: 70926612-70926612 on Build GRCh38
rs780709311 Chr.X: 70926640-70926640 on Build GRCh38
rs866104948 Chr.X: 70926672-70926672 on Build GRCh38
rs751557831 Chr.X: 70926767-70926767 on Build GRCh38
rs780964394 Chr.X: 70926881-70926881 on Build GRCh38
rs779673693 Chr.X: 70926787-70926787 on Build GRCh38
rs760915001 Chr.X: 70926563-70926563 on Build GRCh38
rs896556505 Chr.X: 70926663-70926663 on Build GRCh38
rs868567946 Chr.X: 70926538-70926538 on Build GRCh38
rs200530998 Chr.X: 70926743-70926743 on Build GRCh38
rs775215692 Chr.X: 70926503-70926503 on Build GRCh38
rs370197571 Chr.X: 70926851-70926851 on Build GRCh38
rs760988627 Chr.X: 70926458-70926458 on Build GRCh38
rs767931807 Chr.X: 70926590-70926590 on Build GRCh38
rs374503535 Chr.X: 70926717-70926717 on Build GRCh38
rs750024372 Chr.X: 70926845-70926845 on Build GRCh38
rs377660999 Chr.X: 70926511-70926511 on Build GRCh38
rs760025171 Chr.X: 70926869-70926869 on Build GRCh38
rs935295950 Chr.X: 70926604-70926604 on Build GRCh38
rs767613894 Chr.X: 70926557-70926557 on Build GRCh38
rs1043929675 Chr.X: 70926573-70926573 on Build GRCh38
rs768511468 Chr.X: 70926524-70926524 on Build GRCh38
rs182088306 Chr.X: 70926784-70926784 on Build GRCh38
rs925506167 Chr.X: 70926853-70926853 on Build GRCh38
rs745498618 Chr.X: 70926493-70926493 on Build GRCh38
rs769507776 Chr.X: 70926498-70926498 on Build GRCh38
rs948902263 Chr.X: 70926644-70926644 on Build GRCh38
rs767075284 Chr.X: 70926811-70926811 on Build GRCh38
rs377561132 Chr.X: 70926834-70926834 on Build GRCh38
rs975878404 Chr.X: 70926823-70926823 on Build GRCh38
rs1013711538 Chr.X: 70926698-70926698 on Build GRCh38
rs762900338 Chr.X: 70926611-70926611 on Build GRCh38
rs755732942 Chr.X: 70926667-70926667 on Build GRCh38
rs749146450 Chr.X: 70926718-70926718 on Build GRCh38
rs6525447 Chr.X: 70926625-70926625 on Build GRCh38
rs369829723 Chr.X: 70926577-70926577 on Build GRCh38
rs981957730 Chr.X: 70926686-70926686 on Build GRCh38
rs750652204 Chr.X: 70926561-70926561 on Build GRCh38
rs745528774 Chr.X: 70926645-70926645 on Build GRCh38
rs189857869 Chr.X: 70926764-70926764 on Build GRCh38
rs4360450 Chr.X: 70926548-70926548 on Build GRCh38
rs774322976 Chr.X: 70926742-70926742 on Build GRCh38
rs976647122 Chr.X: 70926420-70926420 on Build GRCh38
rs865857295 Chr.X: 70926588-70926588 on Build GRCh38
rs774417127 Chr.X: 70926441-70926441 on Build GRCh38
rs930585312 Chr.X: 70926813-70926813 on Build GRCh38
rs761858509 Chr.X: 70926550-70926550 on Build GRCh38
rs755771269 Chr.X: 70926895-70926895 on Build GRCh38
rs916114011 Chr.X: 70926592-70926592 on Build GRCh38
rs185787766 Chr.X: 70926635-70926635 on Build GRCh38
rs768429002 Chr.X: 70926724-70926724 on Build GRCh38
rs766875762 Chr.X: 70926576-70926576 on Build GRCh38
rs772076861 Chr.X: 70926824-70926824 on Build GRCh38
rs1024540643 Chr.X: 70926714-70926714 on Build GRCh38
rs901602168 Chr.X: 70926863-70926863 on Build GRCh38
rs199939808 Chr.X: 70926835-70926835 on Build GRCh38
rs753171914 Chr.X: 70926888-70926888 on Build GRCh38

View Full Product Details