Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71580417-71580941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACRC
Entrez Gene ID
93953
Gene Name
acidic repeat containing
Gene Aliases
NAAR1
UniGene
Hs.135167
Gene Chromosome Location
Chr.X: 71578024-71613583 on Build GRCh38
Transcripts
NM_052957.4 XM_006724716.3 XM_011531064.1 XM_017029965.1

SNP Detail

SNP ID Chromosome Location
rs956685265 Chr.X: 71580839-71580839 on Build GRCh38
rs1028485540 Chr.X: 71580464-71580464 on Build GRCh38
rs767425296 Chr.X: 71580780-71580781 on Build GRCh38
rs1046365121 Chr.X: 71580805-71580805 on Build GRCh38
rs1008460057 Chr.X: 71580492-71580492 on Build GRCh38
rs1052570590 Chr.X: 71580471-71580471 on Build GRCh38
rs373105527 Chr.X: 71580725-71580725 on Build GRCh38
rs138602001 Chr.X: 71580861-71580861 on Build GRCh38
rs979293849 Chr.X: 71580820-71580820 on Build GRCh38
rs113849620 Chr.X: 71580650-71580650 on Build GRCh38
rs892955564 Chr.X: 71580583-71580583 on Build GRCh38
rs773088638 Chr.X: 71580773-71580773 on Build GRCh38
rs760714525 Chr.X: 71580774-71580774 on Build GRCh38
rs995639183 Chr.X: 71580631-71580631 on Build GRCh38
rs995690661 Chr.X: 71580448-71580448 on Build GRCh38
rs765038011 Chr.X: 71580469-71580469 on Build GRCh38
rs781748302 Chr.X: 71580869-71580869 on Build GRCh38
rs1045305191 Chr.X: 71580557-71580557 on Build GRCh38
rs773227640 Chr.X: 71580798-71580799 on Build GRCh38
rs112973179 Chr.X: 71580766-71580766 on Build GRCh38
rs12834159 Chr.X: 71580860-71580860 on Build GRCh38
rs781340339 Chr.X: 71580792-71580792 on Build GRCh38
rs757531858 Chr.X: 71580867-71580867 on Build GRCh38
rs752919557 Chr.X: 71580834-71580834 on Build GRCh38
rs997409283 Chr.X: 71580751-71580751 on Build GRCh38
rs758230172 Chr.X: 71580540-71580540 on Build GRCh38
rs756664321 Chr.X: 71580863-71580863 on Build GRCh38
rs370258215 Chr.X: 71580899-71580899 on Build GRCh38
rs377384243 Chr.X: 71580858-71580858 on Build GRCh38
rs1004196715 Chr.X: 71580716-71580716 on Build GRCh38
rs949669690 Chr.X: 71580544-71580544 on Build GRCh38
rs746349614 Chr.X: 71580870-71580870 on Build GRCh38
rs1020062208 Chr.X: 71580717-71580717 on Build GRCh38
rs7050103 Chr.X: 71580453-71580453 on Build GRCh38
rs990567372 Chr.X: 71580682-71580682 on Build GRCh38
rs377049000 Chr.X: 71580872-71580872 on Build GRCh38
rs905348339 Chr.X: 71580568-71580568 on Build GRCh38
rs760682963 Chr.X: 71580802-71580803 on Build GRCh38
rs753694751 Chr.X: 71580793-71580793 on Build GRCh38
rs949187618 Chr.X: 71580802-71580802 on Build GRCh38
rs957988262 Chr.X: 71580592-71580592 on Build GRCh38
rs1048463978 Chr.X: 71580664-71580664 on Build GRCh38
rs1019808758 Chr.X: 71580505-71580505 on Build GRCh38
rs966423402 Chr.X: 71580525-71580526 on Build GRCh38
rs778502805 Chr.X: 71580897-71580897 on Build GRCh38
rs925066428 Chr.X: 71580570-71580570 on Build GRCh38
rs750249737 Chr.X: 71580538-71580538 on Build GRCh38
rs373420187 Chr.X: 71580850-71580850 on Build GRCh38
rs916451686 Chr.X: 71580787-71580787 on Build GRCh38
rs887227551 Chr.X: 71580676-71580676 on Build GRCh38
rs953158606 Chr.X: 71580801-71580801 on Build GRCh38
rs73552708 Chr.X: 71580832-71580832 on Build GRCh38
rs759716604 Chr.X: 71580800-71580800 on Build GRCh38
rs761825295 Chr.X: 71580446-71580446 on Build GRCh38
rs766144259 Chr.X: 71580619-71580619 on Build GRCh38

View Full Product Details