Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 80438518-80439024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM46D
Entrez Gene ID
169966
Gene Name
family with sequence similarity 46 member D
Gene Aliases
CT1.26, CT112
UniGene
Hs.367959
Gene Chromosome Location
Chr.X: 80335504-80445311 on Build GRCh38
Transcripts
NM_001170574.1 NM_152630.4

SNP Detail

SNP ID Chromosome Location
rs964963430 Chr.X: 80438921-80438921 on Build GRCh38
rs1027312238 Chr.X: 80438971-80438971 on Build GRCh38
rs894914240 Chr.X: 80438593-80438593 on Build GRCh38
rs1024603090 Chr.X: 80438729-80438729 on Build GRCh38
rs989360721 Chr.X: 80438923-80438923 on Build GRCh38
rs757345366 Chr.X: 80438573-80438573 on Build GRCh38
rs778123333 Chr.X: 80438965-80438965 on Build GRCh38
rs902616948 Chr.X: 80438733-80438733 on Build GRCh38
rs999550951 Chr.X: 80438796-80438796 on Build GRCh38
rs754558480 Chr.X: 80438804-80438804 on Build GRCh38
rs1044092321 Chr.X: 80438697-80438697 on Build GRCh38
rs950471598 Chr.X: 80438985-80438985 on Build GRCh38
rs1011835567 Chr.X: 80438607-80438607 on Build GRCh38
rs946739258 Chr.X: 80438640-80438642 on Build GRCh38
rs890280969 Chr.X: 80438872-80438872 on Build GRCh38
rs970578114 Chr.X: 80438737-80438737 on Build GRCh38
rs112062503 Chr.X: 80438579-80438579 on Build GRCh38
rs1017526313 Chr.X: 80438904-80438904 on Build GRCh38
rs6652954 Chr.X: 80438901-80438901 on Build GRCh38
rs1052925429 Chr.X: 80438853-80438853 on Build GRCh38
rs749282131 Chr.X: 80438827-80438827 on Build GRCh38
rs182296069 Chr.X: 80438622-80438622 on Build GRCh38
rs1009114811 Chr.X: 80438888-80438888 on Build GRCh38
rs1001072442 Chr.X: 80438540-80438540 on Build GRCh38
rs994488376 Chr.X: 80438777-80438777 on Build GRCh38

View Full Product Details