Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 84062274-84062793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS6KA6
Entrez Gene ID
27330
Gene Name
ribosomal protein S6 kinase A6
Gene Aliases
PP90RSK4, RSK4
UniGene
Hs.234478
Gene Chromosome Location
Chr.X: 84058346-84187935 on Build GRCh38
Transcripts
NM_014496.4 XM_017029423.1

SNP Detail

SNP ID Chromosome Location
rs759138606 Chr.X: 84062597-84062597 on Build GRCh38
rs752114871 Chr.X: 84062631-84062634 on Build GRCh38
rs919437914 Chr.X: 84062321-84062321 on Build GRCh38
rs1033088405 Chr.X: 84062618-84062618 on Build GRCh38
rs771907941 Chr.X: 84062335-84062336 on Build GRCh38
rs917288609 Chr.X: 84062665-84062665 on Build GRCh38
rs1007995644 Chr.X: 84062648-84062648 on Build GRCh38
rs760102068 Chr.X: 84062683-84062683 on Build GRCh38
rs888105542 Chr.X: 84062376-84062376 on Build GRCh38
rs180933416 Chr.X: 84062491-84062491 on Build GRCh38
rs773446191 Chr.X: 84062318-84062318 on Build GRCh38
rs189789582 Chr.X: 84062408-84062408 on Build GRCh38
rs1045508097 Chr.X: 84062546-84062546 on Build GRCh38
rs950141501 Chr.X: 84062733-84062733 on Build GRCh38
rs1017622473 Chr.X: 84062691-84062691 on Build GRCh38
rs892027211 Chr.X: 84062630-84062630 on Build GRCh38
rs913567418 Chr.X: 84062457-84062457 on Build GRCh38
rs979328129 Chr.X: 84062543-84062543 on Build GRCh38
rs184893031 Chr.X: 84062370-84062370 on Build GRCh38
rs5922906 Chr.X: 84062587-84062587 on Build GRCh38
rs957380552 Chr.X: 84062639-84062639 on Build GRCh38
rs903025799 Chr.X: 84062597-84062598 on Build GRCh38
rs766325269 Chr.X: 84062684-84062684 on Build GRCh38
rs976223427 Chr.X: 84062305-84062305 on Build GRCh38
rs965836760 Chr.X: 84062406-84062406 on Build GRCh38
rs757635652 Chr.X: 84062744-84062744 on Build GRCh38
rs1019088514 Chr.X: 84062331-84062331 on Build GRCh38
rs894362142 Chr.X: 84062423-84062423 on Build GRCh38
rs764654848 Chr.X: 84062603-84062603 on Build GRCh38
rs1036446580 Chr.X: 84062743-84062743 on Build GRCh38
rs749660987 Chr.X: 84062480-84062480 on Build GRCh38
rs998892448 Chr.X: 84062614-84062614 on Build GRCh38
rs16979793 Chr.X: 84062564-84062564 on Build GRCh38
rs1046887174 Chr.X: 84062345-84062345 on Build GRCh38
rs937817461 Chr.X: 84062581-84062581 on Build GRCh38
rs1014186387 Chr.X: 84062468-84062468 on Build GRCh38

View Full Product Details