Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 85279318-85279830 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POF1B
Entrez Gene ID
79983
Gene Name
premature ovarian failure, 1B
Gene Aliases
POF, POF2B
UniGene
Hs.267038
Gene Chromosome Location
Chr.X: 85277389-85379743 on Build GRCh38
Transcripts
NM_024921.3 XM_005262203.3

SNP Detail

SNP ID Chromosome Location
rs1020031451 Chr.X: 85279363-85279363 on Build GRCh38
rs764597122 Chr.X: 85279466-85279469 on Build GRCh38
rs934412667 Chr.X: 85279491-85279491 on Build GRCh38
rs773134028 Chr.X: 85279756-85279756 on Build GRCh38
rs759722764 Chr.X: 85279428-85279429 on Build GRCh38
rs779587305 Chr.X: 85279370-85279370 on Build GRCh38
rs767407495 Chr.X: 85279389-85279389 on Build GRCh38
rs769929846 Chr.X: 85279505-85279505 on Build GRCh38
rs551418925 Chr.X: 85279541-85279541 on Build GRCh38
rs910834044 Chr.X: 85279565-85279566 on Build GRCh38
rs772899957 Chr.X: 85279404-85279404 on Build GRCh38
rs993996751 Chr.X: 85279726-85279726 on Build GRCh38
rs73513635 Chr.X: 85279613-85279613 on Build GRCh38
rs373475107 Chr.X: 85279436-85279436 on Build GRCh38
rs73627349 Chr.X: 85279780-85279780 on Build GRCh38
rs897880418 Chr.X: 85279703-85279703 on Build GRCh38
rs748816246 Chr.X: 85279384-85279384 on Build GRCh38
rs372539151 Chr.X: 85279379-85279379 on Build GRCh38
rs892837108 Chr.X: 85279531-85279531 on Build GRCh38
rs761595726 Chr.X: 85279387-85279387 on Build GRCh38
rs181248568 Chr.X: 85279803-85279803 on Build GRCh38
rs749132991 Chr.X: 85279766-85279766 on Build GRCh38
rs1040582321 Chr.X: 85279700-85279700 on Build GRCh38
rs747104592 Chr.X: 85279375-85279376 on Build GRCh38
rs905344907 Chr.X: 85279627-85279627 on Build GRCh38
rs768784756 Chr.X: 85279376-85279376 on Build GRCh38
rs753921003 Chr.X: 85279430-85279430 on Build GRCh38
rs776879430 Chr.X: 85279377-85279378 on Build GRCh38
rs762909156 Chr.X: 85279376-85279379 on Build GRCh38
rs765619812 Chr.X: 85279435-85279436 on Build GRCh38
rs781043288 Chr.X: 85279476-85279477 on Build GRCh38
rs758862317 Chr.X: 85279378-85279378 on Build GRCh38
rs1015601116 Chr.X: 85279800-85279800 on Build GRCh38
rs2227090 Chr.X: 85279377-85279377 on Build GRCh38
rs748327638 Chr.X: 85279382-85279382 on Build GRCh38
rs768556295 Chr.X: 85279386-85279386 on Build GRCh38
rs363789 Chr.X: 85279477-85279479 on Build GRCh38
rs944836318 Chr.X: 85279567-85279567 on Build GRCh38
rs1050656138 Chr.X: 85279565-85279565 on Build GRCh38
rs759741335 Chr.X: 85279461-85279461 on Build GRCh38
rs1049018262 Chr.X: 85279751-85279751 on Build GRCh38
rs973973843 Chr.X: 85279804-85279804 on Build GRCh38
rs766542303 Chr.X: 85279417-85279417 on Build GRCh38
rs763383566 Chr.X: 85279436-85279438 on Build GRCh38
rs879249530 Chr.X: 85279484-85279484 on Build GRCh38
rs796394268 Chr.X: 85279376-85279377 on Build GRCh38
rs749297111 Chr.X: 85279748-85279748 on Build GRCh38
rs142391747 Chr.X: 85279801-85279801 on Build GRCh38
rs112147894 Chr.X: 85279376-85279377 on Build GRCh38
rs935718249 Chr.X: 85279504-85279504 on Build GRCh38
rs1011109380 Chr.X: 85279587-85279587 on Build GRCh38
rs377237007 Chr.X: 85279408-85279408 on Build GRCh38
rs769331961 Chr.X: 85279381-85279381 on Build GRCh38
rs1002762236 Chr.X: 85279696-85279696 on Build GRCh38

View Full Product Details