Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 93673049-93673585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAP1L3
Entrez Gene ID
4675
Gene Name
nucleosome assembly protein 1 like 3
Gene Aliases
MB20, NPL3
UniGene
Hs.21365
Gene Chromosome Location
Chr.X: 93670926-93673683 on Build GRCh38
Transcripts
NM_004538.5

SNP Detail

SNP ID Chromosome Location
rs369510120 Chr.X: 93673192-93673192 on Build GRCh38
rs745323704 Chr.X: 93673094-93673094 on Build GRCh38
rs201582641 Chr.X: 93673224-93673224 on Build GRCh38
rs1008801684 Chr.X: 93673544-93673544 on Build GRCh38
rs772389834 Chr.X: 93673153-93673154 on Build GRCh38
rs62641622 Chr.X: 93673115-93673115 on Build GRCh38
rs190447192 Chr.X: 93673136-93673136 on Build GRCh38
rs775226866 Chr.X: 93673100-93673100 on Build GRCh38
rs149908317 Chr.X: 93673081-93673081 on Build GRCh38
rs144990323 Chr.X: 93673082-93673082 on Build GRCh38
rs746672924 Chr.X: 93673124-93673125 on Build GRCh38
rs758655977 Chr.X: 93673124-93673124 on Build GRCh38
rs372945820 Chr.X: 93673324-93673324 on Build GRCh38
rs148673858 Chr.X: 93673174-93673174 on Build GRCh38
rs777443854 Chr.X: 93673125-93673130 on Build GRCh38
rs769786203 Chr.X: 93673099-93673099 on Build GRCh38
rs760945470 Chr.X: 93673276-93673276 on Build GRCh38
rs566146584 Chr.X: 93673438-93673438 on Build GRCh38
rs760001145 Chr.X: 93673316-93673316 on Build GRCh38
rs770684313 Chr.X: 93673085-93673085 on Build GRCh38
rs756086573 Chr.X: 93673126-93673126 on Build GRCh38
rs750104084 Chr.X: 93673348-93673348 on Build GRCh38
rs1043456626 Chr.X: 93673148-93673174 on Build GRCh38
rs1004047936 Chr.X: 93673188-93673188 on Build GRCh38
rs762058020 Chr.X: 93673113-93673113 on Build GRCh38
rs943786071 Chr.X: 93673546-93673546 on Build GRCh38
rs759440847 Chr.X: 93673102-93673103 on Build GRCh38
rs1016128221 Chr.X: 93673238-93673238 on Build GRCh38
rs959339031 Chr.X: 93673413-93673413 on Build GRCh38
rs767682634 Chr.X: 93673114-93673114 on Build GRCh38
rs954782413 Chr.X: 93673516-93673516 on Build GRCh38
rs745751828 Chr.X: 93673137-93673163 on Build GRCh38
rs754317513 Chr.X: 93673115-93673116 on Build GRCh38
rs967312822 Chr.X: 93673460-93673460 on Build GRCh38
rs62635803 Chr.X: 93673108-93673108 on Build GRCh38
rs762061463 Chr.X: 93673109-93673110 on Build GRCh38
rs753931874 Chr.X: 93673312-93673312 on Build GRCh38
rs772293013 Chr.X: 93673274-93673274 on Build GRCh38
rs756992591 Chr.X: 93673172-93673172 on Build GRCh38
rs768843887 Chr.X: 93673110-93673115 on Build GRCh38
rs748285907 Chr.X: 93673154-93673156 on Build GRCh38
rs924380754 Chr.X: 93673541-93673541 on Build GRCh38
rs973091496 Chr.X: 93673150-93673150 on Build GRCh38
rs990139752 Chr.X: 93673422-93673422 on Build GRCh38
rs757028815 Chr.X: 93673127-93673128 on Build GRCh38
rs192311076 Chr.X: 93673107-93673107 on Build GRCh38
rs182641612 Chr.X: 93673347-93673347 on Build GRCh38
rs776931563 Chr.X: 93673214-93673214 on Build GRCh38
rs764725592 Chr.X: 93673118-93673119 on Build GRCh38
rs1045684 Chr.X: 93673378-93673378 on Build GRCh38
rs769918522 Chr.X: 93673136-93673137 on Build GRCh38
rs762947325 Chr.X: 93673105-93673105 on Build GRCh38
rs1029461447 Chr.X: 93673410-93673410 on Build GRCh38
rs766425655 Chr.X: 93673305-93673305 on Build GRCh38
rs779414158 Chr.X: 93673074-93673074 on Build GRCh38
rs761134785 Chr.X: 93673113-93673136 on Build GRCh38
rs765858439 Chr.X: 93673122-93673122 on Build GRCh38
rs767867439 Chr.X: 93673110-93673118 on Build GRCh38
rs753291433 Chr.X: 93673123-93673123 on Build GRCh38
rs377311138 Chr.X: 93673340-93673340 on Build GRCh38
rs770732139 Chr.X: 93673088-93673089 on Build GRCh38
rs996566783 Chr.X: 93673441-93673442 on Build GRCh38
rs946206396 Chr.X: 93673353-93673353 on Build GRCh38
rs920361625 Chr.X: 93673152-93673153 on Build GRCh38
rs749236135 Chr.X: 93673231-93673231 on Build GRCh38
rs778694008 Chr.X: 93673447-93673447 on Build GRCh38
rs377497549 Chr.X: 93673159-93673159 on Build GRCh38
rs748157767 Chr.X: 93673469-93673469 on Build GRCh38
rs761543067 Chr.X: 93673254-93673254 on Build GRCh38
rs932297382 Chr.X: 93673554-93673554 on Build GRCh38
rs774558833 Chr.X: 93673137-93673142 on Build GRCh38
rs776598582 Chr.X: 93673103-93673123 on Build GRCh38
rs750030880 Chr.X: 93673142-93673142 on Build GRCh38
rs753126678 Chr.X: 93673322-93673322 on Build GRCh38
rs1029296098 Chr.X: 93673102-93673102 on Build GRCh38
rs758027449 Chr.X: 93673121-93673122 on Build GRCh38
rs768554262 Chr.X: 93673241-93673241 on Build GRCh38
rs766911140 Chr.X: 93673130-93673130 on Build GRCh38
rs768533905 Chr.X: 93673434-93673434 on Build GRCh38
rs760078060 Chr.X: 93673116-93673136 on Build GRCh38
rs774640663 Chr.X: 93673110-93673112 on Build GRCh38
rs776311937 Chr.X: 93673088-93673088 on Build GRCh38
rs755775133 Chr.X: 93673183-93673183 on Build GRCh38
rs750521102 Chr.X: 93673121-93673121 on Build GRCh38
rs752265568 Chr.X: 93673119-93673136 on Build GRCh38
rs1043510426 Chr.X: 93673379-93673379 on Build GRCh38
rs765790824 Chr.X: 93673317-93673317 on Build GRCh38
rs1040786721 Chr.X: 93673548-93673548 on Build GRCh38
rs920940068 Chr.X: 93673549-93673549 on Build GRCh38
rs755496970 Chr.X: 93673137-93673137 on Build GRCh38
rs982103855 Chr.X: 93673280-93673280 on Build GRCh38
rs774159327 Chr.X: 93673244-93673244 on Build GRCh38

View Full Product Details