Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 100628668-100629207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN6
Entrez Gene ID
7105
Gene Name
tetraspanin 6
Gene Aliases
T245, TM4SF6, TSPAN-6
UniGene
Hs.43233
Gene Chromosome Location
Chr.X: 100627108-100637104 on Build GRCh38
Transcripts
NM_001278740.1 NM_001278741.1 NM_001278742.1 NM_001278743.1 NM_003270.3 XM_011531018.1

SNP Detail

SNP ID Chromosome Location
rs181408156 Chr.X: 100629020-100629020 on Build GRCh38
rs961155774 Chr.X: 100628851-100628851 on Build GRCh38
rs972800600 Chr.X: 100628880-100628880 on Build GRCh38
rs907206788 Chr.X: 100628725-100628725 on Build GRCh38
rs112075334 Chr.X: 100629169-100629169 on Build GRCh38
rs773438572 Chr.X: 100629016-100629016 on Build GRCh38
rs1006424109 Chr.X: 100628759-100628760 on Build GRCh38
rs911677872 Chr.X: 100628996-100628996 on Build GRCh38
rs767492786 Chr.X: 100628817-100628817 on Build GRCh38
rs955215228 Chr.X: 100628997-100628997 on Build GRCh38
rs1045767443 Chr.X: 100628717-100628717 on Build GRCh38
rs953053144 Chr.X: 100628886-100628886 on Build GRCh38
rs989372485 Chr.X: 100628854-100628854 on Build GRCh38
rs911148856 Chr.X: 100629029-100629029 on Build GRCh38
rs1010193103 Chr.X: 100628698-100628698 on Build GRCh38
rs895161917 Chr.X: 100628750-100628750 on Build GRCh38
rs1031233877 Chr.X: 100628826-100628826 on Build GRCh38
rs921820632 Chr.X: 100628862-100628862 on Build GRCh38
rs922257814 Chr.X: 100628952-100628952 on Build GRCh38
rs1037222822 Chr.X: 100629048-100629048 on Build GRCh38
rs1046237183 Chr.X: 100629087-100629087 on Build GRCh38
rs971213051 Chr.X: 100628813-100628813 on Build GRCh38
rs987541714 Chr.X: 100628982-100628982 on Build GRCh38
rs999396426 Chr.X: 100628816-100628816 on Build GRCh38
rs946102279 Chr.X: 100629085-100629085 on Build GRCh38
rs907179137 Chr.X: 100629138-100629138 on Build GRCh38
rs756176724 Chr.X: 100629159-100629159 on Build GRCh38
rs1025060032 Chr.X: 100628786-100628786 on Build GRCh38
rs952161907 Chr.X: 100628753-100628753 on Build GRCh38
rs913313788 Chr.X: 100629062-100629062 on Build GRCh38
rs1001755243 Chr.X: 100628734-100628734 on Build GRCh38
rs370488845 Chr.X: 100628965-100628965 on Build GRCh38
rs1016674912 Chr.X: 100628760-100628760 on Build GRCh38
rs79521063 Chr.X: 100628722-100628722 on Build GRCh38
rs1054258181 Chr.X: 100629058-100629058 on Build GRCh38
rs957843429 Chr.X: 100628838-100628838 on Build GRCh38
rs752674834 Chr.X: 100628976-100628978 on Build GRCh38
rs1012219050 Chr.X: 100628773-100628773 on Build GRCh38

View Full Product Details