Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 100632992-100633528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN6
Entrez Gene ID
7105
Gene Name
tetraspanin 6
Gene Aliases
T245, TM4SF6, TSPAN-6
UniGene
Hs.43233
Gene Chromosome Location
Chr.X: 100627108-100637104 on Build GRCh38
Transcripts
NM_001278740.1 NM_001278741.1 NM_001278742.1 NM_001278743.1 NM_003270.3 XM_011531018.1

SNP Detail

SNP ID Chromosome Location
rs867360514 Chr.X: 100633070-100633070 on Build GRCh38
rs749195747 Chr.X: 100633268-100633268 on Build GRCh38
rs762476388 Chr.X: 100633179-100633179 on Build GRCh38
rs760433475 Chr.X: 100633485-100633485 on Build GRCh38
rs770934344 Chr.X: 100633365-100633365 on Build GRCh38
rs754383668 Chr.X: 100633147-100633147 on Build GRCh38
rs915694579 Chr.X: 100633468-100633468 on Build GRCh38
rs775956142 Chr.X: 100633430-100633430 on Build GRCh38
rs747185293 Chr.X: 100633364-100633364 on Build GRCh38
rs764619818 Chr.X: 100633491-100633491 on Build GRCh38
rs905871954 Chr.X: 100633285-100633285 on Build GRCh38
rs778916459 Chr.X: 100633298-100633298 on Build GRCh38
rs752467243 Chr.X: 100633224-100633224 on Build GRCh38
rs937323113 Chr.X: 100633341-100633341 on Build GRCh38
rs770907503 Chr.X: 100633275-100633275 on Build GRCh38
rs776752997 Chr.X: 100633375-100633375 on Build GRCh38
rs899107575 Chr.X: 100633075-100633075 on Build GRCh38
rs760952738 Chr.X: 100633057-100633057 on Build GRCh38
rs753436909 Chr.X: 100633498-100633498 on Build GRCh38
rs922334801 Chr.X: 100633236-100633236 on Build GRCh38
rs755870439 Chr.X: 100633250-100633250 on Build GRCh38
rs192635520 Chr.X: 100633218-100633218 on Build GRCh38
rs770328851 Chr.X: 100633410-100633410 on Build GRCh38
rs1037691573 Chr.X: 100633065-100633065 on Build GRCh38
rs371846945 Chr.X: 100633428-100633428 on Build GRCh38
rs140549013 Chr.X: 100633267-100633267 on Build GRCh38
rs933512777 Chr.X: 100633251-100633251 on Build GRCh38
rs751162370 Chr.X: 100633200-100633200 on Build GRCh38
rs759091780 Chr.X: 100633502-100633502 on Build GRCh38
rs745947062 Chr.X: 100633380-100633380 on Build GRCh38
rs769026698 Chr.X: 100633457-100633457 on Build GRCh38
rs1008945280 Chr.X: 100633164-100633165 on Build GRCh38
rs1002043793 Chr.X: 100633103-100633103 on Build GRCh38
rs754607258 Chr.X: 100633212-100633212 on Build GRCh38
rs1010890162 Chr.X: 100633210-100633210 on Build GRCh38
rs775885243 Chr.X: 100633051-100633051 on Build GRCh38
rs190418709 Chr.X: 100633180-100633180 on Build GRCh38
rs1022484284 Chr.X: 100633262-100633262 on Build GRCh38
rs76568447 Chr.X: 100633480-100633480 on Build GRCh38
rs761215689 Chr.X: 100633297-100633297 on Build GRCh38
rs968594597 Chr.X: 100633150-100633150 on Build GRCh38
rs983610161 Chr.X: 100633257-100633257 on Build GRCh38
rs866472504 Chr.X: 100633029-100633029 on Build GRCh38
rs942240652 Chr.X: 100633274-100633274 on Build GRCh38
rs184384144 Chr.X: 100633143-100633143 on Build GRCh38
rs763250646 Chr.X: 100633437-100633437 on Build GRCh38
rs1006258601 Chr.X: 100633058-100633058 on Build GRCh38
rs753177093 Chr.X: 100633115-100633115 on Build GRCh38
rs766124468 Chr.X: 100633490-100633490 on Build GRCh38
rs1035748553 Chr.X: 100633109-100633109 on Build GRCh38

View Full Product Details