Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23967348-23967916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRSF10
Entrez Gene ID
10772
Gene Name
serine and arginine rich splicing factor 10
Gene Aliases
FUSIP1, FUSIP2, NSSR, PPP1R149, SFRS13, SFRS13A, SRp38, SRrp40, TASR, TASR1, TASR2
UniGene
Hs.3530
Gene Chromosome Location
Chr.1: 23964347-23980927 on Build GRCh38
Transcripts
NM_001191005.2 NM_001191006.2 NM_001191007.2 NM_001191009.2 NM_001300936.1 NM_001300937.1 NM_006625.5 NM_054016.3 NR_034035.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3804582 c.549C>G p.I183M Chr.1: 23967666-23967666 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1051583560 Chr.1: 23967533-23967533 on Build GRCh38
rs905842791 Chr.1: 23967760-23967760 on Build GRCh38
rs878912124 Chr.1: 23967586-23967586 on Build GRCh38
rs568954623 Chr.1: 23967580-23967580 on Build GRCh38
rs1057170288 Chr.1: 23967848-23967848 on Build GRCh38
rs9424411 Chr.1: 23967872-23967872 on Build GRCh38
rs4276860 Chr.1: 23967759-23967759 on Build GRCh38
rs1001513038 Chr.1: 23967764-23967764 on Build GRCh38
rs34040379 Chr.1: 23967442-23967443 on Build GRCh38
rs890411047 Chr.1: 23967596-23967596 on Build GRCh38
rs1010566731 Chr.1: 23967789-23967789 on Build GRCh38
rs35880755 Chr.1: 23967400-23967401 on Build GRCh38

View Full Product Details