Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 16709354-16709905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC144A
Entrez Gene ID
9720
Gene Name
coiled-coil domain containing 144A
Gene Aliases
-
UniGene
Hs.745246
Gene Chromosome Location
Chr.17: 16666768-16774999 on Build GRCh38
Transcripts
NM_014695.2 NR_130142.1 XM_011524089.2 XM_017025429.1 XM_017025430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5691907 c.1353A>G p.T451T Chr.17: 16709410-16709410 on Build GRCh38
336498 c.1298C>A p.P433Q Chr.17: 16709355-16709355 on Build GRCh38
1588673 c.1355A>C p.K452T Chr.17: 16709412-16709412 on Build GRCh38
1588672 c.1402A>C p.S468R Chr.17: 16709459-16709459 on Build GRCh38
976013 c.1501A>C p.K501Q Chr.17: 16709558-16709558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530227813 Chr.17: 16709841-16709841 on Build GRCh38
rs758971425 Chr.17: 16709536-16709536 on Build GRCh38
rs1014666704 Chr.17: 16709657-16709657 on Build GRCh38
rs550376205 Chr.17: 16709859-16709859 on Build GRCh38
rs774352522 Chr.17: 16709434-16709434 on Build GRCh38
rs767225436 Chr.17: 16709449-16709449 on Build GRCh38
rs763430123 Chr.17: 16709641-16709641 on Build GRCh38
rs992868086 Chr.17: 16709421-16709421 on Build GRCh38
rs925897564 Chr.17: 16709808-16709808 on Build GRCh38
rs967367214 Chr.17: 16709769-16709769 on Build GRCh38
rs1006027113 Chr.17: 16709398-16709398 on Build GRCh38
rs777989517 Chr.17: 16709556-16709556 on Build GRCh38
rs766701211 Chr.17: 16709537-16709537 on Build GRCh38
rs2621609 Chr.17: 16709879-16709879 on Build GRCh38
rs748109673 Chr.17: 16709568-16709568 on Build GRCh38
rs935860047 Chr.17: 16709472-16709472 on Build GRCh38
rs777966356 Chr.17: 16709679-16709679 on Build GRCh38
rs562909488 Chr.17: 16709651-16709651 on Build GRCh38
rs143021188 Chr.17: 16709789-16709789 on Build GRCh38
rs778053377 Chr.17: 16709577-16709577 on Build GRCh38
rs546377720 Chr.17: 16709423-16709423 on Build GRCh38
rs778825888 Chr.17: 16709593-16709593 on Build GRCh38
rs541151679 Chr.17: 16709428-16709428 on Build GRCh38
rs760307516 Chr.17: 16709636-16709636 on Build GRCh38
rs113734367 Chr.17: 16709638-16709638 on Build GRCh38
rs527732682 Chr.17: 16709786-16709786 on Build GRCh38
rs749083042 Chr.17: 16709610-16709612 on Build GRCh38
rs367718304 Chr.17: 16709646-16709646 on Build GRCh38
rs368458434 Chr.17: 16709580-16709580 on Build GRCh38
rs374355538 Chr.17: 16709550-16709550 on Build GRCh38
rs751970885 Chr.17: 16709540-16709540 on Build GRCh38
rs957420404 Chr.17: 16709843-16709843 on Build GRCh38
rs374867098 Chr.17: 16709599-16709599 on Build GRCh38
rs759948806 Chr.17: 16709653-16709653 on Build GRCh38
rs544967407 Chr.17: 16709654-16709654 on Build GRCh38
rs769575959 Chr.17: 16709404-16709404 on Build GRCh38
rs757107880 Chr.17: 16709663-16709663 on Build GRCh38
rs755105619 Chr.17: 16709542-16709542 on Build GRCh38
rs564960312 Chr.17: 16709708-16709708 on Build GRCh38
rs756355442 Chr.17: 16709659-16709659 on Build GRCh38
rs958946252 Chr.17: 16709408-16709408 on Build GRCh38
rs748280431 Chr.17: 16709399-16709399 on Build GRCh38
rs866157604 Chr.17: 16709392-16709392 on Build GRCh38
rs371251531 Chr.17: 16709521-16709521 on Build GRCh38
rs773285758 Chr.17: 16709405-16709405 on Build GRCh38
rs897502855 Chr.17: 16709543-16709543 on Build GRCh38
rs756312720 Chr.17: 16709409-16709410 on Build GRCh38
rs944367921 Chr.17: 16709514-16709514 on Build GRCh38
rs910324487 Chr.17: 16709495-16709495 on Build GRCh38
rs759076736 Chr.17: 16709445-16709445 on Build GRCh38
rs752947599 Chr.17: 16709655-16709655 on Build GRCh38
rs368571934 Chr.17: 16709491-16709491 on Build GRCh38
rs371753347 Chr.17: 16709581-16709581 on Build GRCh38
rs192192037 Chr.17: 16709456-16709456 on Build GRCh38
rs771560111 Chr.17: 16709616-16709616 on Build GRCh38
rs765608301 Chr.17: 16709502-16709502 on Build GRCh38
rs543644438 Chr.17: 16709576-16709576 on Build GRCh38
rs925763310 Chr.17: 16709458-16709458 on Build GRCh38
rs776158688 Chr.17: 16709640-16709640 on Build GRCh38
rs754017904 Chr.17: 16709684-16709684 on Build GRCh38
rs775226278 Chr.17: 16709635-16709635 on Build GRCh38
rs567785366 Chr.17: 16709839-16709839 on Build GRCh38

View Full Product Details