Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42299741-42300296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAT5A
Entrez Gene ID
6776
Gene Name
signal transducer and activator of transcription 5A
Gene Aliases
MGF, STAT5
UniGene
Hs.437058
Gene Chromosome Location
Chr.17: 42287547-42311943 on Build GRCh38
Transcripts
NM_001288718.1 NM_001288719.1 NM_001288720.1 NM_003152.3 XM_005257624.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6996871 c.605C>T p.T202M Chr.17: 42299805-42299805 on Build GRCh38
6884544 c.634G>A p.E212K Chr.17: 42299834-42299834 on Build GRCh38
1383352 c.819C>T p.D273D Chr.17: 42300267-42300267 on Build GRCh38
1383351 c.559G>T p.A187S Chr.17: 42299759-42299759 on Build GRCh38
6203209 c.789G>T p.G263G Chr.17: 42300237-42300237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008751297 Chr.17: 42299915-42299915 on Build GRCh38
rs781434526 Chr.17: 42299883-42299883 on Build GRCh38
rs766259233 Chr.17: 42299790-42299790 on Build GRCh38
rs542759072 Chr.17: 42299876-42299876 on Build GRCh38
rs946027832 Chr.17: 42299887-42299887 on Build GRCh38
rs375378402 Chr.17: 42299806-42299806 on Build GRCh38
rs577203457 Chr.17: 42300273-42300273 on Build GRCh38
rs2230123 Chr.17: 42299826-42299826 on Build GRCh38
rs561106561 Chr.17: 42299917-42299917 on Build GRCh38
rs754740482 Chr.17: 42299791-42299791 on Build GRCh38
rs772357085 Chr.17: 42299924-42299924 on Build GRCh38
rs750660966 Chr.17: 42300268-42300268 on Build GRCh38
rs770419080 Chr.17: 42300096-42300096 on Build GRCh38
rs760140777 Chr.17: 42299863-42299863 on Build GRCh38
rs950380490 Chr.17: 42299779-42299779 on Build GRCh38
rs1023962786 Chr.17: 42300059-42300059 on Build GRCh38
rs776108843 Chr.17: 42299820-42299820 on Build GRCh38
rs746131312 Chr.17: 42299890-42299890 on Build GRCh38
rs547365506 Chr.17: 42300014-42300014 on Build GRCh38
rs1051041483 Chr.17: 42299888-42299888 on Build GRCh38
rs759535072 Chr.17: 42299773-42299773 on Build GRCh38
rs774846322 Chr.17: 42299838-42299838 on Build GRCh38
rs913437925 Chr.17: 42300198-42300198 on Build GRCh38
rs374346157 Chr.17: 42299805-42299805 on Build GRCh38
rs765087246 Chr.17: 42299877-42299877 on Build GRCh38
rs990723381 Chr.17: 42300170-42300170 on Build GRCh38
rs942718644 Chr.17: 42300267-42300267 on Build GRCh38
rs765171270 Chr.17: 42300139-42300139 on Build GRCh38
rs200134221 Chr.17: 42299781-42299781 on Build GRCh38
rs537222147 Chr.17: 42300106-42300106 on Build GRCh38
rs549988812 Chr.17: 42299943-42299943 on Build GRCh38
rs748498942 Chr.17: 42299918-42299918 on Build GRCh38
rs995087540 Chr.17: 42299994-42299994 on Build GRCh38
rs762089778 Chr.17: 42299839-42299839 on Build GRCh38
rs536302118 Chr.17: 42300042-42300042 on Build GRCh38
rs779303545 Chr.17: 42299914-42299914 on Build GRCh38
rs554381637 Chr.17: 42300063-42300063 on Build GRCh38
rs558824323 Chr.17: 42300207-42300207 on Build GRCh38
rs912541717 Chr.17: 42300240-42300240 on Build GRCh38
rs759057425 Chr.17: 42300138-42300138 on Build GRCh38
rs900306818 Chr.17: 42299980-42299980 on Build GRCh38
rs1027367612 Chr.17: 42299998-42299998 on Build GRCh38
rs980326386 Chr.17: 42300072-42300072 on Build GRCh38
rs370086862 Chr.17: 42299906-42299906 on Build GRCh38
rs767141937 Chr.17: 42299849-42299849 on Build GRCh38
rs763843816 Chr.17: 42300247-42300247 on Build GRCh38
rs758149061 Chr.17: 42299879-42299879 on Build GRCh38
rs772954017 Chr.17: 42299896-42299896 on Build GRCh38
rs747188957 Chr.17: 42299804-42299804 on Build GRCh38
rs772823677 Chr.17: 42299925-42299925 on Build GRCh38
rs142186647 Chr.17: 42299765-42299765 on Build GRCh38
rs527783523 Chr.17: 42299850-42299850 on Build GRCh38
rs756240110 Chr.17: 42299893-42299893 on Build GRCh38
rs762777302 Chr.17: 42300234-42300234 on Build GRCh38
rs367568442 Chr.17: 42299825-42299825 on Build GRCh38
rs780338666 Chr.17: 42299903-42299903 on Build GRCh38
rs371007974 Chr.17: 42299799-42299799 on Build GRCh38
rs374276628 Chr.17: 42300013-42300013 on Build GRCh38
rs369438268 Chr.17: 42299819-42299819 on Build GRCh38
rs569837514 Chr.17: 42300082-42300082 on Build GRCh38
rs576311406 Chr.17: 42299763-42299763 on Build GRCh38
rs1038893567 Chr.17: 42300268-42300268 on Build GRCh38
rs749808850 Chr.17: 42299910-42299910 on Build GRCh38
rs571592630 Chr.17: 42299979-42299979 on Build GRCh38
rs927548430 Chr.17: 42300081-42300081 on Build GRCh38
rs531804028 Chr.17: 42299942-42299942 on Build GRCh38
rs565540435 Chr.17: 42300039-42300039 on Build GRCh38
rs374315116 Chr.17: 42299911-42299911 on Build GRCh38
rs745891584 Chr.17: 42299811-42299811 on Build GRCh38

View Full Product Details