Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73729478-73730069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPRKB
Entrez Gene ID
51002
Gene Name
TP53RK binding protein
Gene Aliases
CGI-121, CGI121
UniGene
Hs.157401
Gene Chromosome Location
Chr.2: 73729830-73740133 on Build GRCh38
Transcripts
NM_016058.2 XM_005264343.3 XM_006712026.3 XM_006712027.3 XM_011532877.2 XM_011532878.2 XM_011532879.2 XM_011532880.2 XM_017004232.1 XM_017004233.1 XM_017004234.1 XM_017004235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3426669 c.449A>C p.K150T Chr.2: 73730022-73730022 on Build GRCh38
3758365 c.474T>C p.I158I Chr.2: 73729997-73729997 on Build GRCh38
1409438 c.475G>A p.G159R Chr.2: 73729996-73729996 on Build GRCh38
5605168 c.468A>C p.E156D Chr.2: 73730003-73730003 on Build GRCh38
1409437 c.476G>T p.G159V Chr.2: 73729995-73729995 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs987010101 Chr.2: 73729684-73729684 on Build GRCh38
rs927420794 Chr.2: 73729783-73729783 on Build GRCh38
rs765629057 Chr.2: 73729968-73729968 on Build GRCh38
rs777224613 Chr.2: 73729948-73729948 on Build GRCh38
rs989440556 Chr.2: 73729744-73729744 on Build GRCh38
rs200885884 Chr.2: 73729507-73729521 on Build GRCh38
rs771658283 Chr.2: 73730037-73730037 on Build GRCh38
rs547471478 Chr.2: 73729915-73729915 on Build GRCh38
rs778740524 Chr.2: 73730027-73730027 on Build GRCh38
rs541169157 Chr.2: 73729713-73729713 on Build GRCh38
rs1037984780 Chr.2: 73729982-73729982 on Build GRCh38
rs765408516 Chr.2: 73729901-73729905 on Build GRCh38
rs948856283 Chr.2: 73729618-73729618 on Build GRCh38
rs754976073 Chr.2: 73730015-73730016 on Build GRCh38
rs767037310 Chr.2: 73729939-73729939 on Build GRCh38
rs747700337 Chr.2: 73729924-73729924 on Build GRCh38
rs369017132 Chr.2: 73729911-73729911 on Build GRCh38
rs963348618 Chr.2: 73729503-73729503 on Build GRCh38
rs771744684 Chr.2: 73729928-73729928 on Build GRCh38
rs377427709 Chr.2: 73729898-73729898 on Build GRCh38
rs12997568 Chr.2: 73729754-73729754 on Build GRCh38
rs749784962 Chr.2: 73730018-73730018 on Build GRCh38
rs751241236 Chr.2: 73729615-73729615 on Build GRCh38
rs112219090 Chr.2: 73729841-73729841 on Build GRCh38
rs555361002 Chr.2: 73729545-73729545 on Build GRCh38
rs967851355 Chr.2: 73729838-73729838 on Build GRCh38
rs34843558 Chr.2: 73729542-73729543 on Build GRCh38
rs534697795 Chr.2: 73729640-73729640 on Build GRCh38
rs769807239 Chr.2: 73729577-73729577 on Build GRCh38
rs889537195 Chr.2: 73729774-73729774 on Build GRCh38
rs765286185 Chr.2: 73729507-73729520 on Build GRCh38
rs371075002 Chr.2: 73730031-73730031 on Build GRCh38
rs149283944 Chr.2: 73729671-73729671 on Build GRCh38
rs7210 Chr.2: 73729997-73729997 on Build GRCh38
rs763140886 Chr.2: 73729990-73729990 on Build GRCh38
rs12713794 Chr.2: 73729719-73729719 on Build GRCh38
rs747097398 Chr.2: 73729894-73729894 on Build GRCh38
rs2969598 Chr.2: 73729521-73729521 on Build GRCh38
rs747967676 Chr.2: 73730029-73730029 on Build GRCh38
rs887101704 Chr.2: 73729801-73729801 on Build GRCh38
rs888642277 Chr.2: 73729702-73729702 on Build GRCh38
rs67805162 Chr.2: 73729814-73729814 on Build GRCh38
rs541879398 Chr.2: 73729794-73729794 on Build GRCh38
rs13020517 Chr.2: 73729617-73729617 on Build GRCh38
rs190278229 Chr.2: 73729804-73729804 on Build GRCh38
rs373441862 Chr.2: 73729935-73729935 on Build GRCh38
rs956304351 Chr.2: 73729740-73729740 on Build GRCh38
rs1005791838 Chr.2: 73729501-73729501 on Build GRCh38
rs773998085 Chr.2: 73729921-73729921 on Build GRCh38
rs760244184 Chr.2: 73729505-73729506 on Build GRCh38
rs777654695 Chr.2: 73729892-73729892 on Build GRCh38
rs953933682 Chr.2: 73729513-73729513 on Build GRCh38
rs772668640 Chr.2: 73729931-73729931 on Build GRCh38
rs961050034 Chr.2: 73729506-73729506 on Build GRCh38
rs374832089 Chr.2: 73729596-73729596 on Build GRCh38
rs201951702 Chr.2: 73729705-73729705 on Build GRCh38
rs375480496 Chr.2: 73729858-73729858 on Build GRCh38
rs1029383400 Chr.2: 73729510-73729510 on Build GRCh38
rs536196504 Chr.2: 73729518-73729532 on Build GRCh38
rs923873738 Chr.2: 73729578-73729578 on Build GRCh38
rs967946262 Chr.2: 73729527-73729528 on Build GRCh38
rs980118578 Chr.2: 73729775-73729775 on Build GRCh38
rs978027153 Chr.2: 73729546-73729546 on Build GRCh38
rs941595499 Chr.2: 73729707-73729707 on Build GRCh38
rs1022112054 Chr.2: 73729807-73729807 on Build GRCh38
rs934619237 Chr.2: 73729597-73729597 on Build GRCh38
rs141777165 Chr.2: 73729960-73729960 on Build GRCh38
rs997823634 Chr.2: 73729507-73729507 on Build GRCh38
rs947340345 Chr.2: 73729755-73729755 on Build GRCh38
rs558915274 Chr.2: 73729643-73729643 on Build GRCh38
rs753710518 Chr.2: 73730010-73730010 on Build GRCh38
rs985310638 Chr.2: 73729522-73729522 on Build GRCh38
rs909986621 Chr.2: 73730011-73730013 on Build GRCh38
rs752979476 Chr.2: 73729976-73729976 on Build GRCh38
rs377577443 Chr.2: 73730005-73730005 on Build GRCh38
rs751404929 Chr.2: 73730003-73730005 on Build GRCh38
rs1022583577 Chr.2: 73729526-73729526 on Build GRCh38
rs993245652 Chr.2: 73729606-73729606 on Build GRCh38
rs748239714 Chr.2: 73729535-73729535 on Build GRCh38
rs202158839 Chr.2: 73729520-73729520 on Build GRCh38
rs754783287 Chr.2: 73730026-73730026 on Build GRCh38
rs1046114 Chr.2: 73729917-73729917 on Build GRCh38
rs1029127495 Chr.2: 73729664-73729664 on Build GRCh38
rs992840124 Chr.2: 73729936-73729936 on Build GRCh38

View Full Product Details