Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74207611-74208204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTHFD2
Entrez Gene ID
10797
Gene Name
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, methenyltetrahydrofolate cyclohydrolase
Gene Aliases
NMDMC
UniGene
Hs.469030
Gene Chromosome Location
Chr.2: 74198563-74215298 on Build GRCh38
Transcripts
NM_006636.3 NR_027405.1 XM_006711924.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1752662 c.73G>C p.D25H Chr.2: 74207796-74207796 on Build GRCh38
5813974 c.381T>A p.D127E Chr.2: 74207798-74207798 on Build GRCh38
5792293 c.409+4A>G p.? Chr.2: 74207830-74207830 on Build GRCh38
6288379 c.307A>G p.M103V Chr.2: 74207724-74207724 on Build GRCh38
1215660 c.40T>G p.L14V Chr.2: 74207763-74207763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902062690 Chr.2: 74207973-74207973 on Build GRCh38
rs760767727 Chr.2: 74207792-74207792 on Build GRCh38
rs768040845 Chr.2: 74207849-74207849 on Build GRCh38
rs759402063 Chr.2: 74207700-74207700 on Build GRCh38
rs990303372 Chr.2: 74208000-74208000 on Build GRCh38
rs761468889 Chr.2: 74207834-74207834 on Build GRCh38
rs1034153686 Chr.2: 74208037-74208037 on Build GRCh38
rs919750626 Chr.2: 74208171-74208171 on Build GRCh38
rs772341902 Chr.2: 74207688-74207688 on Build GRCh38
rs375166259 Chr.2: 74207888-74207889 on Build GRCh38
rs367575757 Chr.2: 74207802-74207802 on Build GRCh38
rs553711937 Chr.2: 74207979-74207979 on Build GRCh38
rs529893372 Chr.2: 74208170-74208170 on Build GRCh38
rs889478376 Chr.2: 74207919-74207919 on Build GRCh38
rs749320177 Chr.2: 74207782-74207784 on Build GRCh38
rs765173316 Chr.2: 74207703-74207703 on Build GRCh38
rs1014850356 Chr.2: 74207897-74207897 on Build GRCh38
rs776651570 Chr.2: 74208101-74208104 on Build GRCh38
rs764402466 Chr.2: 74207646-74207646 on Build GRCh38
rs752465086 Chr.2: 74207714-74207714 on Build GRCh38
rs961278038 Chr.2: 74208140-74208140 on Build GRCh38
rs577874465 Chr.2: 74207864-74207864 on Build GRCh38
rs919704583 Chr.2: 74207818-74207818 on Build GRCh38
rs903960529 Chr.2: 74208012-74208012 on Build GRCh38
rs1049883489 Chr.2: 74207882-74207882 on Build GRCh38
rs779413862 Chr.2: 74207753-74207753 on Build GRCh38
rs1005394200 Chr.2: 74207844-74207844 on Build GRCh38
rs774757232 Chr.2: 74207674-74207674 on Build GRCh38
rs972560172 Chr.2: 74207759-74207759 on Build GRCh38
rs769195354 Chr.2: 74207745-74207745 on Build GRCh38
rs953665803 Chr.2: 74207993-74207993 on Build GRCh38
rs968155800 Chr.2: 74207668-74207668 on Build GRCh38
rs558882295 Chr.2: 74207854-74207854 on Build GRCh38
rs775545708 Chr.2: 74207800-74207800 on Build GRCh38
rs748382194 Chr.2: 74207757-74207757 on Build GRCh38
rs772392236 Chr.2: 74208083-74208086 on Build GRCh38
rs773272911 Chr.2: 74207698-74207698 on Build GRCh38
rs762032359 Chr.2: 74207814-74207814 on Build GRCh38
rs749729793 Chr.2: 74207743-74207743 on Build GRCh38
rs375474924 Chr.2: 74207950-74207950 on Build GRCh38
rs1021112144 Chr.2: 74207991-74207991 on Build GRCh38
rs776288126 Chr.2: 74208181-74208181 on Build GRCh38
rs370643260 Chr.2: 74207734-74207734 on Build GRCh38
rs774752295 Chr.2: 74207682-74207682 on Build GRCh38
rs765861213 Chr.2: 74208160-74208160 on Build GRCh38
rs537048762 Chr.2: 74207793-74207793 on Build GRCh38
rs931063971 Chr.2: 74207868-74207868 on Build GRCh38
rs760189914 Chr.2: 74207953-74207953 on Build GRCh38
rs938364416 Chr.2: 74208111-74208111 on Build GRCh38
rs545272678 Chr.2: 74207905-74207905 on Build GRCh38
rs894308461 Chr.2: 74207763-74207763 on Build GRCh38
rs772358344 Chr.2: 74207773-74207773 on Build GRCh38
rs10209904 Chr.2: 74208031-74208031 on Build GRCh38
rs780574345 Chr.2: 74207740-74207740 on Build GRCh38
rs368064055 Chr.2: 74208129-74208129 on Build GRCh38
rs780685596 Chr.2: 74207731-74207731 on Build GRCh38
rs768299038 Chr.2: 74208058-74208059 on Build GRCh38
rs373728570 Chr.2: 74207735-74207735 on Build GRCh38
rs767149883 Chr.2: 74207850-74207850 on Build GRCh38
rs780977517 Chr.2: 74207668-74207669 on Build GRCh38
rs750861744 Chr.2: 74207892-74207892 on Build GRCh38
rs922227513 Chr.2: 74207984-74207984 on Build GRCh38
rs1024937548 Chr.2: 74208121-74208121 on Build GRCh38
rs560975500 Chr.2: 74208090-74208090 on Build GRCh38
rs754433549 Chr.2: 74207878-74207878 on Build GRCh38
rs756899613 Chr.2: 74207724-74207724 on Build GRCh38
rs114367481 Chr.2: 74208068-74208068 on Build GRCh38
rs747340999 Chr.2: 74208056-74208057 on Build GRCh38
rs769034178 Chr.2: 74207673-74207673 on Build GRCh38
rs751273218 Chr.2: 74207723-74207723 on Build GRCh38
rs959369166 Chr.2: 74208054-74208054 on Build GRCh38
rs762110737 Chr.2: 74207685-74207685 on Build GRCh38
rs1040610233 Chr.2: 74207965-74207965 on Build GRCh38
rs773183383 Chr.2: 74207784-74207784 on Build GRCh38
rs952123650 Chr.2: 74207831-74207831 on Build GRCh38
rs190636717 Chr.2: 74207899-74207899 on Build GRCh38
rs945794865 Chr.2: 74208045-74208045 on Build GRCh38
rs928265620 Chr.2: 74208109-74208109 on Build GRCh38
rs763944779 Chr.2: 74207826-74207826 on Build GRCh38
rs749619298 Chr.2: 74207663-74207663 on Build GRCh38
rs765787524 Chr.2: 74207877-74207877 on Build GRCh38

View Full Product Details