Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 97113201-97113756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD36
Entrez Gene ID
375248
Gene Name
ankyrin repeat domain 36
Gene Aliases
UNQ2430
UniGene
Hs.541894
Gene Chromosome Location
Chr.2: 97113186-97264521 on Build GRCh38
Transcripts
NM_001164315.1 XM_006712514.3 XM_011511130.1 XM_011511131.1 XM_017004009.1 XM_017004010.1 XM_017004011.1 XM_017004012.1 XM_017004013.1 XM_017004014.1 XM_017004015.1 XM_017004016.1 XM_017004017.1 XM_017004018.1 XM_017004019.1 XM_017004020.1 XM_017004021.1 XM_017004022.1 XM_017004023.1 XM_017004024.1 XM_017004025.1 XM_017004026.1 XM_017004027.1 XM_017004028.1 XM_017004029.1 XM_017004030.1 XM_017004031.1 XM_017004032.1 XM_017004033.1 XM_017004034.1 XM_017004035.1 XM_017004036.1 XM_017004037.1 XM_017004038.1 XM_017004039.1 XM_017004040.1 XM_017004041.1 XM_017004042.1 XM_017004043.1 XM_017004044.1 XM_017004045.1 XM_017004046.1 XM_017004047.1 XM_017004048.1 XM_017004049.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7209004 c.5A>C p.E2A Chr.2: 97113744-97113744 on Build GRCh38
1532902 c.11G>A p.G4D Chr.2: 97113750-97113750 on Build GRCh38
7209003 c.5A>C p.E2A Chr.2: 97113744-97113744 on Build GRCh38
2826028 c.13A>G p.K5E Chr.2: 97113752-97113752 on Build GRCh38
2826029 c.13A>G p.K5E Chr.2: 97113752-97113752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759742645 Chr.2: 97113693-97113693 on Build GRCh38
rs774754940 Chr.2: 97113461-97113461 on Build GRCh38
rs972525963 Chr.2: 97113265-97113265 on Build GRCh38
rs747836220 Chr.2: 97113693-97113714 on Build GRCh38
rs368520397 Chr.2: 97113631-97113631 on Build GRCh38
rs1018878574 Chr.2: 97113612-97113612 on Build GRCh38
rs3878989 Chr.2: 97113303-97113303 on Build GRCh38
rs965278057 Chr.2: 97113621-97113621 on Build GRCh38
rs528028494 Chr.2: 97113395-97113395 on Build GRCh38
rs1008889833 Chr.2: 97113402-97113402 on Build GRCh38
rs548813872 Chr.2: 97113223-97113223 on Build GRCh38
rs918386218 Chr.2: 97113456-97113456 on Build GRCh38
rs762774979 Chr.2: 97113711-97113711 on Build GRCh38
rs749916465 Chr.2: 97113720-97113720 on Build GRCh38
rs765498151 Chr.2: 97113708-97113708 on Build GRCh38
rs570178734 Chr.2: 97113608-97113608 on Build GRCh38
rs983958667 Chr.2: 97113671-97113671 on Build GRCh38
rs757281592 Chr.2: 97113227-97113227 on Build GRCh38
rs1056783253 Chr.2: 97113527-97113527 on Build GRCh38
rs879606714 Chr.2: 97113666-97113666 on Build GRCh38
rs1050976987 Chr.2: 97113565-97113565 on Build GRCh38
rs540156263 Chr.2: 97113688-97113688 on Build GRCh38
rs1048067773 Chr.2: 97113307-97113307 on Build GRCh38
rs188657605 Chr.2: 97113219-97113219 on Build GRCh38
rs905487810 Chr.2: 97113308-97113308 on Build GRCh38
rs781219548 Chr.2: 97113565-97113569 on Build GRCh38
rs1028994623 Chr.2: 97113393-97113393 on Build GRCh38
rs3878990 Chr.2: 97113361-97113361 on Build GRCh38
rs927060997 Chr.2: 97113677-97113677 on Build GRCh38
rs1000652478 Chr.2: 97113385-97113385 on Build GRCh38
rs568750193 Chr.2: 97113359-97113359 on Build GRCh38
rs3866778 Chr.2: 97113660-97113660 on Build GRCh38
rs1041887764 Chr.2: 97113360-97113360 on Build GRCh38
rs775258747 Chr.2: 97113709-97113709 on Build GRCh38
rs3878987 Chr.2: 97113275-97113275 on Build GRCh38
rs952688358 Chr.2: 97113641-97113641 on Build GRCh38
rs1010434765 Chr.2: 97113355-97113356 on Build GRCh38
rs937033491 Chr.2: 97113495-97113495 on Build GRCh38
rs779922756 Chr.2: 97113732-97113732 on Build GRCh38
rs946253925 Chr.2: 97113536-97113536 on Build GRCh38
rs547779533 Chr.2: 97113423-97113423 on Build GRCh38
rs931040047 Chr.2: 97113304-97113304 on Build GRCh38
rs537622043 Chr.2: 97113635-97113635 on Build GRCh38
rs763995764 Chr.2: 97113714-97113714 on Build GRCh38
rs570881572 Chr.2: 97113428-97113428 on Build GRCh38
rs984318149 Chr.2: 97113468-97113468 on Build GRCh38
rs553302847 Chr.2: 97113729-97113729 on Build GRCh38
rs58302585 Chr.2: 97113343-97113343 on Build GRCh38
rs1041914092 Chr.2: 97113548-97113548 on Build GRCh38
rs1057513370 Chr.2: 97113334-97113334 on Build GRCh38
rs371922875 Chr.2: 97113719-97113719 on Build GRCh38
rs539875882 Chr.2: 97113433-97113433 on Build GRCh38
rs1019518237 Chr.2: 97113419-97113419 on Build GRCh38
rs751465684 Chr.2: 97113715-97113715 on Build GRCh38
rs889192292 Chr.2: 97113578-97113578 on Build GRCh38
rs3866779 Chr.2: 97113661-97113661 on Build GRCh38
rs1000365637 Chr.2: 97113554-97113554 on Build GRCh38
rs187519227 Chr.2: 97113378-97113378 on Build GRCh38
rs904649342 Chr.2: 97113552-97113552 on Build GRCh38
rs937020809 Chr.2: 97113317-97113317 on Build GRCh38
rs1814507 Chr.2: 97113645-97113645 on Build GRCh38
rs191678423 Chr.2: 97113454-97113454 on Build GRCh38
rs917525748 Chr.2: 97113529-97113529 on Build GRCh38
rs943906354 Chr.2: 97113252-97113252 on Build GRCh38
rs896058633 Chr.2: 97113354-97113354 on Build GRCh38
rs971594700 Chr.2: 97113366-97113391 on Build GRCh38
rs927055943 Chr.2: 97113470-97113470 on Build GRCh38
rs912476308 Chr.2: 97113248-97113248 on Build GRCh38
rs3878988 Chr.2: 97113278-97113278 on Build GRCh38
rs994006970 Chr.2: 97113628-97113628 on Build GRCh38
rs3866777 Chr.2: 97113604-97113604 on Build GRCh38

View Full Product Details