Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138803472-138804043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM213
Entrez Gene ID
155006
Gene Name
transmembrane protein 213
Gene Aliases
-
UniGene
Hs.567729
Gene Chromosome Location
Chr.7: 138797487-138806759 on Build GRCh38
Transcripts
NM_001085429.1 XM_005250167.3

SNP Detail

SNP ID Chromosome Location
rs1003332623 Chr.7: 138803526-138803526 on Build GRCh38
rs1054256547 Chr.7: 138803710-138803710 on Build GRCh38
rs971433000 Chr.7: 138803514-138803514 on Build GRCh38
rs540009105 Chr.7: 138803751-138803751 on Build GRCh38
rs946287296 Chr.7: 138803823-138803823 on Build GRCh38
rs898889189 Chr.7: 138803804-138803804 on Build GRCh38
rs1034827464 Chr.7: 138803862-138803862 on Build GRCh38
rs958845854 Chr.7: 138803596-138803596 on Build GRCh38
rs922145142 Chr.7: 138803713-138803713 on Build GRCh38
rs931798982 Chr.7: 138803709-138803709 on Build GRCh38
rs1020740481 Chr.7: 138803513-138803513 on Build GRCh38
rs751941665 Chr.7: 138803855-138803855 on Build GRCh38
rs971033655 Chr.7: 138803574-138803574 on Build GRCh38
rs540182802 Chr.7: 138803704-138803704 on Build GRCh38
rs190051130 Chr.7: 138803672-138803672 on Build GRCh38
rs533073369 Chr.7: 138803746-138803746 on Build GRCh38
rs3087006 Chr.7: 138803797-138803798 on Build GRCh38
rs542368943 Chr.7: 138803575-138803575 on Build GRCh38
rs573345462 Chr.7: 138803570-138803570 on Build GRCh38
rs555459950 Chr.7: 138803648-138803648 on Build GRCh38
rs1041639806 Chr.7: 138803802-138803802 on Build GRCh38
rs772284415 Chr.7: 138803985-138803985 on Build GRCh38
rs148449016 Chr.7: 138803890-138803890 on Build GRCh38
rs966088137 Chr.7: 138804007-138804007 on Build GRCh38
rs894601366 Chr.7: 138803870-138803870 on Build GRCh38
rs751054322 Chr.7: 138803569-138803569 on Build GRCh38
rs956631244 Chr.7: 138803969-138803970 on Build GRCh38
rs377546191 Chr.7: 138803779-138803779 on Build GRCh38
rs1054527335 Chr.7: 138803803-138803803 on Build GRCh38
rs956080110 Chr.7: 138803643-138803643 on Build GRCh38
rs1027118956 Chr.7: 138803891-138803891 on Build GRCh38
rs527270502 Chr.7: 138803498-138803498 on Build GRCh38
rs944404877 Chr.7: 138803703-138803703 on Build GRCh38
rs969189213 Chr.7: 138803996-138803996 on Build GRCh38
rs1010538663 Chr.7: 138803756-138803756 on Build GRCh38
rs940297146 Chr.7: 138803780-138803780 on Build GRCh38
rs1034458773 Chr.7: 138803578-138803578 on Build GRCh38
rs983415838 Chr.7: 138803980-138803980 on Build GRCh38
rs4999707 Chr.7: 138803814-138803814 on Build GRCh38
rs34623032 Chr.7: 138803779-138803780 on Build GRCh38
rs2084663 Chr.7: 138803742-138803742 on Build GRCh38
rs1035647723 Chr.7: 138803594-138803594 on Build GRCh38
rs1021645399 Chr.7: 138803977-138803977 on Build GRCh38
rs1019877203 Chr.7: 138803999-138803999 on Build GRCh38
rs777980218 Chr.7: 138803662-138803662 on Build GRCh38
rs766367688 Chr.7: 138803536-138803536 on Build GRCh38
rs1037387536 Chr.7: 138803783-138803784 on Build GRCh38
rs921715501 Chr.7: 138803707-138803707 on Build GRCh38
rs1010163825 Chr.7: 138803972-138803972 on Build GRCh38
rs907491897 Chr.7: 138803679-138803679 on Build GRCh38
rs1003413302 Chr.7: 138803893-138803893 on Build GRCh38
rs375045221 Chr.7: 138803798-138803798 on Build GRCh38
rs752327844 Chr.7: 138803647-138803647 on Build GRCh38
rs373508381 Chr.7: 138803830-138803830 on Build GRCh38
rs113809059 Chr.7: 138803678-138803678 on Build GRCh38
rs767085882 Chr.7: 138803614-138803614 on Build GRCh38
rs80190966 Chr.7: 138803946-138803946 on Build GRCh38

View Full Product Details