Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 26748276-26748845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRA1A
Entrez Gene ID
148
Gene Name
adrenoceptor alpha 1A
Gene Aliases
ADRA1C, ADRA1L1, ALPHA1AAR
UniGene
Hs.52931
Gene Chromosome Location
Chr.8: 26738113-26870994 on Build GRCh38
Transcripts
NM_001322503.1 NM_033303.4 NR_136343.1 XM_006716292.3 XM_017013094.1 XM_017013095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4408287 c.1410C>T p.D470D Chr.8: 26748608-26748608 on Build GRCh38
5716464 c.1087G>A p.D363N Chr.8: 26748545-26748545 on Build GRCh38
3927855 c.1376C>T p.A459V Chr.8: 26748642-26748642 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs950685781 Chr.8: 26748302-26748302 on Build GRCh38
rs61758626 Chr.8: 26748353-26748353 on Build GRCh38
rs771401155 Chr.8: 26748366-26748366 on Build GRCh38
rs895262347 Chr.8: 26748478-26748478 on Build GRCh38
rs539122253 Chr.8: 26748663-26748663 on Build GRCh38
rs61758627 Chr.8: 26748516-26748516 on Build GRCh38
rs1027506604 Chr.8: 26748512-26748512 on Build GRCh38
rs546240661 Chr.8: 26748330-26748330 on Build GRCh38
rs551144996 Chr.8: 26748401-26748401 on Build GRCh38
rs1026096361 Chr.8: 26748480-26748480 on Build GRCh38
rs571058158 Chr.8: 26748468-26748468 on Build GRCh38
rs61759694 Chr.8: 26748738-26748738 on Build GRCh38
rs544593378 Chr.8: 26748685-26748685 on Build GRCh38
rs935725340 Chr.8: 26748359-26748359 on Build GRCh38
rs1042999893 Chr.8: 26748463-26748463 on Build GRCh38
rs866960633 Chr.8: 26748619-26748619 on Build GRCh38
rs974405085 Chr.8: 26748348-26748348 on Build GRCh38
rs1055070501 Chr.8: 26748677-26748677 on Build GRCh38
rs754627109 Chr.8: 26748751-26748754 on Build GRCh38
rs965041152 Chr.8: 26748580-26748580 on Build GRCh38
rs948349035 Chr.8: 26748708-26748708 on Build GRCh38
rs192933532 Chr.8: 26748820-26748820 on Build GRCh38
rs184509584 Chr.8: 26748681-26748681 on Build GRCh38
rs989689949 Chr.8: 26748360-26748360 on Build GRCh38
rs748996690 Chr.8: 26748750-26748751 on Build GRCh38
rs534870257 Chr.8: 26748751-26748751 on Build GRCh38
rs560844906 Chr.8: 26748787-26748787 on Build GRCh38
rs1055041538 Chr.8: 26748474-26748474 on Build GRCh38
rs146880108 Chr.8: 26748469-26748469 on Build GRCh38
rs892789708 Chr.8: 26748438-26748439 on Build GRCh38
rs142441467 Chr.8: 26748307-26748307 on Build GRCh38
rs1028646970 Chr.8: 26748564-26748564 on Build GRCh38
rs915373381 Chr.8: 26748394-26748394 on Build GRCh38
rs930755267 Chr.8: 26748751-26748751 on Build GRCh38
rs61758629 Chr.8: 26748676-26748676 on Build GRCh38
rs566500446 Chr.8: 26748596-26748596 on Build GRCh38
rs921714814 Chr.8: 26748352-26748352 on Build GRCh38
rs753577687 Chr.8: 26748751-26748752 on Build GRCh38
rs192467915 Chr.8: 26748608-26748608 on Build GRCh38
rs1040220480 Chr.8: 26748805-26748805 on Build GRCh38
rs373833046 Chr.8: 26748540-26748540 on Build GRCh38
rs532751366 Chr.8: 26748358-26748358 on Build GRCh38
rs35884587 Chr.8: 26748770-26748770 on Build GRCh38
rs1006273059 Chr.8: 26748816-26748816 on Build GRCh38
rs61759696 Chr.8: 26748814-26748814 on Build GRCh38
rs932029206 Chr.8: 26748774-26748774 on Build GRCh38
rs967242338 Chr.8: 26748610-26748610 on Build GRCh38
rs61758630 Chr.8: 26748721-26748721 on Build GRCh38
rs56246211 Chr.8: 26748517-26748518 on Build GRCh38
rs990278704 Chr.8: 26748607-26748607 on Build GRCh38
rs934421618 Chr.8: 26748616-26748616 on Build GRCh38
rs397897201 Chr.8: 26748452-26748452 on Build GRCh38
rs1003158049 Chr.8: 26748476-26748476 on Build GRCh38
rs573581541 Chr.8: 26748759-26748760 on Build GRCh38
rs558772084 Chr.8: 26748627-26748627 on Build GRCh38
rs933452528 Chr.8: 26748339-26748339 on Build GRCh38
rs1004808632 Chr.8: 26748530-26748530 on Build GRCh38
rs975032604 Chr.8: 26748585-26748585 on Build GRCh38
rs995873951 Chr.8: 26748545-26748545 on Build GRCh38
rs34923219 Chr.8: 26748441-26748441 on Build GRCh38
rs777433956 Chr.8: 26748746-26748746 on Build GRCh38
rs61758628 Chr.8: 26748518-26748518 on Build GRCh38
rs1005160119 Chr.8: 26748788-26748788 on Build GRCh38
rs560450236 Chr.8: 26748333-26748333 on Build GRCh38
rs1012855418 Chr.8: 26748441-26748441 on Build GRCh38
rs61759695 Chr.8: 26748747-26748747 on Build GRCh38
rs995311441 Chr.8: 26748479-26748479 on Build GRCh38
rs553241251 Chr.8: 26748568-26748568 on Build GRCh38
rs923478980 Chr.8: 26748310-26748310 on Build GRCh38
rs868130110 Chr.8: 26748611-26748611 on Build GRCh38
rs554972791 Chr.8: 26748733-26748733 on Build GRCh38
rs945662174 Chr.8: 26748408-26748408 on Build GRCh38
rs957163254 Chr.8: 26748584-26748584 on Build GRCh38
rs965666159 Chr.8: 26748327-26748327 on Build GRCh38
rs1049576358 Chr.8: 26748769-26748769 on Build GRCh38
rs578174680 Chr.8: 26748597-26748610 on Build GRCh38
rs375528473 Chr.8: 26748306-26748306 on Build GRCh38

View Full Product Details