Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 77206000-77206482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS13A
Entrez Gene ID
23230
Gene Name
vacuolar protein sorting 13 homolog A
Gene Aliases
CHAC, CHOREIN
UniGene
Hs.459790
Gene Chromosome Location
Chr.9: 77177353-77421537 on Build GRCh38
Transcripts
NM_001018037.1 NM_001018038.2 NM_015186.3 NM_033305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6820192 c.320T>G p.L107R Chr.9: 77206014-77206014 on Build GRCh38
6820193 c.320T>G p.L107R Chr.9: 77206014-77206014 on Build GRCh38
4788812 c.333G>A p.K111K Chr.9: 77206027-77206027 on Build GRCh38
286456 c.348A>C p.K116N Chr.9: 77206042-77206042 on Build GRCh38
3217968 c.307G>C p.E103Q Chr.9: 77206001-77206001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886374999 Chr.9: 77206345-77206346 on Build GRCh38
rs7044740 Chr.9: 77206346-77206346 on Build GRCh38
rs753443354 Chr.9: 77206125-77206125 on Build GRCh38
rs745532800 Chr.9: 77206345-77206348 on Build GRCh38
rs201491206 Chr.9: 77206348-77206348 on Build GRCh38
rs760150991 Chr.9: 77206108-77206108 on Build GRCh38
rs761456498 Chr.9: 77206129-77206129 on Build GRCh38
rs774335892 Chr.9: 77206347-77206348 on Build GRCh38
rs996409167 Chr.9: 77206366-77206366 on Build GRCh38
rs990264776 Chr.9: 77206025-77206025 on Build GRCh38
rs765816986 Chr.9: 77206327-77206328 on Build GRCh38
rs758838089 Chr.9: 77206029-77206029 on Build GRCh38
rs759460937 Chr.9: 77206087-77206087 on Build GRCh38
rs554301406 Chr.9: 77206068-77206068 on Build GRCh38
rs141045905 Chr.9: 77206300-77206302 on Build GRCh38
rs45495494 Chr.9: 77206329-77206329 on Build GRCh38
rs771891218 Chr.9: 77206089-77206089 on Build GRCh38
rs964742584 Chr.9: 77206094-77206094 on Build GRCh38
rs747054782 Chr.9: 77206038-77206038 on Build GRCh38
rs780664594 Chr.9: 77206031-77206031 on Build GRCh38
rs926078279 Chr.9: 77206265-77206265 on Build GRCh38
rs779423321 Chr.9: 77206332-77206332 on Build GRCh38
rs571906446 Chr.9: 77206409-77206409 on Build GRCh38
rs58682676 Chr.9: 77206328-77206329 on Build GRCh38
rs753369621 Chr.9: 77206357-77206357 on Build GRCh38
rs576250969 Chr.9: 77206293-77206293 on Build GRCh38
rs73651497 Chr.9: 77206155-77206155 on Build GRCh38
rs775568234 Chr.9: 77206107-77206107 on Build GRCh38
rs763964169 Chr.9: 77206285-77206287 on Build GRCh38
rs781540482 Chr.9: 77206066-77206066 on Build GRCh38
rs189951909 Chr.9: 77206134-77206134 on Build GRCh38
rs556307196 Chr.9: 77206254-77206254 on Build GRCh38
rs61192750 Chr.9: 77206356-77206357 on Build GRCh38
rs1012592116 Chr.9: 77206422-77206422 on Build GRCh38
rs768897133 Chr.9: 77206053-77206053 on Build GRCh38
rs72275336 Chr.9: 77206084-77206093 on Build GRCh38
rs923559153 Chr.9: 77206331-77206331 on Build GRCh38
rs1014283290 Chr.9: 77206322-77206322 on Build GRCh38
rs1045782520 Chr.9: 77206443-77206443 on Build GRCh38
rs946258563 Chr.9: 77206169-77206169 on Build GRCh38
rs558281308 Chr.9: 77206407-77206407 on Build GRCh38
rs1043262160 Chr.9: 77206259-77206259 on Build GRCh38
rs942196006 Chr.9: 77206358-77206358 on Build GRCh38
rs568407702 Chr.9: 77206085-77206094 on Build GRCh38
rs776724808 Chr.9: 77206329-77206330 on Build GRCh38
rs763646015 Chr.9: 77206122-77206122 on Build GRCh38
rs71930615 Chr.9: 77206325-77206326 on Build GRCh38
rs774023870 Chr.9: 77206085-77206085 on Build GRCh38
rs1055925202 Chr.9: 77206292-77206292 on Build GRCh38
rs58922469 Chr.9: 77206345-77206346 on Build GRCh38
rs963203598 Chr.9: 77206354-77206354 on Build GRCh38
rs894177219 Chr.9: 77206410-77206410 on Build GRCh38

View Full Product Details