Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 87625288-87625865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL4
Entrez Gene ID
56062
Gene Name
kelch like family member 4
Gene Aliases
DKELCHL, KHL4
UniGene
Hs.49075
Gene Chromosome Location
Chr.X: 87517541-87670050 on Build GRCh38
Transcripts
NM_019117.4 NM_057162.2

SNP Detail

SNP ID Chromosome Location
rs775595775 Chr.X: 87625812-87625812 on Build GRCh38
rs185061011 Chr.X: 87625606-87625606 on Build GRCh38
rs778952516 Chr.X: 87625618-87625618 on Build GRCh38
rs959656720 Chr.X: 87625375-87625375 on Build GRCh38
rs371495295 Chr.X: 87625602-87625602 on Build GRCh38
rs1026703103 Chr.X: 87625514-87625514 on Build GRCh38
rs944326970 Chr.X: 87625692-87625692 on Build GRCh38
rs995245716 Chr.X: 87625469-87625469 on Build GRCh38
rs1038374373 Chr.X: 87625735-87625735 on Build GRCh38
rs1017980160 Chr.X: 87625448-87625448 on Build GRCh38
rs899562793 Chr.X: 87625459-87625459 on Build GRCh38
rs757568603 Chr.X: 87625787-87625787 on Build GRCh38
rs937478413 Chr.X: 87625334-87625334 on Build GRCh38
rs1025488589 Chr.X: 87625404-87625405 on Build GRCh38
rs951194594 Chr.X: 87625520-87625520 on Build GRCh38
rs754504540 Chr.X: 87625821-87625821 on Build GRCh38
rs748890314 Chr.X: 87625809-87625810 on Build GRCh38
rs755854259 Chr.X: 87625583-87625583 on Build GRCh38
rs1041954783 Chr.X: 87625418-87625418 on Build GRCh38
rs763501599 Chr.X: 87625760-87625760 on Build GRCh38
rs375911367 Chr.X: 87625837-87625837 on Build GRCh38
rs181279490 Chr.X: 87625601-87625601 on Build GRCh38
rs765334133 Chr.X: 87625738-87625738 on Build GRCh38
rs189289791 Chr.X: 87625524-87625524 on Build GRCh38
rs768303034 Chr.X: 87625613-87625613 on Build GRCh38
rs989784466 Chr.X: 87625515-87625515 on Build GRCh38
rs752028176 Chr.X: 87625785-87625785 on Build GRCh38
rs866140721 Chr.X: 87625625-87625625 on Build GRCh38
rs997690399 Chr.X: 87625321-87625321 on Build GRCh38
rs753175118 Chr.X: 87625739-87625739 on Build GRCh38
rs927337929 Chr.X: 87625589-87625589 on Build GRCh38
rs759568131 Chr.X: 87625731-87625731 on Build GRCh38
rs970702535 Chr.X: 87625563-87625563 on Build GRCh38
rs748334747 Chr.X: 87625438-87625438 on Build GRCh38
rs899469012 Chr.X: 87625809-87625809 on Build GRCh38
rs760799264 Chr.X: 87625694-87625694 on Build GRCh38
rs748134809 Chr.X: 87625630-87625630 on Build GRCh38
rs773114497 Chr.X: 87625666-87625666 on Build GRCh38
rs893282846 Chr.X: 87625373-87625373 on Build GRCh38
rs753575913 Chr.X: 87625816-87625816 on Build GRCh38
rs995150730 Chr.X: 87625830-87625830 on Build GRCh38
rs886593327 Chr.X: 87625669-87625669 on Build GRCh38
rs377161003 Chr.X: 87625591-87625591 on Build GRCh38
rs1054607842 Chr.X: 87625353-87625353 on Build GRCh38
rs267606526 Chr.X: 87625651-87625651 on Build GRCh38
rs946217946 Chr.X: 87625380-87625380 on Build GRCh38
rs891369492 Chr.X: 87625657-87625657 on Build GRCh38
rs1008302225 Chr.X: 87625443-87625443 on Build GRCh38
rs750023093 Chr.X: 87625571-87625571 on Build GRCh38
rs771114776 Chr.X: 87625727-87625727 on Build GRCh38
rs748081744 Chr.X: 87625836-87625836 on Build GRCh38
rs911115508 Chr.X: 87625523-87625523 on Build GRCh38
rs776734978 Chr.X: 87625730-87625730 on Build GRCh38
rs1006470045 Chr.X: 87625337-87625337 on Build GRCh38
rs754000445 Chr.X: 87625556-87625556 on Build GRCh38
rs778613516 Chr.X: 87625829-87625829 on Build GRCh38
rs771738040 Chr.X: 87625634-87625634 on Build GRCh38
rs757409017 Chr.X: 87625775-87625775 on Build GRCh38
rs1016294292 Chr.X: 87625356-87625356 on Build GRCh38
rs764227718 Chr.X: 87625324-87625324 on Build GRCh38
rs773163161 Chr.X: 87625555-87625555 on Build GRCh38

View Full Product Details