Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 24584971-24585528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP21
Entrez Gene ID
57584
Gene Name
Rho GTPase activating protein 21
Gene Aliases
ARHGAP10
UniGene
Hs.524195
Gene Chromosome Location
Chr.10: 24583609-24725421 on Build GRCh38
Transcripts
NM_020824.3 XM_005252542.3 XM_005252544.4 XM_011519602.2 XM_011519603.2 XM_011519604.2 XM_011519605.2 XM_011519606.2 XM_011519607.2 XM_017016459.1 XM_017016460.1 XM_017016461.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6065451 c.5194G>T p.V1732L Chr.10: 24585092-24585092 on Build GRCh38
4573555 c.5235A>G p.L1745L Chr.10: 24585051-24585051 on Build GRCh38
4573556 c.4913G>A p.S1638N Chr.10: 24585373-24585373 on Build GRCh38
6449423 c.5243C>T p.P1748L Chr.10: 24585043-24585043 on Build GRCh38
3978398 c.5042C>A p.T1681K Chr.10: 24585244-24585244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774601648 Chr.10: 24585033-24585033 on Build GRCh38
rs759311460 Chr.10: 24585334-24585334 on Build GRCh38
rs952012144 Chr.10: 24585039-24585039 on Build GRCh38
rs758648434 Chr.10: 24585371-24585371 on Build GRCh38
rs369715868 Chr.10: 24585486-24585486 on Build GRCh38
rs1143062 Chr.10: 24585360-24585360 on Build GRCh38
rs778138919 Chr.10: 24585019-24585019 on Build GRCh38
rs759249503 Chr.10: 24585252-24585252 on Build GRCh38
rs781358377 Chr.10: 24585000-24585000 on Build GRCh38
rs755309505 Chr.10: 24585438-24585438 on Build GRCh38
rs375200534 Chr.10: 24585416-24585416 on Build GRCh38
rs770060490 Chr.10: 24585008-24585008 on Build GRCh38
rs750504576 Chr.10: 24585425-24585425 on Build GRCh38
rs748253481 Chr.10: 24585006-24585006 on Build GRCh38
rs772723167 Chr.10: 24585326-24585326 on Build GRCh38
rs776516666 Chr.10: 24585474-24585474 on Build GRCh38
rs764764197 Chr.10: 24585083-24585083 on Build GRCh38
rs752660459 Chr.10: 24585257-24585257 on Build GRCh38
rs1133897 Chr.10: 24585509-24585509 on Build GRCh38
rs143603189 Chr.10: 24585398-24585398 on Build GRCh38
rs1143051 Chr.10: 24585457-24585457 on Build GRCh38
rs760540248 Chr.10: 24585412-24585412 on Build GRCh38
rs753096674 Chr.10: 24585451-24585451 on Build GRCh38
rs542841578 Chr.10: 24585226-24585226 on Build GRCh38
rs761307567 Chr.10: 24585079-24585079 on Build GRCh38
rs772625929 Chr.10: 24585227-24585227 on Build GRCh38
rs753880730 Chr.10: 24585357-24585357 on Build GRCh38
rs747471541 Chr.10: 24585319-24585319 on Build GRCh38
rs772580600 Chr.10: 24585075-24585075 on Build GRCh38
rs779820992 Chr.10: 24585465-24585465 on Build GRCh38
rs746202739 Chr.10: 24585190-24585190 on Build GRCh38
rs961689351 Chr.10: 24585178-24585178 on Build GRCh38
rs772490575 Chr.10: 24585205-24585205 on Build GRCh38
rs756232268 Chr.10: 24585169-24585169 on Build GRCh38
rs1143063 Chr.10: 24585348-24585348 on Build GRCh38
rs760630338 Chr.10: 24585353-24585353 on Build GRCh38
rs907648633 Chr.10: 24585138-24585138 on Build GRCh38
rs765963241 Chr.10: 24585243-24585243 on Build GRCh38
rs758736473 Chr.10: 24585307-24585307 on Build GRCh38
rs563608795 Chr.10: 24585468-24585468 on Build GRCh38
rs543832702 Chr.10: 24585419-24585419 on Build GRCh38
rs1143067 Chr.10: 24585278-24585278 on Build GRCh38
rs375897262 Chr.10: 24585293-24585293 on Build GRCh38
rs556765739 Chr.10: 24585091-24585091 on Build GRCh38
rs763923481 Chr.10: 24585356-24585356 on Build GRCh38
rs758555476 Chr.10: 24585426-24585426 on Build GRCh38
rs746142063 Chr.10: 24585297-24585297 on Build GRCh38
rs770327232 Chr.10: 24585384-24585384 on Build GRCh38
rs749514952 Chr.10: 24585027-24585027 on Build GRCh38
rs200596333 Chr.10: 24585292-24585292 on Build GRCh38
rs545273821 Chr.10: 24585335-24585335 on Build GRCh38
rs756676471 Chr.10: 24585452-24585452 on Build GRCh38
rs771305026 Chr.10: 24585032-24585032 on Build GRCh38
rs748733945 Chr.10: 24585383-24585383 on Build GRCh38
rs1143057 Chr.10: 24585404-24585404 on Build GRCh38
rs375019689 Chr.10: 24585363-24585363 on Build GRCh38
rs746707798 Chr.10: 24585467-24585467 on Build GRCh38
rs754891098 Chr.10: 24585151-24585151 on Build GRCh38
rs368102858 Chr.10: 24585260-24585260 on Build GRCh38
rs148825465 Chr.10: 24585381-24585381 on Build GRCh38
rs762603544 Chr.10: 24585232-24585232 on Build GRCh38
rs375505844 Chr.10: 24585258-24585258 on Build GRCh38
rs368694505 Chr.10: 24585313-24585313 on Build GRCh38
rs1110020 Chr.10: 24585405-24585405 on Build GRCh38
rs571130567 Chr.10: 24585009-24585009 on Build GRCh38
rs529388513 Chr.10: 24585496-24585496 on Build GRCh38
rs773934691 Chr.10: 24585390-24585390 on Build GRCh38
rs147761053 Chr.10: 24585332-24585332 on Build GRCh38
rs1001456949 Chr.10: 24585295-24585295 on Build GRCh38
rs1143072 Chr.10: 24585147-24585147 on Build GRCh38
rs761215786 Chr.10: 24585216-24585216 on Build GRCh38
rs372262105 Chr.10: 24585414-24585414 on Build GRCh38
rs753875325 Chr.10: 24585291-24585291 on Build GRCh38
rs755395699 Chr.10: 24585378-24585378 on Build GRCh38
rs769780369 Chr.10: 24585487-24585487 on Build GRCh38
rs1143070 Chr.10: 24585180-24585180 on Build GRCh38
rs746742561 Chr.10: 24585220-24585220 on Build GRCh38
rs372601705 Chr.10: 24585137-24585137 on Build GRCh38
rs770544831 Chr.10: 24585331-24585331 on Build GRCh38
rs748737080 Chr.10: 24585327-24585327 on Build GRCh38
rs368333258 Chr.10: 24585396-24585396 on Build GRCh38
rs766578541 Chr.10: 24585433-24585433 on Build GRCh38
rs1143052 Chr.10: 24585429-24585429 on Build GRCh38
rs1143069 Chr.10: 24585184-24585184 on Build GRCh38
rs145888629 Chr.10: 24585325-24585325 on Build GRCh38
rs367808073 Chr.10: 24585417-24585417 on Build GRCh38
rs142036127 Chr.10: 24585182-24585182 on Build GRCh38
rs1143058 Chr.10: 24585376-24585376 on Build GRCh38
rs767462034 Chr.10: 24585253-24585253 on Build GRCh38
rs751288085 Chr.10: 24585248-24585248 on Build GRCh38
rs375430948 Chr.10: 24585441-24585441 on Build GRCh38
rs1143059 Chr.10: 24585373-24585373 on Build GRCh38
rs752752522 Chr.10: 24585165-24585165 on Build GRCh38
rs775458794 Chr.10: 24585345-24585345 on Build GRCh38
rs3206453 Chr.10: 24585421-24585421 on Build GRCh38
rs775941789 Chr.10: 24585207-24585207 on Build GRCh38
rs771061399 Chr.10: 24585186-24585186 on Build GRCh38
rs1143053 Chr.10: 24585411-24585411 on Build GRCh38
rs922818994 Chr.10: 24585212-24585212 on Build GRCh38
rs1143060 Chr.10: 24585372-24585372 on Build GRCh38
rs368982209 Chr.10: 24585093-24585093 on Build GRCh38
rs1143054 Chr.10: 24585407-24585407 on Build GRCh38
rs750053031 Chr.10: 24585084-24585084 on Build GRCh38
rs1143050 Chr.10: 24585462-24585462 on Build GRCh38
rs867323833 Chr.10: 24585183-24585183 on Build GRCh38
rs553242970 Chr.10: 24585259-24585259 on Build GRCh38
rs868309592 Chr.10: 24585397-24585397 on Build GRCh38
rs763018622 Chr.10: 24585506-24585506 on Build GRCh38
rs778048970 Chr.10: 24585172-24585172 on Build GRCh38
rs1143075 Chr.10: 24585110-24585110 on Build GRCh38
rs1143061 Chr.10: 24585362-24585362 on Build GRCh38
rs371210209 Chr.10: 24585437-24585437 on Build GRCh38
rs776033597 Chr.10: 24585077-24585077 on Build GRCh38
rs781264880 Chr.10: 24585158-24585158 on Build GRCh38
rs776145004 Chr.10: 24585103-24585103 on Build GRCh38
rs1143055 Chr.10: 24585406-24585406 on Build GRCh38
rs745356491 Chr.10: 24585458-24585458 on Build GRCh38
rs1143076 Chr.10: 24585102-24585102 on Build GRCh38
rs537066065 Chr.10: 24585051-24585051 on Build GRCh38
rs747391102 Chr.10: 24585375-24585375 on Build GRCh38
rs1143073 Chr.10: 24585120-24585120 on Build GRCh38
rs1143071 Chr.10: 24585162-24585162 on Build GRCh38
rs370977656 Chr.10: 24585104-24585104 on Build GRCh38
rs913286043 Chr.10: 24585456-24585456 on Build GRCh38

View Full Product Details