Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33209832-33210308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RING1
Entrez Gene ID
6015
Gene Name
ring finger protein 1
Gene Aliases
RING1A, RNF1
UniGene
Hs.631989
Gene Chromosome Location
Chr.6: 33208509-33212722 on Build GRCh38
Transcripts
NM_002931.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3669521 c.394C>T p.H132Y Chr.6: 33210069-33210069 on Build GRCh38
5525525 c.261C>T p.C87C Chr.6: 33209936-33209936 on Build GRCh38
5525524 c.262C>T p.R88* Chr.6: 33209937-33209937 on Build GRCh38
1568400 c.368G>A p.R123Q Chr.6: 33210043-33210043 on Build GRCh38
4393718 c.341G>A p.R114Q Chr.6: 33210016-33210016 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145706732 Chr.6: 33210260-33210260 on Build GRCh38
rs962691999 Chr.6: 33210167-33210167 on Build GRCh38
rs185890541 Chr.6: 33209940-33209940 on Build GRCh38
rs778030870 Chr.6: 33210172-33210172 on Build GRCh38
rs796687310 Chr.6: 33209871-33209871 on Build GRCh38
rs138399139 Chr.6: 33209960-33209960 on Build GRCh38
rs932620853 Chr.6: 33210247-33210247 on Build GRCh38
rs766484158 Chr.6: 33210145-33210145 on Build GRCh38
rs774028517 Chr.6: 33210108-33210108 on Build GRCh38
rs998079087 Chr.6: 33210032-33210032 on Build GRCh38
rs753806303 Chr.6: 33210152-33210152 on Build GRCh38
rs780074052 Chr.6: 33209973-33209973 on Build GRCh38
rs778190373 Chr.6: 33210014-33210014 on Build GRCh38
rs377682205 Chr.6: 33210015-33210015 on Build GRCh38
rs145844429 Chr.6: 33209933-33209933 on Build GRCh38
rs762381214 Chr.6: 33209931-33209931 on Build GRCh38
rs182594008 Chr.6: 33209873-33209873 on Build GRCh38
rs747437599 Chr.6: 33209890-33209890 on Build GRCh38
rs775970204 Chr.6: 33210064-33210064 on Build GRCh38
rs370939105 Chr.6: 33210143-33210143 on Build GRCh38
rs537492111 Chr.6: 33210277-33210277 on Build GRCh38
rs750192143 Chr.6: 33210136-33210136 on Build GRCh38
rs943356767 Chr.6: 33210271-33210271 on Build GRCh38
rs772430673 Chr.6: 33209878-33209878 on Build GRCh38
rs1039490976 Chr.6: 33209977-33209977 on Build GRCh38
rs144721528 Chr.6: 33209900-33209900 on Build GRCh38
rs768258382 Chr.6: 33210183-33210191 on Build GRCh38
rs213208 Chr.6: 33210233-33210233 on Build GRCh38
rs761398688 Chr.6: 33210123-33210123 on Build GRCh38
rs962951094 Chr.6: 33209956-33209956 on Build GRCh38
rs773565706 Chr.6: 33209889-33209889 on Build GRCh38
rs757121144 Chr.6: 33209938-33209938 on Build GRCh38
rs766551289 Chr.6: 33210270-33210270 on Build GRCh38
rs758591908 Chr.6: 33210153-33210153 on Build GRCh38
rs770027759 Chr.6: 33210125-33210125 on Build GRCh38
rs373469632 Chr.6: 33210232-33210232 on Build GRCh38
rs763222816 Chr.6: 33209994-33209994 on Build GRCh38
rs960512158 Chr.6: 33210211-33210211 on Build GRCh38
rs1043584866 Chr.6: 33210240-33210240 on Build GRCh38
rs1006447276 Chr.6: 33210133-33210133 on Build GRCh38
rs61736933 Chr.6: 33210005-33210005 on Build GRCh38
rs748308232 Chr.6: 33209981-33209981 on Build GRCh38
rs750187590 Chr.6: 33209954-33209954 on Build GRCh38
rs765725767 Chr.6: 33209918-33209918 on Build GRCh38
rs774992909 Chr.6: 33209930-33209930 on Build GRCh38
rs565944825 Chr.6: 33210221-33210221 on Build GRCh38
rs868769018 Chr.6: 33209953-33209953 on Build GRCh38
rs771143710 Chr.6: 33210023-33210023 on Build GRCh38
rs373037542 Chr.6: 33210173-33210173 on Build GRCh38
rs759991167 Chr.6: 33209910-33209910 on Build GRCh38
rs767181284 Chr.6: 33209951-33209951 on Build GRCh38
rs770204859 Chr.6: 33210063-33210063 on Build GRCh38
rs757433349 Chr.6: 33210181-33210181 on Build GRCh38
rs925262532 Chr.6: 33210261-33210261 on Build GRCh38
rs547927195 Chr.6: 33210071-33210071 on Build GRCh38
rs763568949 Chr.6: 33210084-33210084 on Build GRCh38
rs751197447 Chr.6: 33209937-33209937 on Build GRCh38
rs925241945 Chr.6: 33210210-33210210 on Build GRCh38
rs369175859 Chr.6: 33210026-33210026 on Build GRCh38
rs746185117 Chr.6: 33210043-33210043 on Build GRCh38

View Full Product Details