Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33263930-33264432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS52
Entrez Gene ID
6293
Gene Name
VPS52, GARP complex subunit
Gene Aliases
ARE1, SAC2, SACM2L, dJ1033B10.5
UniGene
Hs.480356
Gene Chromosome Location
Chr.6: 33250272-33271965 on Build GRCh38
Transcripts
NM_001289174.1 NM_001289175.1 NM_001289176.1 NM_022553.5 XM_011514797.1 XM_011514798.2 XM_011514799.1 XM_017011177.1 XM_017011178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5870369 c.1531C>T p.R511C Chr.6: 33264097-33264097 on Build GRCh38
6820720 c.1554C>T p.S518S Chr.6: 33264074-33264074 on Build GRCh38
1568399 c.1470C>T p.S490S Chr.6: 33264428-33264428 on Build GRCh38
1077977 c.1474C>T p.R492* Chr.6: 33264424-33264424 on Build GRCh38
3349529 c.1592G>A p.R531Q Chr.6: 33264036-33264036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748991793 Chr.6: 33264360-33264360 on Build GRCh38
rs777963890 Chr.6: 33264023-33264023 on Build GRCh38
rs549125615 Chr.6: 33264300-33264300 on Build GRCh38
rs781654019 Chr.6: 33264347-33264347 on Build GRCh38
rs747311309 Chr.6: 33264332-33264332 on Build GRCh38
rs954735380 Chr.6: 33264230-33264230 on Build GRCh38
rs760052833 Chr.6: 33263975-33263975 on Build GRCh38
rs765419047 Chr.6: 33264145-33264145 on Build GRCh38
rs747540827 Chr.6: 33264027-33264027 on Build GRCh38
rs772451675 Chr.6: 33264022-33264022 on Build GRCh38
rs768134425 Chr.6: 33264368-33264368 on Build GRCh38
rs460568 Chr.6: 33264248-33264248 on Build GRCh38
rs780301860 Chr.6: 33263990-33263990 on Build GRCh38
rs755320816 Chr.6: 33263997-33263997 on Build GRCh38
rs757220709 Chr.6: 33263988-33263988 on Build GRCh38
rs767613503 Chr.6: 33264386-33264386 on Build GRCh38
rs762692222 Chr.6: 33264073-33264073 on Build GRCh38
rs754682846 Chr.6: 33264326-33264327 on Build GRCh38
rs977922952 Chr.6: 33264306-33264306 on Build GRCh38
rs924669738 Chr.6: 33264320-33264320 on Build GRCh38
rs567365122 Chr.6: 33264375-33264375 on Build GRCh38
rs548548380 Chr.6: 33264377-33264377 on Build GRCh38
rs370078243 Chr.6: 33264094-33264094 on Build GRCh38
rs781666258 Chr.6: 33263993-33263994 on Build GRCh38
rs759040822 Chr.6: 33264049-33264049 on Build GRCh38
rs752665333 Chr.6: 33264405-33264405 on Build GRCh38
rs752499446 Chr.6: 33263964-33263964 on Build GRCh38
rs527333584 Chr.6: 33264304-33264304 on Build GRCh38
rs776125760 Chr.6: 33264041-33264041 on Build GRCh38
rs763909768 Chr.6: 33264406-33264406 on Build GRCh38
rs752994849 Chr.6: 33264149-33264149 on Build GRCh38
rs200453261 Chr.6: 33264150-33264150 on Build GRCh38
rs773254313 Chr.6: 33264398-33264398 on Build GRCh38
rs769313005 Chr.6: 33264354-33264354 on Build GRCh38
rs551574533 Chr.6: 33264071-33264071 on Build GRCh38
rs760650425 Chr.6: 33264399-33264399 on Build GRCh38
rs763983027 Chr.6: 33263976-33263976 on Build GRCh38
rs761693768 Chr.6: 33264385-33264385 on Build GRCh38
rs377192361 Chr.6: 33263998-33263998 on Build GRCh38
rs974802038 Chr.6: 33264205-33264205 on Build GRCh38
rs973707309 Chr.6: 33264255-33264255 on Build GRCh38
rs573952590 Chr.6: 33264112-33264112 on Build GRCh38
rs757600726 Chr.6: 33263981-33263981 on Build GRCh38
rs374574456 Chr.6: 33264133-33264133 on Build GRCh38
rs375050499 Chr.6: 33264280-33264280 on Build GRCh38
rs213202 Chr.6: 33264278-33264278 on Build GRCh38
rs778248240 Chr.6: 33264362-33264363 on Build GRCh38
rs763980037 Chr.6: 33264074-33264074 on Build GRCh38
rs903299177 Chr.6: 33264004-33264004 on Build GRCh38
rs747631677 Chr.6: 33264383-33264383 on Build GRCh38
rs1033351711 Chr.6: 33264274-33264274 on Build GRCh38
rs751478385 Chr.6: 33263981-33263981 on Build GRCh38
rs745428517 Chr.6: 33264349-33264349 on Build GRCh38
rs757356152 Chr.6: 33264102-33264102 on Build GRCh38
rs1001184413 Chr.6: 33264132-33264132 on Build GRCh38
rs751263838 Chr.6: 33264088-33264088 on Build GRCh38
rs761854873 Chr.6: 33264096-33264096 on Build GRCh38
rs1042154930 Chr.6: 33264410-33264410 on Build GRCh38
rs375773684 Chr.6: 33264036-33264036 on Build GRCh38
rs149782269 Chr.6: 33264068-33264068 on Build GRCh38
rs769490620 Chr.6: 33264064-33264064 on Build GRCh38
rs764805606 Chr.6: 33263962-33263962 on Build GRCh38
rs923281149 Chr.6: 33264224-33264224 on Build GRCh38
rs369245452 Chr.6: 33264315-33264315 on Build GRCh38
rs760394050 Chr.6: 33264357-33264357 on Build GRCh38
rs936105692 Chr.6: 33264326-33264326 on Build GRCh38
rs141469953 Chr.6: 33264202-33264202 on Build GRCh38
rs768379569 Chr.6: 33264299-33264299 on Build GRCh38
rs997111398 Chr.6: 33264108-33264108 on Build GRCh38
rs959515227 Chr.6: 33264277-33264277 on Build GRCh38
rs201918121 Chr.6: 33263999-33263999 on Build GRCh38
rs371946607 Chr.6: 33264336-33264336 on Build GRCh38
rs937602948 Chr.6: 33264321-33264321 on Build GRCh38
rs755087840 Chr.6: 33264136-33264136 on Build GRCh38
rs763069543 Chr.6: 33264108-33264109 on Build GRCh38

View Full Product Details