Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 177280700-177281249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRINP2
Entrez Gene ID
57795
Gene Name
BMP/retinoic acid inducible neural specific 2
Gene Aliases
DBCCR1L2, FAM5B
UniGene
Hs.495918
Gene Chromosome Location
Chr.1: 177170922-177282422 on Build GRCh38
Transcripts
NM_021165.3 XM_005245379.1 XM_011509826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6123080 c.2052G>T p.Q684H Chr.1: 177281228-177281228 on Build GRCh38
260811 c.1797G>A p.E599E Chr.1: 177280973-177280973 on Build GRCh38
4828554 c.1793C>G p.S598C Chr.1: 177280969-177280969 on Build GRCh38
219450 c.1775C>A p.P592H Chr.1: 177280951-177280951 on Build GRCh38
4795329 c.1528C>T p.Q510* Chr.1: 177280704-177280704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777331988 Chr.1: 177281207-177281207 on Build GRCh38
rs764250922 Chr.1: 177281169-177281169 on Build GRCh38
rs750293974 Chr.1: 177280755-177280755 on Build GRCh38
rs758546554 Chr.1: 177281046-177281046 on Build GRCh38
rs148283069 Chr.1: 177280959-177280960 on Build GRCh38
rs759731438 Chr.1: 177281028-177281028 on Build GRCh38
rs771635814 Chr.1: 177281139-177281139 on Build GRCh38
rs201211995 Chr.1: 177280796-177280796 on Build GRCh38
rs775161210 Chr.1: 177280937-177280960 on Build GRCh38
rs751886595 Chr.1: 177281075-177281075 on Build GRCh38
rs369281014 Chr.1: 177281180-177281180 on Build GRCh38
rs559412885 Chr.1: 177280944-177280944 on Build GRCh38
rs145369948 Chr.1: 177280904-177280904 on Build GRCh38
rs773095070 Chr.1: 177280835-177280835 on Build GRCh38
rs768120673 Chr.1: 177280960-177280960 on Build GRCh38
rs906064649 Chr.1: 177281227-177281227 on Build GRCh38
rs760459965 Chr.1: 177281010-177281010 on Build GRCh38
rs765482953 Chr.1: 177281161-177281161 on Build GRCh38
rs753787662 Chr.1: 177281027-177281027 on Build GRCh38
rs746653373 Chr.1: 177281219-177281219 on Build GRCh38
rs1007881440 Chr.1: 177280826-177280826 on Build GRCh38
rs546046745 Chr.1: 177280844-177280844 on Build GRCh38
rs1038211415 Chr.1: 177281080-177281080 on Build GRCh38
rs779310557 Chr.1: 177280930-177280930 on Build GRCh38
rs752832498 Chr.1: 177281168-177281168 on Build GRCh38
rs796395753 Chr.1: 177281026-177281026 on Build GRCh38
rs1008842478 Chr.1: 177280881-177280881 on Build GRCh38
rs767469848 Chr.1: 177280751-177280751 on Build GRCh38
rs753028699 Chr.1: 177281039-177281039 on Build GRCh38
rs1012208156 Chr.1: 177281188-177281188 on Build GRCh38
rs142394323 Chr.1: 177280960-177280961 on Build GRCh38
rs767191417 Chr.1: 177280855-177280855 on Build GRCh38
rs776759875 Chr.1: 177281157-177281157 on Build GRCh38
rs890837441 Chr.1: 177280795-177280795 on Build GRCh38
rs775265813 Chr.1: 177280739-177280739 on Build GRCh38
rs779220314 Chr.1: 177280786-177280786 on Build GRCh38
rs1043166162 Chr.1: 177281125-177281125 on Build GRCh38
rs780481004 Chr.1: 177281194-177281194 on Build GRCh38
rs532244642 Chr.1: 177280824-177280824 on Build GRCh38
rs760418353 Chr.1: 177281145-177281145 on Build GRCh38
rs768210254 Chr.1: 177281122-177281122 on Build GRCh38
rs372116341 Chr.1: 177281131-177281131 on Build GRCh38
rs748351328 Chr.1: 177280933-177280933 on Build GRCh38
rs543200445 Chr.1: 177280766-177280766 on Build GRCh38
rs764625234 Chr.1: 177280727-177280727 on Build GRCh38
rs946224963 Chr.1: 177281065-177281065 on Build GRCh38
rs376484034 Chr.1: 177280827-177280827 on Build GRCh38
rs759536092 Chr.1: 177281159-177281159 on Build GRCh38
rs201432278 Chr.1: 177281203-177281203 on Build GRCh38
rs772872398 Chr.1: 177281141-177281141 on Build GRCh38
rs151314763 Chr.1: 177280837-177280837 on Build GRCh38
rs191738799 Chr.1: 177280957-177280957 on Build GRCh38
rs201494475 Chr.1: 177280978-177280978 on Build GRCh38
rs771351231 Chr.1: 177280953-177280953 on Build GRCh38
rs773810112 Chr.1: 177280821-177280821 on Build GRCh38
rs763946593 Chr.1: 177280746-177280746 on Build GRCh38
rs773949181 Chr.1: 177280841-177280841 on Build GRCh38
rs771572595 Chr.1: 177281197-177281197 on Build GRCh38
rs763754550 Chr.1: 177280838-177280838 on Build GRCh38
rs757609460 Chr.1: 177281175-177281175 on Build GRCh38
rs763172017 Chr.1: 177280747-177280747 on Build GRCh38
rs143029054 Chr.1: 177281132-177281132 on Build GRCh38
rs971088191 Chr.1: 177280917-177280917 on Build GRCh38
rs150033962 Chr.1: 177280872-177280872 on Build GRCh38
rs372209946 Chr.1: 177281114-177281114 on Build GRCh38
rs756265400 Chr.1: 177280758-177280758 on Build GRCh38
rs183912741 Chr.1: 177281061-177281061 on Build GRCh38
rs147592131 Chr.1: 177280979-177280979 on Build GRCh38
rs746349332 Chr.1: 177281115-177281115 on Build GRCh38
rs762582867 Chr.1: 177280740-177280740 on Build GRCh38
rs919791053 Chr.1: 177281090-177281090 on Build GRCh38
rs200578567 Chr.1: 177281079-177281079 on Build GRCh38
rs772887548 Chr.1: 177280998-177280998 on Build GRCh38
rs770730573 Chr.1: 177281153-177281153 on Build GRCh38
rs761440262 Chr.1: 177280995-177280995 on Build GRCh38
rs1050924102 Chr.1: 177281143-177281143 on Build GRCh38
rs970243412 Chr.1: 177280878-177280878 on Build GRCh38
rs993065363 Chr.1: 177281044-177281044 on Build GRCh38
rs758886283 Chr.1: 177280943-177280943 on Build GRCh38
rs766271373 Chr.1: 177280874-177280874 on Build GRCh38
rs753980724 Chr.1: 177280888-177280888 on Build GRCh38
rs774037708 Chr.1: 177280967-177280967 on Build GRCh38
rs755255376 Chr.1: 177280769-177280769 on Build GRCh38
rs765386717 Chr.1: 177281032-177281032 on Build GRCh38
rs771967341 Chr.1: 177280997-177280997 on Build GRCh38
rs755061077 Chr.1: 177280928-177280928 on Build GRCh38
rs1057243379 Chr.1: 177280737-177280737 on Build GRCh38
rs199882325 Chr.1: 177281226-177281226 on Build GRCh38
rs750430809 Chr.1: 177280859-177280859 on Build GRCh38
rs766249875 Chr.1: 177281011-177281011 on Build GRCh38
rs140006840 Chr.1: 177280791-177280791 on Build GRCh38
rs868647928 Chr.1: 177280956-177280956 on Build GRCh38
rs756995285 Chr.1: 177280749-177280749 on Build GRCh38
rs372195886 Chr.1: 177281190-177281190 on Build GRCh38
rs747622333 Chr.1: 177280948-177280948 on Build GRCh38
rs904581046 Chr.1: 177280806-177280806 on Build GRCh38
rs772010605 Chr.1: 177280829-177280829 on Build GRCh38
rs373675933 Chr.1: 177281088-177281088 on Build GRCh38
rs779985104 Chr.1: 177280759-177280759 on Build GRCh38
rs777143325 Chr.1: 177280955-177280955 on Build GRCh38
rs1053176889 Chr.1: 177281138-177281138 on Build GRCh38
rs149846085 Chr.1: 177281210-177281210 on Build GRCh38
rs139918070 Chr.1: 177280868-177280868 on Build GRCh38

View Full Product Details