Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186448256-186448781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102724919
Entrez Gene ID
102724919
Gene Name
uncharacterized LOC102724919
Gene Aliases
-
UniGene
Hs.576493
Gene Chromosome Location
Chr.1: 186435161-186470291 on Build GRCh38
Transcripts
NR_126002.1 NM_002597.4 NM_022576.3 XM_011509603.2

Gene Symbol
PDC
Entrez Gene ID
5132
Gene Name
phosducin
Gene Aliases
MEKA, PHD, PhLOP, PhLP
UniGene
Hs.654381
Gene Chromosome Location
Chr.1: 186443566-186461108 on Build GRCh38
Transcripts
NR_126002.1 NM_002597.4 NM_022576.3 XM_011509603.2

SNP Detail

SNP ID Chromosome Location
rs922854897 Chr.1: 186448386-186448386 on Build GRCh38
rs553869371 Chr.1: 186448367-186448367 on Build GRCh38
rs147246273 Chr.1: 186448467-186448467 on Build GRCh38
rs6672847 Chr.1: 186448602-186448602 on Build GRCh38
rs150554538 Chr.1: 186448708-186448708 on Build GRCh38
rs1026227292 Chr.1: 186448310-186448310 on Build GRCh38
rs917678984 Chr.1: 186448691-186448691 on Build GRCh38
rs1048774672 Chr.1: 186448458-186448458 on Build GRCh38
rs979743847 Chr.1: 186448639-186448639 on Build GRCh38
rs556949151 Chr.1: 186448508-186448508 on Build GRCh38
rs117176421 Chr.1: 186448351-186448351 on Build GRCh38
rs369675127 Chr.1: 186448632-186448632 on Build GRCh38
rs1007130749 Chr.1: 186448472-186448472 on Build GRCh38
rs959569386 Chr.1: 186448650-186448650 on Build GRCh38
rs989052106 Chr.1: 186448369-186448369 on Build GRCh38
rs139556853 Chr.1: 186448727-186448727 on Build GRCh38
rs969847787 Chr.1: 186448345-186448345 on Build GRCh38
rs559630515 Chr.1: 186448677-186448677 on Build GRCh38
rs779593647 Chr.1: 186448451-186448451 on Build GRCh38
rs925524684 Chr.1: 186448641-186448641 on Build GRCh38
rs895751390 Chr.1: 186448546-186448546 on Build GRCh38
rs1023114188 Chr.1: 186448342-186448342 on Build GRCh38
rs74134843 Chr.1: 186448487-186448487 on Build GRCh38
rs992509455 Chr.1: 186448326-186448327 on Build GRCh38
rs533402593 Chr.1: 186448678-186448678 on Build GRCh38
rs758845654 Chr.1: 186448571-186448571 on Build GRCh38
rs918280069 Chr.1: 186448721-186448721 on Build GRCh38
rs2774529 Chr.1: 186448704-186448704 on Build GRCh38
rs956155803 Chr.1: 186448687-186448687 on Build GRCh38
rs911270326 Chr.1: 186448431-186448431 on Build GRCh38
rs541234088 Chr.1: 186448659-186448659 on Build GRCh38
rs925505207 Chr.1: 186448339-186448339 on Build GRCh38
rs539158220 Chr.1: 186448590-186448590 on Build GRCh38
rs6672638 Chr.1: 186448352-186448352 on Build GRCh38
rs945627702 Chr.1: 186448301-186448301 on Build GRCh38
rs966091210 Chr.1: 186448563-186448563 on Build GRCh38
rs574200327 Chr.1: 186448637-186448637 on Build GRCh38
rs142358592 Chr.1: 186448516-186448516 on Build GRCh38
rs555669506 Chr.1: 186448682-186448682 on Build GRCh38
rs567436267 Chr.1: 186448412-186448412 on Build GRCh38
rs548949429 Chr.1: 186448409-186448409 on Build GRCh38
rs6659683 Chr.1: 186448318-186448318 on Build GRCh38
rs999942395 Chr.1: 186448623-186448623 on Build GRCh38
rs534636432 Chr.1: 186448413-186448413 on Build GRCh38
rs899001743 Chr.1: 186448463-186448463 on Build GRCh38
rs949064044 Chr.1: 186448404-186448404 on Build GRCh38
rs6672836 Chr.1: 186448565-186448565 on Build GRCh38
rs967102242 Chr.1: 186448627-186448627 on Build GRCh38
rs553322721 Chr.1: 186448456-186448456 on Build GRCh38

View Full Product Details