Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 90878921-90879500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPP30
Entrez Gene ID
10556
Gene Name
ribonuclease P/MRP subunit p30
Gene Aliases
TSG15
UniGene
Hs.139120
Gene Chromosome Location
Chr.10: 90859062-90908558 on Build GRCh38
Transcripts
NM_001104546.1 NM_006413.4 XM_011539120.2 XM_011539121.2 XM_017015475.1 XM_017015476.1 XM_017015477.1 XM_017015478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6039581 c.335delT p.H114fs*7 Chr.10: 90879127-90879127 on Build GRCh38
6769326 c.302A>C p.D101A Chr.10: 90879094-90879094 on Build GRCh38
5548349 c.377C>T p.A126V Chr.10: 90879103-90879103 on Build GRCh38
4937761 c.286A>G p.R96G Chr.10: 90879078-90879078 on Build GRCh38
6039580 c.335delT p.H114fs*7 Chr.10: 90879127-90879127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982757247 Chr.10: 90878998-90878998 on Build GRCh38
rs748326572 Chr.10: 90879160-90879160 on Build GRCh38
rs890094751 Chr.10: 90879177-90879177 on Build GRCh38
rs750526145 Chr.10: 90879029-90879029 on Build GRCh38
rs915019780 Chr.10: 90879478-90879478 on Build GRCh38
rs765698946 Chr.10: 90879105-90879105 on Build GRCh38
rs111724886 Chr.10: 90878957-90878958 on Build GRCh38
rs1004213725 Chr.10: 90879321-90879322 on Build GRCh38
rs770893004 Chr.10: 90879171-90879171 on Build GRCh38
rs546061535 Chr.10: 90879369-90879369 on Build GRCh38
rs147125295 Chr.10: 90879110-90879110 on Build GRCh38
rs747161058 Chr.10: 90879036-90879036 on Build GRCh38
rs762893913 Chr.10: 90879065-90879065 on Build GRCh38
rs766943901 Chr.10: 90879177-90879177 on Build GRCh38
rs994469493 Chr.10: 90879341-90879341 on Build GRCh38
rs1015555751 Chr.10: 90879327-90879327 on Build GRCh38
rs900024231 Chr.10: 90879295-90879295 on Build GRCh38
rs780305589 Chr.10: 90879034-90879034 on Build GRCh38
rs1057483086 Chr.10: 90879275-90879275 on Build GRCh38
rs950545727 Chr.10: 90879069-90879069 on Build GRCh38
rs147735522 Chr.10: 90879131-90879131 on Build GRCh38
rs200490313 Chr.10: 90879169-90879169 on Build GRCh38
rs934796612 Chr.10: 90879240-90879240 on Build GRCh38
rs773069991 Chr.10: 90879064-90879064 on Build GRCh38
rs779621853 Chr.10: 90879022-90879022 on Build GRCh38
rs369794466 Chr.10: 90879154-90879154 on Build GRCh38
rs148154173 Chr.10: 90878952-90878953 on Build GRCh38
rs569084133 Chr.10: 90879480-90879480 on Build GRCh38
rs762305187 Chr.10: 90879095-90879095 on Build GRCh38
rs757679133 Chr.10: 90879017-90879017 on Build GRCh38
rs770376071 Chr.10: 90879079-90879079 on Build GRCh38
rs766093849 Chr.10: 90878983-90878984 on Build GRCh38
rs758484260 Chr.10: 90879032-90879032 on Build GRCh38
rs201170208 Chr.10: 90879158-90879158 on Build GRCh38
rs924698775 Chr.10: 90879192-90879192 on Build GRCh38
rs1044645771 Chr.10: 90879304-90879304 on Build GRCh38
rs996073675 Chr.10: 90879407-90879407 on Build GRCh38
rs773901416 Chr.10: 90879085-90879086 on Build GRCh38
rs767244151 Chr.10: 90879085-90879085 on Build GRCh38
rs751709810 Chr.10: 90879133-90879133 on Build GRCh38
rs952850414 Chr.10: 90878981-90878981 on Build GRCh38
rs769721933 Chr.10: 90879057-90879057 on Build GRCh38
rs12242142 Chr.10: 90879002-90879002 on Build GRCh38
rs968674052 Chr.10: 90879441-90879441 on Build GRCh38
rs201044592 Chr.10: 90878953-90878954 on Build GRCh38
rs947389375 Chr.10: 90879173-90879173 on Build GRCh38
rs970694020 Chr.10: 90879365-90879365 on Build GRCh38
rs759005938 Chr.10: 90879119-90879119 on Build GRCh38
rs754138092 Chr.10: 90879091-90879091 on Build GRCh38
rs752316160 Chr.10: 90879321-90879321 on Build GRCh38
rs1027051090 Chr.10: 90878956-90878956 on Build GRCh38
rs896008813 Chr.10: 90879296-90879296 on Build GRCh38
rs773707268 Chr.10: 90879176-90879176 on Build GRCh38
rs768963585 Chr.10: 90879039-90879039 on Build GRCh38
rs780827517 Chr.10: 90879051-90879051 on Build GRCh38
rs964590669 Chr.10: 90879401-90879401 on Build GRCh38
rs55802732 Chr.10: 90878951-90878952 on Build GRCh38
rs556896805 Chr.10: 90879225-90879225 on Build GRCh38
rs745437678 Chr.10: 90879185-90879185 on Build GRCh38
rs767279095 Chr.10: 90879368-90879368 on Build GRCh38
rs1022225711 Chr.10: 90879418-90879418 on Build GRCh38
rs898795029 Chr.10: 90879320-90879320 on Build GRCh38
rs75192201 Chr.10: 90878950-90878951 on Build GRCh38
rs113145555 Chr.10: 90879147-90879147 on Build GRCh38
rs766593272 Chr.10: 90879424-90879424 on Build GRCh38
rs764063269 Chr.10: 90879439-90879439 on Build GRCh38
rs374963629 Chr.10: 90879056-90879056 on Build GRCh38
rs185359415 Chr.10: 90878995-90878995 on Build GRCh38
rs761540066 Chr.10: 90879291-90879291 on Build GRCh38
rs759076589 Chr.10: 90879084-90879084 on Build GRCh38
rs565849953 Chr.10: 90879080-90879080 on Build GRCh38
rs566526930 Chr.10: 90879213-90879213 on Build GRCh38
rs974689490 Chr.10: 90879159-90879159 on Build GRCh38
rs552303706 Chr.10: 90879367-90879367 on Build GRCh38
rs377053803 Chr.10: 90879303-90879303 on Build GRCh38
rs754266112 Chr.10: 90879014-90879014 on Build GRCh38
rs769523183 Chr.10: 90879162-90879162 on Build GRCh38
rs781547144 Chr.10: 90879152-90879152 on Build GRCh38
rs397812678 Chr.10: 90878954-90878955 on Build GRCh38
rs760771865 Chr.10: 90879061-90879062 on Build GRCh38
rs376252104 Chr.10: 90879370-90879370 on Build GRCh38
rs575482569 Chr.10: 90879454-90879458 on Build GRCh38
rs777570254 Chr.10: 90879168-90879168 on Build GRCh38
rs944307767 Chr.10: 90879234-90879235 on Build GRCh38

View Full Product Details