Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 58611879-58612475 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP91
Entrez Gene ID
80829
Gene Name
ZFP91 zinc finger protein
Gene Aliases
DMS-8, DSM-8, FKSG11, PZF, ZFP-91, ZNF757
UniGene
Hs.524920
Gene Chromosome Location
Chr.11: 58579114-58621550 on Build GRCh38
Transcripts
NM_001197051.1 NM_053023.4 NR_024091.1

Gene Symbol
ZFP91-CNTF
Entrez Gene ID
386607
Gene Name
ZFP91-CNTF readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.524920
Gene Chromosome Location
Chr.11: 58579114-58625733 on Build GRCh38
Transcripts
NM_001197051.1 NM_053023.4 NR_024091.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6826848 c.871C>T p.R291* Chr.11: 58612291-58612291 on Build GRCh38
3450012 c.889C>T p.P297S Chr.11: 58612309-58612309 on Build GRCh38
3450011 c.889C>T p.P297S Chr.11: 58612309-58612309 on Build GRCh38
6826847 c.871C>T p.R291* Chr.11: 58612291-58612291 on Build GRCh38
4560883 c.860G>A p.R287K Chr.11: 58612280-58612280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034645004 Chr.11: 58612040-58612040 on Build GRCh38
rs1052036019 Chr.11: 58611945-58611947 on Build GRCh38
rs750034172 Chr.11: 58612310-58612310 on Build GRCh38
rs747675932 Chr.11: 58612371-58612371 on Build GRCh38
rs766681812 Chr.11: 58611927-58611927 on Build GRCh38
rs768139018 Chr.11: 58612274-58612274 on Build GRCh38
rs899249271 Chr.11: 58612359-58612359 on Build GRCh38
rs779956088 Chr.11: 58612358-58612358 on Build GRCh38
rs763536713 Chr.11: 58612192-58612193 on Build GRCh38
rs745673709 Chr.11: 58612260-58612260 on Build GRCh38
rs199553022 Chr.11: 58612313-58612313 on Build GRCh38
rs936302471 Chr.11: 58611904-58611905 on Build GRCh38
rs568197313 Chr.11: 58611957-58611957 on Build GRCh38
rs776128009 Chr.11: 58612281-58612281 on Build GRCh38
rs936034208 Chr.11: 58612060-58612060 on Build GRCh38
rs754627818 Chr.11: 58612366-58612366 on Build GRCh38
rs746741085 Chr.11: 58612269-58612269 on Build GRCh38
rs894244617 Chr.11: 58612212-58612212 on Build GRCh38
rs376012550 Chr.11: 58611934-58611934 on Build GRCh38
rs929566529 Chr.11: 58612131-58612131 on Build GRCh38
rs890306592 Chr.11: 58612264-58612264 on Build GRCh38
rs991351132 Chr.11: 58611966-58611966 on Build GRCh38
rs990148143 Chr.11: 58612061-58612061 on Build GRCh38
rs910664784 Chr.11: 58612071-58612071 on Build GRCh38
rs772630557 Chr.11: 58612263-58612263 on Build GRCh38
rs886739343 Chr.11: 58611912-58611912 on Build GRCh38
rs149644873 Chr.11: 58612119-58612119 on Build GRCh38
rs775143010 Chr.11: 58611940-58611940 on Build GRCh38
rs971324126 Chr.11: 58612013-58612013 on Build GRCh38
rs977274923 Chr.11: 58612354-58612354 on Build GRCh38
rs1016588124 Chr.11: 58612192-58612192 on Build GRCh38
rs994840920 Chr.11: 58612386-58612386 on Build GRCh38
rs191528384 Chr.11: 58612320-58612320 on Build GRCh38
rs779395306 Chr.11: 58611928-58611928 on Build GRCh38
rs753772902 Chr.11: 58612227-58612227 on Build GRCh38
rs779541281 Chr.11: 58612267-58612267 on Build GRCh38
rs1004937768 Chr.11: 58612001-58612001 on Build GRCh38
rs183467698 Chr.11: 58612017-58612017 on Build GRCh38
rs779827248 Chr.11: 58612402-58612402 on Build GRCh38
rs139680226 Chr.11: 58612191-58612192 on Build GRCh38
rs890261799 Chr.11: 58612000-58612000 on Build GRCh38
rs182271764 Chr.11: 58612338-58612338 on Build GRCh38
rs750414228 Chr.11: 58612346-58612346 on Build GRCh38
rs567082716 Chr.11: 58612123-58612123 on Build GRCh38
rs544280159 Chr.11: 58612314-58612314 on Build GRCh38
rs751112611 Chr.11: 58612362-58612362 on Build GRCh38
rs535496393 Chr.11: 58611982-58611982 on Build GRCh38
rs778811517 Chr.11: 58612253-58612253 on Build GRCh38
rs549449974 Chr.11: 58612023-58612023 on Build GRCh38
rs534378166 Chr.11: 58612152-58612152 on Build GRCh38
rs774645473 Chr.11: 58612304-58612304 on Build GRCh38
rs1024117634 Chr.11: 58611988-58611988 on Build GRCh38
rs780636457 Chr.11: 58612369-58612369 on Build GRCh38
rs748633292 Chr.11: 58612245-58612246 on Build GRCh38
rs765245419 Chr.11: 58612344-58612344 on Build GRCh38
rs767996632 Chr.11: 58612312-58612312 on Build GRCh38
rs888035851 Chr.11: 58612189-58612189 on Build GRCh38
rs923671098 Chr.11: 58612184-58612184 on Build GRCh38
rs201852935 Chr.11: 58612325-58612325 on Build GRCh38
rs771455127 Chr.11: 58612292-58612292 on Build GRCh38
rs560163236 Chr.11: 58612343-58612343 on Build GRCh38
rs763995481 Chr.11: 58612018-58612018 on Build GRCh38
rs376953189 Chr.11: 58612266-58612266 on Build GRCh38
rs957297944 Chr.11: 58612403-58612403 on Build GRCh38
rs769337242 Chr.11: 58612376-58612376 on Build GRCh38
rs963372206 Chr.11: 58612029-58612029 on Build GRCh38
rs555542772 Chr.11: 58612191-58612191 on Build GRCh38
rs943426602 Chr.11: 58612094-58612094 on Build GRCh38
rs371368430 Chr.11: 58612349-58612349 on Build GRCh38
rs757267656 Chr.11: 58612249-58612249 on Build GRCh38
rs749726346 Chr.11: 58612288-58612288 on Build GRCh38
rs778491059 Chr.11: 58612190-58612191 on Build GRCh38
rs188008158 Chr.11: 58612033-58612033 on Build GRCh38
rs996065067 Chr.11: 58611939-58611939 on Build GRCh38
rs2509921 Chr.11: 58612058-58612058 on Build GRCh38
rs931899916 Chr.11: 58612226-58612226 on Build GRCh38
rs766941977 Chr.11: 58612169-58612169 on Build GRCh38

View Full Product Details