Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49599700-49600299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM186B
Entrez Gene ID
84070
Gene Name
family with sequence similarity 186 member B
Gene Aliases
C12orf25
UniGene
Hs.524406
Gene Chromosome Location
Chr.12: 49582875-49605710 on Build GRCh38
Transcripts
NM_032130.2 NR_027450.1 XM_006719625.2 XM_006719626.2 XM_006719627.3 XM_011538796.2 XM_011538797.2 XM_017020008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4682795 c.1337A>G p.Q446R Chr.12: 49600033-49600033 on Build GRCh38
1705567 c.1423A>T p.S475C Chr.12: 49600217-49600217 on Build GRCh38
6659096 c.1884G>A p.K628K Chr.12: 49599756-49599756 on Build GRCh38
5815918 c.1577T>A p.L526Q Chr.12: 49600063-49600063 on Build GRCh38
7111437 c.1089C>G p.L363L Chr.12: 49600281-49600281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376802867 Chr.12: 49600132-49600132 on Build GRCh38
rs773976789 Chr.12: 49599797-49599797 on Build GRCh38
rs747733317 Chr.12: 49600001-49600001 on Build GRCh38
rs535110127 Chr.12: 49599917-49599917 on Build GRCh38
rs772008879 Chr.12: 49600006-49600006 on Build GRCh38
rs760890443 Chr.12: 49600255-49600255 on Build GRCh38
rs1040157548 Chr.12: 49599836-49599836 on Build GRCh38
rs566685124 Chr.12: 49600256-49600256 on Build GRCh38
rs778234525 Chr.12: 49599857-49599857 on Build GRCh38
rs776610710 Chr.12: 49600026-49600026 on Build GRCh38
rs370586607 Chr.12: 49600158-49600158 on Build GRCh38
rs371259662 Chr.12: 49599911-49599911 on Build GRCh38
rs148848955 Chr.12: 49600271-49600271 on Build GRCh38
rs944109383 Chr.12: 49600122-49600122 on Build GRCh38
rs933711881 Chr.12: 49599757-49599757 on Build GRCh38
rs776499332 Chr.12: 49600135-49600135 on Build GRCh38
rs375738935 Chr.12: 49600230-49600230 on Build GRCh38
rs770748895 Chr.12: 49600133-49600133 on Build GRCh38
rs137909183 Chr.12: 49600248-49600248 on Build GRCh38
rs777016632 Chr.12: 49600264-49600264 on Build GRCh38
rs757007321 Chr.12: 49600197-49600197 on Build GRCh38
rs930209645 Chr.12: 49599972-49599972 on Build GRCh38
rs546333213 Chr.12: 49600032-49600032 on Build GRCh38
rs1045212890 Chr.12: 49600240-49600240 on Build GRCh38
rs775212053 Chr.12: 49599778-49599778 on Build GRCh38
rs765347376 Chr.12: 49600246-49600246 on Build GRCh38
rs896649632 Chr.12: 49600005-49600005 on Build GRCh38
rs768197102 Chr.12: 49599981-49599981 on Build GRCh38
rs12299908 Chr.12: 49599983-49599983 on Build GRCh38
rs201221640 Chr.12: 49600048-49600048 on Build GRCh38
rs766850153 Chr.12: 49600052-49600052 on Build GRCh38
rs752982311 Chr.12: 49599854-49599854 on Build GRCh38
rs146791191 Chr.12: 49599881-49599881 on Build GRCh38
rs777094375 Chr.12: 49599770-49599770 on Build GRCh38
rs372553683 Chr.12: 49599861-49599861 on Build GRCh38
rs758503302 Chr.12: 49599855-49599855 on Build GRCh38
rs150032579 Chr.12: 49599809-49599809 on Build GRCh38
rs74239917 Chr.12: 49600019-49600019 on Build GRCh38
rs780578524 Chr.12: 49599956-49599956 on Build GRCh38
rs1000946854 Chr.12: 49600251-49600251 on Build GRCh38
rs772737486 Chr.12: 49600145-49600146 on Build GRCh38
rs747540745 Chr.12: 49599755-49599755 on Build GRCh38
rs374835327 Chr.12: 49599959-49599959 on Build GRCh38
rs745752039 Chr.12: 49600149-49600149 on Build GRCh38
rs200290691 Chr.12: 49600183-49600183 on Build GRCh38
rs761974024 Chr.12: 49600037-49600037 on Build GRCh38
rs376716288 Chr.12: 49599899-49599899 on Build GRCh38
rs761847616 Chr.12: 49600153-49600153 on Build GRCh38
rs772558743 Chr.12: 49600045-49600046 on Build GRCh38
rs761495376 Chr.12: 49600143-49600150 on Build GRCh38
rs146948851 Chr.12: 49600187-49600187 on Build GRCh38
rs764273824 Chr.12: 49599937-49599937 on Build GRCh38
rs748284721 Chr.12: 49600232-49600232 on Build GRCh38
rs745823010 Chr.12: 49600220-49600220 on Build GRCh38
rs142744818 Chr.12: 49600175-49600175 on Build GRCh38
rs771218796 Chr.12: 49600259-49600259 on Build GRCh38
rs771389627 Chr.12: 49599765-49599765 on Build GRCh38
rs200786079 Chr.12: 49599918-49599918 on Build GRCh38
rs754446694 Chr.12: 49600078-49600078 on Build GRCh38
rs767609528 Chr.12: 49599805-49599805 on Build GRCh38
rs765000122 Chr.12: 49600146-49600148 on Build GRCh38
rs571026024 Chr.12: 49599870-49599870 on Build GRCh38
rs771086324 Chr.12: 49599762-49599773 on Build GRCh38
rs150032302 Chr.12: 49599850-49599850 on Build GRCh38
rs913059016 Chr.12: 49599902-49599902 on Build GRCh38
rs867554059 Chr.12: 49600022-49600022 on Build GRCh38
rs150140309 Chr.12: 49600033-49600033 on Build GRCh38
rs762337069 Chr.12: 49600168-49600169 on Build GRCh38
rs774561616 Chr.12: 49600035-49600035 on Build GRCh38
rs768744459 Chr.12: 49600150-49600150 on Build GRCh38
rs52824916 Chr.12: 49599895-49599895 on Build GRCh38
rs758798875 Chr.12: 49599720-49599720 on Build GRCh38
rs763969730 Chr.12: 49599795-49599795 on Build GRCh38
rs372928425 Chr.12: 49599896-49599896 on Build GRCh38
rs1038445571 Chr.12: 49599782-49599782 on Build GRCh38
rs200005083 Chr.12: 49599947-49599947 on Build GRCh38
rs374816762 Chr.12: 49600188-49600188 on Build GRCh38
rs760701513 Chr.12: 49600009-49600009 on Build GRCh38
rs747829471 Chr.12: 49600085-49600085 on Build GRCh38
rs199944340 Chr.12: 49600079-49600079 on Build GRCh38
rs541823903 Chr.12: 49600194-49600194 on Build GRCh38
rs370657151 Chr.12: 49599946-49599946 on Build GRCh38
rs557621248 Chr.12: 49600045-49600045 on Build GRCh38
rs148050129 Chr.12: 49599953-49599953 on Build GRCh38
rs543736560 Chr.12: 49600169-49600169 on Build GRCh38
rs775998573 Chr.12: 49600122-49600124 on Build GRCh38
rs754362439 Chr.12: 49600072-49600072 on Build GRCh38
rs758087218 Chr.12: 49600103-49600103 on Build GRCh38
rs139362702 Chr.12: 49599779-49599779 on Build GRCh38
rs773097118 Chr.12: 49600007-49600007 on Build GRCh38
rs994392582 Chr.12: 49599811-49599811 on Build GRCh38
rs767899751 Chr.12: 49600041-49600041 on Build GRCh38
rs778505020 Chr.12: 49600084-49600084 on Build GRCh38
rs780983455 Chr.12: 49600205-49600205 on Build GRCh38
rs774701559 Chr.12: 49599961-49599961 on Build GRCh38
rs369228240 Chr.12: 49599892-49599892 on Build GRCh38
rs896868175 Chr.12: 49599776-49599776 on Build GRCh38
rs150891582 Chr.12: 49599772-49599772 on Build GRCh38
rs111881897 Chr.12: 49600156-49600156 on Build GRCh38
rs1032328206 Chr.12: 49600074-49600074 on Build GRCh38
rs759753697 Chr.12: 49599814-49599814 on Build GRCh38
rs756537392 Chr.12: 49599955-49599955 on Build GRCh38
rs200799283 Chr.12: 49599869-49599869 on Build GRCh38
rs372452686 Chr.12: 49600222-49600222 on Build GRCh38
rs761666903 Chr.12: 49599799-49599799 on Build GRCh38
rs748999703 Chr.12: 49599891-49599891 on Build GRCh38
rs373354688 Chr.12: 49599936-49599936 on Build GRCh38
rs569309521 Chr.12: 49599912-49599912 on Build GRCh38
rs371185924 Chr.12: 49600037-49600037 on Build GRCh38
rs145528972 Chr.12: 49599863-49599863 on Build GRCh38
rs143625590 Chr.12: 49599879-49599879 on Build GRCh38
rs778851005 Chr.12: 49599944-49599944 on Build GRCh38
rs777945294 Chr.12: 49599750-49599750 on Build GRCh38
rs138755693 Chr.12: 49600174-49600174 on Build GRCh38
rs774374047 Chr.12: 49600151-49600151 on Build GRCh38
rs986615654 Chr.12: 49600172-49600172 on Build GRCh38
rs759951365 Chr.12: 49600267-49600267 on Build GRCh38
rs201041491 Chr.12: 49599773-49599773 on Build GRCh38
rs752298682 Chr.12: 49600184-49600184 on Build GRCh38
rs779637034 Chr.12: 49599725-49599725 on Build GRCh38
rs143376289 Chr.12: 49600269-49600269 on Build GRCh38
rs954448103 Chr.12: 49599990-49599990 on Build GRCh38

View Full Product Details