Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49460499-49460923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATS2
Entrez Gene ID
65244
Gene Name
spermatogenesis associated serine rich 2
Gene Aliases
Nbla00526, P59SCR, SCR59, SPATA10
UniGene
Hs.654826
Gene Chromosome Location
Chr.12: 49366905-49527441 on Build GRCh38
Transcripts
NM_001293285.1 NM_001293286.1 NM_023071.3 NR_120663.1 XM_011538690.2 XM_011538691.2 XM_017019840.1

SNP Detail

SNP ID Chromosome Location
rs745750342 Chr.12: 49460601-49460601 on Build GRCh38
rs889514292 Chr.12: 49460670-49460670 on Build GRCh38
rs1013735888 Chr.12: 49460818-49460818 on Build GRCh38
rs768250854 Chr.12: 49460732-49460732 on Build GRCh38
rs1024213232 Chr.12: 49460837-49460837 on Build GRCh38
rs969610081 Chr.12: 49460881-49460881 on Build GRCh38
rs548618103 Chr.12: 49460600-49460600 on Build GRCh38
rs773058105 Chr.12: 49460750-49460751 on Build GRCh38
rs537269078 Chr.12: 49460691-49460691 on Build GRCh38
rs563672949 Chr.12: 49460584-49460584 on Build GRCh38
rs187575557 Chr.12: 49460674-49460674 on Build GRCh38
rs557273811 Chr.12: 49460701-49460701 on Build GRCh38
rs781325508 Chr.12: 49460593-49460593 on Build GRCh38
rs552669942 Chr.12: 49460725-49460725 on Build GRCh38
rs774050166 Chr.12: 49460742-49460742 on Build GRCh38
rs534726751 Chr.12: 49460633-49460633 on Build GRCh38
rs868325848 Chr.12: 49460861-49460861 on Build GRCh38
rs878938401 Chr.12: 49460804-49460804 on Build GRCh38
rs145184265 Chr.12: 49460548-49460548 on Build GRCh38
rs774795528 Chr.12: 49460622-49460622 on Build GRCh38
rs4590957 Chr.12: 49460887-49460887 on Build GRCh38
rs182998612 Chr.12: 49460539-49460539 on Build GRCh38
rs993866784 Chr.12: 49460738-49460738 on Build GRCh38
rs528850098 Chr.12: 49460595-49460595 on Build GRCh38
rs138121732 Chr.12: 49460619-49460619 on Build GRCh38
rs557848547 Chr.12: 49460648-49460648 on Build GRCh38
rs757084104 Chr.12: 49460572-49460572 on Build GRCh38
rs536446811 Chr.12: 49460524-49460524 on Build GRCh38
rs772891635 Chr.12: 49460532-49460532 on Build GRCh38
rs142418644 Chr.12: 49460654-49460654 on Build GRCh38

View Full Product Details